Search
Search

| Assay Name: | hsa-miR-153 |
|---|---|
| miRBase Accession Number: | MI0010335 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | UUGCAUAGUCACAAAAGUGA |
| Species: | Chicken, Chrysemys picta, Dog, Gorilla gorilla, Macaca nemestrina, Pig, Rhesus monkey, Xenopus tropicalis |
| Product Type: | TaqMan™ MicroRNA Assay |
Gene Family ID: | MIPF0000050, mir-153 |
Mature miRNA Sequence: | UUGCAUAGUCACAAAAGUGA |
| Species | miRBase ID | miRBase Accession Number | |
|---|---|---|---|
| Chicken | gga-miR-153-3p | MIMAT0001119 | |
| Chrysemys picta | cpi-miR-153-3p | MIMAT0037788 | |
| Dog | cfa-miR-153 | MIMAT0009840 | |
| Gorilla gorilla | ggo-miR-153 | MIMAT0002274 | |
| Macaca nemestrina | mne-miR-153 | MIMAT0002273 | |
| Pig | ssc-miR-153 | MIMAT0002160 | |
| View More Rows | |||
| Rhesus monkey | mml-miR-153-3p | MIMAT0002271 | |
| Xenopus tropicalis | xtr-miR-153 | MIMAT0003607 | |