Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › PM11293
          Assay Name: mmu-miR-497a-5p
          miRBase Accession Number: MI0004636
          miRBase Version: v22.1
          Chromosome Location: Chr.11: 70234717 - 70234800 [+] on Build GRCm38
          Mature miRNA Sequence: CAGCAGCACACUGUGGUUUGUA
          Species: Mouse, Rat, Bovine, Cricetulus griseus, Guinea pig, Microcebus murinus, Pan troglodytes, Rabbit, Tupaia chinensis
          Product Type: Ambion® Pre-miR™ miRNA Precursor
          Assay ID PM11293
          Size
          Availability Made To Order
          Catalog # AM17100
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Family ID:

          MIPF0000231, mir-497

          Mature miRNA Sequence:

          CAGCAGCACACUGUGGUUUGUA

          miRBase Details:



          Species miRBase ID miRBase Accession Number
          Mouse mmu-miR-497a-5p MIMAT0003453
          Rat rno-miR-497-5p MIMAT0003383
          Bovine bta-miR-497 MIMAT0004343
          Cricetulus griseus cgr-miR-497-5p MIMAT0023970
          Guinea pig cpo-miR-497-5p MIMAT0047295
          Microcebus murinus mmr-miR-497 MIMAT0049173
          View More Rows
          Pan troglodytes ptr-miR-497 MIMAT0012803
          Rabbit ocu-miR-497-5p MIMAT0048487
          Tupaia chinensis tch-miR-497-5p MIMAT0036558

          Stem-loop Details



          Mouse

          Stem-loop ID mmu-mir-497a
          Stem-loop Accession # MI0004636
          Stem-loop Sequence
          CCUGCCCCCGCCCCAGCAGCACACUGUGGUUUGUACGGCACUGUGGCCACGUCCAAACCACACUGUGGUGUUAGAGCGAGGGUA
          Chromosome Location Chr. 11 - 70234717 - 70234800 [+] on Build GRCm38
          Mature MicroRNA miRBase Accession #: MIMAT0003453
          miRBase ID: mmu-miR-497a-5p
          Mature miRNA Sequence: CAGCAGCACACUGUGGUUUGUA
          Chromosome Location: Chr. 11 - 70234717 - 70234800 [+] on Build GRCm38

          Rat

          Stem-loop ID rno-mir-497
          Stem-loop Accession # MI0003724
          Stem-loop Sequence
          CCAGCAGCACACUGUGGUUUGUACGGCACUGUGGCCACGUCCAAACCACACUGUGGUGUUAGAGCGAGG
          Chromosome Location Chr. 10 - 56844977 - 56845045 [+] on Build Rnor_6.0
          Mature MicroRNA miRBase Accession #: MIMAT0003383
          miRBase ID: rno-miR-497-5p
          Mature miRNA Sequence: CAGCAGCACACUGUGGUUUGUA
          Chromosome Location: Chr. 10 - 56844977 - 56845045 [+] on Build Rnor_6.0

          Bovine

          Stem-loop ID bta-mir-497
          Stem-loop Accession # MI0005467
          Stem-loop Sequence
          CCACCCCAGUCCUGCUCCCGCCCCAGCAGCACACUGUGGUUUGUACGGCACUGUGGCCACGUCCAAACCACACUGUGGUGUUAGAGCGAGGGUGGGGGAGGCACCGCUGAGG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0004343
          miRBase ID: bta-miR-497
          Mature miRNA Sequence: CAGCAGCACACUGUGGUUUGUA
          Chromosome Location: NA

          Cricetulus griseus

          Stem-loop ID cgr-mir-497
          Stem-loop Accession # MI0020532
          Stem-loop Sequence
          CUCCCGCCCCAGCAGCACACUGUGGUUUGUACGGCACUGUGGCCACGUCCAAACCACACUGUGGUGUUAGAGCGAGGGUGG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0023970
          miRBase ID: cgr-miR-497-5p
          Mature miRNA Sequence: CAGCAGCACACUGUGGUUUGUA
          Chromosome Location: NA

          Guinea pig

          Stem-loop ID cpo-mir-497
          Stem-loop Accession # MI0038797
          Stem-loop Sequence
          CAGCAGCACACUGUGGUUUGUACGGCACUGUGGCCACGUCCAAACCACACUGUGGUGUUAGAGCGAGGGUGGG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0047295
          miRBase ID: cpo-miR-497-5p
          Mature miRNA Sequence: CAGCAGCACACUGUGGUUUGUA
          Chromosome Location: NA

          Microcebus murinus

          Stem-loop ID mmr-mir-497
          Stem-loop Accession # MI0039891
          Stem-loop Sequence
          CAGCAGCACACUGUGGUUUGUACGGCACUGUGGCCACGUCCAAACCACACUGUGGUGUUAGAGCGAG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0049173
          miRBase ID: mmr-miR-497
          Mature miRNA Sequence: CAGCAGCACACUGUGGUUUGUA
          Chromosome Location: NA

          Pan troglodytes

          Stem-loop ID ptr-mir-497
          Stem-loop Accession # MI0012565
          Stem-loop Sequence
          CCAGCAGCACACUGUGGUUUGUACGGCACUGUGGCCACGUCCAAACCACACUGUGGUGUUAGAGCGAGG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0012803
          miRBase ID: ptr-miR-497
          Mature miRNA Sequence: CAGCAGCACACUGUGGUUUGUA
          Chromosome Location: NA

          Rabbit

          Stem-loop ID ocu-mir-497
          Stem-loop Accession # MI0039426
          Stem-loop Sequence
          CAGCAGCACACUGUGGUUUGUACGGCACUGUGGCCACGUCCAAACCACACUGUGGUGUUAGA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0048487
          miRBase ID: ocu-miR-497-5p
          Mature miRNA Sequence: CAGCAGCACACUGUGGUUUGUA
          Chromosome Location: NA

          Tupaia chinensis

          Stem-loop ID tch-mir-497
          Stem-loop Accession # MI0031218
          Stem-loop Sequence
          CAGCAGCACACUGUGGUUUGUACGGCACUGUGGCCACGUCCAAACCACACUGUGGUGUUAGA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0036558
          miRBase ID: tch-miR-497-5p
          Mature miRNA Sequence: CAGCAGCACACUGUGGUUUGUA
          Chromosome Location: NA


          Back To Top

          More Information




          Related Products

          TaqMan™ Pri-miRNA Assay : Rn03465143_pri
          TaqMan™ Pri-miRNA Assay : Mm03307967_pri
          Ambion® Anti-miR™ miRNA Inhibitor : AM11293
          TaqMan™ MicroRNA Assay : 001346
          mirVana® miRNA inhibitor : MH11293
          mirVana® miRNA mimic : MC11293
          TaqMan™ Advanced miRNA Assay : rno481298_mir
          TaqMan™ Advanced miRNA Assay : mmu482607_mir


          miRBase Alias:

          Mouse: mmu-miR-497-5p (v20)
          Mouse: mmu-miR-497 (v17)
          Rat: rno-miR-497 (v18)

          Back To Top

          Related Products

          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-x75fr:80/100.66.77.4:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline