Search
Search

| Assay Name: | mmu-miR-497a-5p |
|---|---|
| miRBase Accession Number: | MI0004636 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | CAGCAGCACACUGUGGUUUGUA |
| Species: | Mouse, Rat, Bovine, Cricetulus griseus, Guinea pig, Microcebus murinus, Pan troglodytes, Rabbit, Tupaia chinensis |
| Product Type: | mirVana® miRNA inhibitor |
Gene Family ID: | MIPF0000231, mir-497 |
Mature miRNA Sequence: | CAGCAGCACACUGUGGUUUGUA |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Mouse | mmu-miR-497a-5p | MIMAT0003453 |
| Rat | rno-miR-497-5p | MIMAT0003383 |
| Bovine | bta-miR-497 | MIMAT0004343 |
| Cricetulus griseus | cgr-miR-497-5p | MIMAT0023970 |
| Guinea pig | cpo-miR-497-5p | MIMAT0047295 |
| Microcebus murinus | mmr-miR-497 | MIMAT0049173 |
| View More Rows | ||
| Pan troglodytes | ptr-miR-497 | MIMAT0012803 |
| Rabbit | ocu-miR-497-5p | MIMAT0048487 |
| Tupaia chinensis | tch-miR-497-5p | MIMAT0036558 |