Search
Search

| Assay Name: | mmu-miR-497 |
|---|---|
| miRBase Accession Number: | MI0004636 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | CAGCAGCACACUGUGGUUUGUA |
| Species: | Mouse, Rat, Bovine, Cricetulus griseus, Guinea pig, Microcebus murinus, Pan troglodytes, Rabbit, Tupaia chinensis |
| Product Type: | TaqMan™ MicroRNA Assay |
Gene Family ID: | MIPF0000231, mir-497 |
Mature miRNA Sequence: | CAGCAGCACACUGUGGUUUGUA |
| Species | miRBase ID | miRBase Accession Number | |
|---|---|---|---|
| Mouse | mmu-miR-497a-5p | MIMAT0003453 | |
| Rat | rno-miR-497-5p | MIMAT0003383 | |
| Bovine | bta-miR-497 | MIMAT0004343 | |
| Cricetulus griseus | cgr-miR-497-5p | MIMAT0023970 | |
| Guinea pig | cpo-miR-497-5p | MIMAT0047295 | |
| Microcebus murinus | mmr-miR-497 | MIMAT0049173 | |
| View More Rows | |||
| Pan troglodytes | ptr-miR-497 | MIMAT0012803 | |
| Rabbit | ocu-miR-497-5p | MIMAT0048487 | |
| Tupaia chinensis | tch-miR-497-5p | MIMAT0036558 | |