Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Food and Beverage
    • Lab Solutions
    • Pharma and Biopharma
    • Real-Time PCR
    • Semiconductor Analysis
    • Clinical and Diagnostics
    • Digital Solutions
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • See all services
  • Help and Support
    • Order Help
    • Digital Solutions
    • Product Support
    • Technical Information
    • Training and Education
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm03307967_pri
          Assay Name:
          mmu-mir-497b
          Stem-loop ID:
          mmu-mir-497a mmu-mir-497b
          Mapped to miRBase Version:
          v22.1
          Chromosome Location:
          Chr.11: 70234717 - 70234800 [+] on Build GRCm38
          Stem-loop Location:
          Chr.11: 70234717 - 70234800 [+] on Build GRCm38 Chr.11: 70234692 - 70234816 [-] on Build GRCm38
          Mature miRNA ID:
          mmu-miR-497a-5p mmu-miR-497a-3p mmu-miR-497b
          Species:
          Mouse
          Product Type:
          TaqMan™ Pri-miRNA Assay
          Assay ID Mm03307967_pri
          Size
          Availability Made To Order
          Catalog # 4427012
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Amplicon Length:

          66

          Chromosome Location:

          Chr. 11 - 70234519 - 70234519 [+] on 70234519 Build GRCm38

          Species:

          Mouse

          miRBase Details:



          Species miRBase ID miRBase Accession Number
          Mouse mmu-miR-497a-5p MIMAT0003453
          Mouse mmu-miR-497a-3p MIMAT0017247
          Mouse mmu-miR-497b MIMAT0031404

          Stem-loop Details



          Mouse

          Stem-loop ID mmu-mir-497a
          Stem-loop Accession # MI0004636
          Stem-loop Sequence
          CCUGCCCCCGCCCCAGCAGCACACUGUGGUUUGUACGGCACUGUGGCCACGUCCAAACCACACUGUGGUGUUAGAGCGAGGGUA
          Chromosome Location Chr. 11 - 70234717 - 70234800 [+] on Build GRCm38
          Mature MicroRNA miRBase Accession #: MIMAT0003453
          miRBase ID: mmu-miR-497a-5p
          Mature miRNA Sequence: CAGCAGCACACUGUGGUUUGUA
          Chromosome Location: Chr. 11 - 70234717 - 70234800 [+] on Build GRCm38

          miRBase Accession #: MIMAT0017247
          miRBase ID: mmu-miR-497a-3p
          Mature miRNA Sequence: CAAACCACACUGUGGUGUUAG
          Chromosome Location: Chr. 11 - 70234717 - 70234800 [+] on Build GRCm38

          Gene Family ID MIPF0000231, mir-497
          Distance to Amplicon 166 bases
          Stem-loop ID mmu-mir-497b
          Stem-loop Accession # MI0026030
          Stem-loop Sequence
          UCAUCGGUGCCUCCCAUACCCUCGCUCUAACACCACAGUGUGGUUUGGACGUGGCCACAGUGCCGUACAAACCACAGUGUGCUGCUGGGGCGGGGGCAGGACUGGGGUGGGUCGAGAGGAAGACA
          Chromosome Location Chr. 11 - 70234692 - 70234816 [-] on Build GRCm38
          Mature MicroRNA miRBase Accession #: MIMAT0031404
          miRBase ID: mmu-miR-497b
          Mature miRNA Sequence: CACCACAGUGUGGUUUGGACGUGG
          Chromosome Location: Chr. 11 - 70234692 - 70234816 [-] on Build GRCm38

          Gene Family ID MIPF0000231, mir-497
          Distance to Amplicon 141 bases

          Back To Top

          More Information




          Related Products

          Ambion® Anti-miR™ miRNA Inhibitor : AM11293
          Ambion® Pre-miR™ miRNA Precursor : PM11293
          TaqMan™ MicroRNA Assay : 001346
          mirVana® miRNA inhibitor : MH11293
          mirVana® miRNA mimic : MC11293
          TaqMan™ Advanced miRNA Assay : rno481298_mir
          TaqMan™ Advanced miRNA Assay : mmu482607_mir
          Ambion® Anti-miR™ miRNA Inhibitor : AM28334
          Ambion® Pre-miR™ miRNA Precursor : PM28334
          TaqMan™ MicroRNA Assay : 466309_mat
          mirVana® miRNA inhibitor : MH28334
          mirVana® miRNA mimic : MC28334
          TaqMan™ Advanced miRNA Assay : mmu481864_mir
          Ambion® Anti-miR™ miRNA Inhibitor : AM20230
          Ambion® Pre-miR™ miRNA Precursor : PM20230
          TaqMan™ MicroRNA Assay : 464318_mat
          mirVana® miRNA inhibitor : MH20230
          mirVana® miRNA mimic : MC20230
          TaqMan™ Advanced miRNA Assay : mmu478946_mir


          miRBase Alias:

          Mouse: mmu-mir-497a (v21)

          Back To Top

          Related Products

          • High Capacity RNA-to-cDNA Kit
          • SuperScript® IV VILO™ Master Mix
          • TaqMan® Universal PCR Master Mix
          • TaqMan® Fast Advanced Master Mix
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-vzwvz:80/100.66.79.173:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline