Search
Search

| Assay Name: | hsa-miR-145-3p |
|---|---|
| miRBase Accession Number: | MI0000461 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | GGAUUCCUGGAAAUACUGUUCU |
| Species: | Human, Artibeus jamaicensis, Chrysemys picta, Gadus morhua, Ophiophagus hannah, Pig, Python bivittatus, Xenopus laevis, Zebrafish |
| Product Type: | Ambion® Pre-miR™ miRNA Precursor |
Gene Family ID: | MIPF0000079, mir-145 |
Mature miRNA Sequence: | GGAUUCCUGGAAAUACUGUUCU |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-145-3p | MIMAT0004601 |
| Artibeus jamaicensis | aja-miR-145 | MIMAT0031138 |
| Chrysemys picta | cpi-miR-145-3p | MIMAT0037773 |
| Gadus morhua | gmo-miR-145-3p | MIMAT0044335 |
| Ophiophagus hannah | oha-miR-145-3p | MIMAT0036728 |
| Pig | ssc-miR-145-3p | MIMAT0022919 |
| View More Rows | ||
| Python bivittatus | pbv-miR-145-3p | MIMAT0038960 |
| Xenopus laevis | xla-miR-145-3p | MIMAT0046461 |
| Zebrafish | dre-miR-145-3p | MIMAT0031976 |