Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › 477915_mir

          Specificity testing was performed using human anti-targets.

          Assay Name: hsa-miR-145-3p
          Stem-loop Accession Number: MI0000461
          miRBase Version: v22.1
          Chromosome Location: Chr.5: 149430646 - 149430733 [+] on Build GRCh38
          Mature miRNA Sequence: GGAUUCCUGGAAAUACUGUUCU
          Species: Human, Artibeus jamaicensis, Chrysemys picta, Gadus morhua, Ophiophagus hannah, Pig, Python bivittatus, Xenopus laevis, Zebrafish
          Product Type: TaqMan™ Advanced miRNA Assay
          Assay ID 477915_mir
          Size S: 250 rxns
          Availability Inventoried
          Catalog # A25576
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Family ID:

          MIPF0000079, mir-145

          Mature miRNA Sequence:

          GGAUUCCUGGAAAUACUGUUCU

          miRBase Details:



          Species miRBase ID miRBase Accession Number
          Human hsa-miR-145-3p MIMAT0004601
          Artibeus jamaicensis aja-miR-145 MIMAT0031138
          Chrysemys picta cpi-miR-145-3p MIMAT0037773
          Gadus morhua gmo-miR-145-3p MIMAT0044335
          Ophiophagus hannah oha-miR-145-3p MIMAT0036728
          Pig ssc-miR-145-3p MIMAT0022919
          View More Rows
          Python bivittatus pbv-miR-145-3p MIMAT0038960
          Xenopus laevis xla-miR-145-3p MIMAT0046461
          Zebrafish dre-miR-145-3p MIMAT0031976

          Stem-loop Details



          Human

          Stem-loop ID hsa-mir-145
          Stem-loop Accession # MI0000461
          Stem-loop Sequence
          CACCUUGUCCUCACGGUCCAGUUUUCCCAGGAAUCCCUUAGAUGCUAAGAUGGGGAUUCCUGGAAAUACUGUUCUUGAGGUCAUGGUU
          Chromosome Location Chr. 5 - 149430646 - 149430733 [+] on Build GRCh38
          Mature MicroRNA miRBase Accession #: MIMAT0004601
          miRBase ID: hsa-miR-145-3p
          Mature miRNA Sequence: GGAUUCCUGGAAAUACUGUUCU
          Chromosome Location: Chr. 5 - 149430646 - 149430733 [+] on Build GRCh38

          Artibeus jamaicensis

          Stem-loop ID aja-mir-145
          Stem-loop Accession # MI0024360
          Stem-loop Sequence
          CACCUUGUCCUCACGGUCCAGUUUUCCCAGGAAUCCCUUAGAUGCUAAGAUGGGGAUUCCUGGAAAUACUGUUCUUGAGGUCAUGGUU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0031138
          miRBase ID: aja-miR-145
          Mature miRNA Sequence: GGAUUCCUGGAAAUACUGUUCU
          Chromosome Location: NA

          Chrysemys picta

          Stem-loop ID cpi-mir-145
          Stem-loop Accession # MI0029488
          Stem-loop Sequence
          GUGUCCUCAGGGUCCAGUUUUCCCAGGAAUCCCUUAGGCACUAUGACGGGGAUUCCUGGAAAUACUGUUCUUGAGGCCAUGGCUCAGCAGCU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0037773
          miRBase ID: cpi-miR-145-3p
          Mature miRNA Sequence: GGAUUCCUGGAAAUACUGUUCU
          Chromosome Location: NA

          Gadus morhua

          Stem-loop ID gmo-mir-145
          Stem-loop Accession # MI0036228
          Stem-loop Sequence
          GUCCAGUUUUCCCAGGAAUCCCUUGUUGAACAUGAAAGCGGGAUUCCUGGAAAUACUGUUCU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0044335
          miRBase ID: gmo-miR-145-3p
          Mature miRNA Sequence: GGAUUCCUGGAAAUACUGUUCU
          Chromosome Location: NA

          Ophiophagus hannah

          Stem-loop ID oha-mir-145
          Stem-loop Accession # MI0031358
          Stem-loop Sequence
          UCGGGGUCCAGUUUUCCCAGGAAUCCCUUAGAAGAUAAGAAGGGGAUUCCUGGAAAUACUGUUCUUGGGGUC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0036728
          miRBase ID: oha-miR-145-3p
          Mature miRNA Sequence: GGAUUCCUGGAAAUACUGUUCU
          Chromosome Location: NA

          Pig

          Stem-loop ID ssc-mir-145
          Stem-loop Accession # MI0002417
          Stem-loop Sequence
          CACCUUGUCCUCACGGUCCAGUUUUCCCAGGAAUCCCUUAGAUGCUGAGAUGGGGAUUCCUGGAAAUACUGUUCUUGAGGUCAUGG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0022919
          miRBase ID: ssc-miR-145-3p
          Mature miRNA Sequence: GGAUUCCUGGAAAUACUGUUCU
          Chromosome Location: NA

          Python bivittatus

          Stem-loop ID pbv-mir-145
          Stem-loop Accession # MI0030243
          Stem-loop Sequence
          ACUAUGUCCUCGGGGUCCAGUUUUCCCAGGAAUCCCUUAGAUGCUAAGAAGGGGAUUCCUGGAAAUACUGUUCUUGGGGUCAUGGCUC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0038960
          miRBase ID: pbv-miR-145-3p
          Mature miRNA Sequence: GGAUUCCUGGAAAUACUGUUCU
          Chromosome Location: NA

          Xenopus laevis

          Stem-loop ID xla-mir-145
          Stem-loop Accession # MI0038256
          Stem-loop Sequence
          CAGUUUUCCCAGGAAUCCCUUGGGUGCUGUGGUGGGGAUUCCUGGAAAUACUGUUCU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0046461
          miRBase ID: xla-miR-145-3p
          Mature miRNA Sequence: GGAUUCCUGGAAAUACUGUUCU
          Chromosome Location: NA

          Zebrafish

          Stem-loop ID dre-mir-145
          Stem-loop Accession # MI0002010
          Stem-loop Sequence
          UCAGUCUUCAUCAUUUCCUCAUCCCCGGGGUCCAGUUUUCCCAGGAAUCCCUUGGGCAAUCGAAAGGGGGAUUCCUGGAAAUACUGUUCUUGGGGUUGGGGCUGGACUACUGA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0031976
          miRBase ID: dre-miR-145-3p
          Mature miRNA Sequence: GGAUUCCUGGAAAUACUGUUCU
          Chromosome Location: NA


          Back To Top

          More Information




          Related Products

          TaqMan™ Pri-miRNA Assay : Hs03303169_pri
          Ambion® Anti-miR™ miRNA Inhibitor : AM13036
          Ambion® Pre-miR™ miRNA Precursor : PM13036
          TaqMan™ MicroRNA Assay : 002149
          mirVana® miRNA inhibitor : MH13036
          mirVana® miRNA mimic : MC13036
          FGMIR : TaqManTM Advanced miRNA Human B 96-well Plate 2
          MIRNA : TaqMan® Advanced miRNA Human B Card
          MIRNA : TaqMan® OpenArray® Human Advanced MicroRNA Panel
          MIRNA : TaqMan® Advanced miRNA Human A and B Cards
          FGMIR : TaqManTM Advanced miRNA Human B Card
          MIRNA : TaqMan® Advanced miRNA Human B 96-well Plates, Standard
          FGMIR : QS TaqmanTM OpenArray Human Advanced miRNA Panel
          MIRNA : TaqMan® Advanced miRNA Human A and B 96-well Plates, Standard
          FGMIR : TaqManTM Advanced miRNA Human B 96-well Plate 2

          Back To Top

          Related Products

          • TaqMan® Advanced miRNA cDNA Synthesis Kit
          • TaqMan® Fast Advanced Master Mix
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-x75fr:80/100.66.77.4:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline