Search
Search

| Assay Name: | hsa-miR-145* |
|---|---|
| miRBase Accession Number: | MI0000461 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | GGAUUCCUGGAAAUACUGUUCU |
| Species: | Human, Artibeus jamaicensis, Chrysemys picta, Gadus morhua, Ophiophagus hannah, Pig, Python bivittatus, Xenopus laevis, Zebrafish |
| Product Type: | TaqMan™ MicroRNA Assay |
Gene Family ID: | MIPF0000079, mir-145 |
Mature miRNA Sequence: | GGAUUCCUGGAAAUACUGUUCU |
| Species | miRBase ID | miRBase Accession Number | |
|---|---|---|---|
| Human | hsa-miR-145-3p | MIMAT0004601 | |
| Artibeus jamaicensis | aja-miR-145 | MIMAT0031138 | |
| Chrysemys picta | cpi-miR-145-3p | MIMAT0037773 | |
| Gadus morhua | gmo-miR-145-3p | MIMAT0044335 | |
| Ophiophagus hannah | oha-miR-145-3p | MIMAT0036728 | |
| Pig | ssc-miR-145-3p | MIMAT0022919 | |
| View More Rows | |||
| Python bivittatus | pbv-miR-145-3p | MIMAT0038960 | |
| Xenopus laevis | xla-miR-145-3p | MIMAT0046461 | |
| Zebrafish | dre-miR-145-3p | MIMAT0031976 | |