Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Instrument Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • Our Instagram
      Our Instagram
    • Our Facebook
      Our Facebook
    • Blog Behind the Bench
      Blog Behind the Bench
    • Customer Experience Center (CEC)
      Customer Experience Center (CEC)
    • Ecommerce Exclusives
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › MC13036
          Assay Name: hsa-miR-145-3p
          miRBase Accession Number: MI0000461
          miRBase Version: v22.1
          Chromosome Location: Chr.5: 149430646 - 149430733 [+] on Build GRCh38
          Mature miRNA Sequence: GGAUUCCUGGAAAUACUGUUCU
          Species: Human, Artibeus jamaicensis, Chrysemys picta, Gadus morhua, Ophiophagus hannah, Pig, Python bivittatus, Xenopus laevis, Zebrafish
          Product Type: mirVana® miRNA mimic
          Assay ID MC13036

          Purification
          Size
          Availability Made To Order
          Catalog # 4464066
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Family ID:

          MIPF0000079, mir-145

          Mature miRNA Sequence:

          GGAUUCCUGGAAAUACUGUUCU

          miRBase Details:



          Species miRBase ID miRBase Accession Number
          Human hsa-miR-145-3p MIMAT0004601
          Artibeus jamaicensis aja-miR-145 MIMAT0031138
          Chrysemys picta cpi-miR-145-3p MIMAT0037773
          Gadus morhua gmo-miR-145-3p MIMAT0044335
          Ophiophagus hannah oha-miR-145-3p MIMAT0036728
          Pig ssc-miR-145-3p MIMAT0022919
          View More Rows
          Python bivittatus pbv-miR-145-3p MIMAT0038960
          Xenopus laevis xla-miR-145-3p MIMAT0046461
          Zebrafish dre-miR-145-3p MIMAT0031976

          Stem-loop Details



          Human

          Stem-loop ID hsa-mir-145
          Stem-loop Accession # MI0000461
          Stem-loop Sequence
          CACCUUGUCCUCACGGUCCAGUUUUCCCAGGAAUCCCUUAGAUGCUAAGAUGGGGAUUCCUGGAAAUACUGUUCUUGAGGUCAUGGUU
          Chromosome Location Chr. 5 - 149430646 - 149430733 [+] on Build GRCh38
          Mature MicroRNA miRBase Accession #: MIMAT0004601
          miRBase ID: hsa-miR-145-3p
          Mature miRNA Sequence: GGAUUCCUGGAAAUACUGUUCU
          Chromosome Location: Chr. 5 - 149430646 - 149430733 [+] on Build GRCh38

          Artibeus jamaicensis

          Stem-loop ID aja-mir-145
          Stem-loop Accession # MI0024360
          Stem-loop Sequence
          CACCUUGUCCUCACGGUCCAGUUUUCCCAGGAAUCCCUUAGAUGCUAAGAUGGGGAUUCCUGGAAAUACUGUUCUUGAGGUCAUGGUU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0031138
          miRBase ID: aja-miR-145
          Mature miRNA Sequence: GGAUUCCUGGAAAUACUGUUCU
          Chromosome Location: NA

          Chrysemys picta

          Stem-loop ID cpi-mir-145
          Stem-loop Accession # MI0029488
          Stem-loop Sequence
          GUGUCCUCAGGGUCCAGUUUUCCCAGGAAUCCCUUAGGCACUAUGACGGGGAUUCCUGGAAAUACUGUUCUUGAGGCCAUGGCUCAGCAGCU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0037773
          miRBase ID: cpi-miR-145-3p
          Mature miRNA Sequence: GGAUUCCUGGAAAUACUGUUCU
          Chromosome Location: NA

          Gadus morhua

          Stem-loop ID gmo-mir-145
          Stem-loop Accession # MI0036228
          Stem-loop Sequence
          GUCCAGUUUUCCCAGGAAUCCCUUGUUGAACAUGAAAGCGGGAUUCCUGGAAAUACUGUUCU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0044335
          miRBase ID: gmo-miR-145-3p
          Mature miRNA Sequence: GGAUUCCUGGAAAUACUGUUCU
          Chromosome Location: NA

          Ophiophagus hannah

          Stem-loop ID oha-mir-145
          Stem-loop Accession # MI0031358
          Stem-loop Sequence
          UCGGGGUCCAGUUUUCCCAGGAAUCCCUUAGAAGAUAAGAAGGGGAUUCCUGGAAAUACUGUUCUUGGGGUC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0036728
          miRBase ID: oha-miR-145-3p
          Mature miRNA Sequence: GGAUUCCUGGAAAUACUGUUCU
          Chromosome Location: NA

          Pig

          Stem-loop ID ssc-mir-145
          Stem-loop Accession # MI0002417
          Stem-loop Sequence
          CACCUUGUCCUCACGGUCCAGUUUUCCCAGGAAUCCCUUAGAUGCUGAGAUGGGGAUUCCUGGAAAUACUGUUCUUGAGGUCAUGG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0022919
          miRBase ID: ssc-miR-145-3p
          Mature miRNA Sequence: GGAUUCCUGGAAAUACUGUUCU
          Chromosome Location: NA

          Python bivittatus

          Stem-loop ID pbv-mir-145
          Stem-loop Accession # MI0030243
          Stem-loop Sequence
          ACUAUGUCCUCGGGGUCCAGUUUUCCCAGGAAUCCCUUAGAUGCUAAGAAGGGGAUUCCUGGAAAUACUGUUCUUGGGGUCAUGGCUC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0038960
          miRBase ID: pbv-miR-145-3p
          Mature miRNA Sequence: GGAUUCCUGGAAAUACUGUUCU
          Chromosome Location: NA

          Xenopus laevis

          Stem-loop ID xla-mir-145
          Stem-loop Accession # MI0038256
          Stem-loop Sequence
          CAGUUUUCCCAGGAAUCCCUUGGGUGCUGUGGUGGGGAUUCCUGGAAAUACUGUUCU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0046461
          miRBase ID: xla-miR-145-3p
          Mature miRNA Sequence: GGAUUCCUGGAAAUACUGUUCU
          Chromosome Location: NA

          Zebrafish

          Stem-loop ID dre-mir-145
          Stem-loop Accession # MI0002010
          Stem-loop Sequence
          UCAGUCUUCAUCAUUUCCUCAUCCCCGGGGUCCAGUUUUCCCAGGAAUCCCUUGGGCAAUCGAAAGGGGGAUUCCUGGAAAUACUGUUCUUGGGGUUGGGGCUGGACUACUGA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0031976
          miRBase ID: dre-miR-145-3p
          Mature miRNA Sequence: GGAUUCCUGGAAAUACUGUUCU
          Chromosome Location: NA


          Back To Top

          More Information




          Related Products

          TaqMan™ Pri-miRNA Assay : Hs03303169_pri
          Ambion® Anti-miR™ miRNA Inhibitor : AM13036
          Ambion® Pre-miR™ miRNA Precursor : PM13036
          TaqMan™ MicroRNA Assay : 002149
          mirVana® miRNA inhibitor : MH13036
          TaqMan™ Advanced miRNA Assay : 477915_mir


          miRBase Alias:

          Human: hsa-miR-145* (v17)

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Find Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Brasil flag icon
          Brasil

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-vzwvz:80/100.66.79.173:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline