Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Food and Beverage
    • Lab Solutions
    • Pharma and Biopharma
    • Real-Time PCR
    • Semiconductor Analysis
    • Clinical and Diagnostics
    • Digital Solutions
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • See all services
  • Help and Support
    • Order Help
    • Digital Solutions
    • Product Support
    • Technical Information
    • Training and Education
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › mmu481661_mir

          Specificity testing was performed using mouse anti-targets.

          Assay Name: mmu-miR-26a-1-3p
          Stem-loop Accession Number: MI0000083
          miRBase Version: v22.1
          Chromosome Location: Chr.3: 37969404 - 37969480 [+] on Build GRCh38
          Mature miRNA Sequence: CCUAUUCUUGGUUACUUGCACG
          Species: Human, Mouse, Cricetulus griseus, Guinea pig, Pteropus alecto, Rhesus monkey
          Product Type: TaqMan™ Advanced miRNA Assay
          Assay ID mmu481661_mir
          Size S: 250 rxns
          Availability Inventoried
          Catalog # A25576
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Family ID:

          MIPF0000043, mir-26

          Mature miRNA Sequence:

          CCUAUUCUUGGUUACUUGCACG

          miRBase Details:



          Species miRBase ID miRBase Accession Number
          Human hsa-miR-26a-1-3p MIMAT0004499
          Mouse mmu-miR-26a-1-3p MIMAT0017020
          Cricetulus griseus cgr-miR-26a MIMAT0041137
          Guinea pig cpo-miR-26a-3p MIMAT0046886
          Pteropus alecto pal-miR-26a-3p MIMAT0040072
          Rhesus monkey mml-miR-26a-1-3p MIMAT0026574

          Stem-loop Details



          Human

          Stem-loop ID hsa-mir-26a-1
          Stem-loop Accession # MI0000083
          Stem-loop Sequence
          GUGGCCUCGUUCAAGUAAUCCAGGAUAGGCUGUGCAGGUCCCAAUGGGCCUAUUCUUGGUUACUUGCACGGGGACGC
          Chromosome Location Chr. 3 - 37969404 - 37969480 [+] on Build GRCh38
          Mature MicroRNA miRBase Accession #: MIMAT0004499
          miRBase ID: hsa-miR-26a-1-3p
          Mature miRNA Sequence: CCUAUUCUUGGUUACUUGCACG
          Chromosome Location: Chr. 3 - 37969404 - 37969480 [+] on Build GRCh38

          Mouse

          Stem-loop ID mmu-mir-26a-1
          Stem-loop Accession # MI0000573
          Stem-loop Sequence
          AAGGCCGUGGCCUCGUUCAAGUAAUCCAGGAUAGGCUGUGCAGGUCCCAAGGGGCCUAUUCUUGGUUACUUGCACGGGGACGCGGGCCUG
          Chromosome Location Chr. 9 - 119031796 - 119031885 [+] on Build GRCm38
          Mature MicroRNA miRBase Accession #: MIMAT0017020
          miRBase ID: mmu-miR-26a-1-3p
          Mature miRNA Sequence: CCUAUUCUUGGUUACUUGCACG
          Chromosome Location: Chr. 9 - 119031796 - 119031885 [+] on Build GRCm38

          Cricetulus griseus

          Stem-loop ID cgr-mir-26a-1
          Stem-loop Accession # MI0033065
          Stem-loop Sequence
          GUGGCCUCGUUCAAGUAAUCCAGGAUAGGCUGUGCAGGUCCCAAGGGGCCUAUUCUUGGUUACUUGCACGGGG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0041137
          miRBase ID: cgr-miR-26a
          Mature miRNA Sequence: CCUAUUCUUGGUUACUUGCACG
          Chromosome Location: NA

          Guinea pig

          Stem-loop ID cpo-mir-26a-1
          Stem-loop Accession # MI0038580
          Stem-loop Sequence
          UUCAAGUAAUCCAGGAUAGGCUGUGCAGGUCCCAACGGGCCUAUUCUUGGUUACUUGCACG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0046886
          miRBase ID: cpo-miR-26a-3p
          Mature miRNA Sequence: CCUAUUCUUGGUUACUUGCACG
          Chromosome Location: NA

          Pteropus alecto

          Stem-loop ID pal-mir-26a-1
          Stem-loop Accession # MI0032479
          Stem-loop Sequence
          UUCAAGUAAUCCAGGAUAGGCUGUGCAGGUCCCAAUGGGCCUAUUCUUGGUUACUUGCACG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0040072
          miRBase ID: pal-miR-26a-3p
          Mature miRNA Sequence: CCUAUUCUUGGUUACUUGCACG
          Chromosome Location: NA

          Rhesus monkey

          Stem-loop ID mml-mir-26a-1
          Stem-loop Accession # MI0002646
          Stem-loop Sequence
          GUGGCCUCGUUCAAGUAAUCCAGGAUAGGCUGUGCAGGUCCCAAUGGGCCUAUUCUUGGUUACUUGCACGGGGACGC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0026574
          miRBase ID: mml-miR-26a-1-3p
          Mature miRNA Sequence: CCUAUUCUUGGUUACUUGCACG
          Chromosome Location: NA


          Back To Top

          More Information




          Related Products

          TaqMan™ Pri-miRNA Assay : Hs03293444_pri
          TaqMan™ Pri-miRNA Assay : Mm03306848_pri
          Ambion® Anti-miR™ miRNA Inhibitor : AM12758
          Ambion® Pre-miR™ miRNA Precursor : PM12758
          TaqMan™ MicroRNA Assay : 002443
          mirVana® miRNA inhibitor : MH12758
          mirVana® miRNA mimic : MC12758
          TaqMan™ Advanced miRNA Assay : 478787_mir

          Back To Top

          Related Products

          • TaqMan® Advanced miRNA cDNA Synthesis Kit
          • TaqMan® Fast Advanced Master Mix
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-b2k9d:80/100.66.79.173:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline