Search
Search

| Assay Name: | hsa-miR-26a-1-3p |
|---|---|
| Stem-loop Accession Number: | MI0000083 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | CCUAUUCUUGGUUACUUGCACG |
| Species: | Human, Mouse, Cricetulus griseus, Guinea pig, Pteropus alecto, Rhesus monkey |
| Product Type: | TaqMan™ Advanced miRNA Assay |
Gene Family ID: | MIPF0000043, mir-26 |
Mature miRNA Sequence: | CCUAUUCUUGGUUACUUGCACG |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-26a-1-3p | MIMAT0004499 |
| Mouse | mmu-miR-26a-1-3p | MIMAT0017020 |
| Cricetulus griseus | cgr-miR-26a | MIMAT0041137 |
| Guinea pig | cpo-miR-26a-3p | MIMAT0046886 |
| Pteropus alecto | pal-miR-26a-3p | MIMAT0040072 |
| Rhesus monkey | mml-miR-26a-1-3p | MIMAT0026574 |