Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Pipettes and Pipette Tips
Lab Centrifuges
Ultra-Low Temperature Freezers
Spectroscopy
Beakers
PCR Equipment and Supplies
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
See all product categories
Applications
Cell Analysis
Lab Equipment
Real-Time PCR
PCR
Chromatography
Cell Culture and Transfection
DNA and RNA Extraction and Analysis
Protein Biology
Flow Cytometry
Chemicals
See all applications and techniques
Services
Custom Services
Lab Informatics
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Instrument Services
Training Services
Unity Lab Services
See all services
Help and Support
Customer Center
Contact Us
Certificates of Analysis and Conformance
Safety Data Sheets (SDS)
Manuals
How to Cite Our Products in a Paper
Instrument Support
Knowledge Base and Product FAQs
Learning Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
Home
›
Search Tool
›
Search Results
› Mm03306848_pri
Assay Name:
mmu-mir-26a-1
Stem-loop ID:
mmu-mir-26a-1
Mapped to miRBase Version:
v22.1
Chromosome Location:
Chr.9: 119031796 - 119031885 [+] on Build GRCm38
Stem-loop Location:
Chr.9: 119031796 - 119031885 [+] on Build GRCm38
Mature miRNA ID:
mmu-miR-26a-1-3p
mmu-miR-26a-5p
Species:
Mouse
Product Type:
TaqMan
™
Pri-miRNA Assay
Assay ID
Mm03306848_pri
Size
S: 360 rxns; 20X
M: 750 rxns; 20X
L: 2900 rxns; 60X
Availability
Made To Order
Catalog #
4427012
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
Assay Details
More Information
Genomic Map
LOADING...
×
Back To Top
Assay Details
Amplicon Length:
73
Chromosome Location:
Chr. 9 - 119032098 - 119032098 [+] on 119032098 Build GRCm38
Species:
Mouse
miRBase Details:
Species
miRBase ID
miRBase Accession Number
Mouse
mmu-miR-26a-1-3p
MIMAT0017020
Mouse
mmu-miR-26a-5p
MIMAT0000533
Stem-loop Details
Mouse
Stem-loop ID
mmu-mir-26a-1
Stem-loop Accession #
MI0000573
Stem-loop Sequence
AAGGCCGUGGCCUCGUUCAAGUAAUCCAGGAUAGGCUGUGCAGGUCCCAAGGGGCCUAUUCUUGGUUACUUGCACGGGGACGCGGGCCUG
Chromosome Location
Chr. 9 - 119031796 - 119031885 [+] on Build GRCm38
Mature MicroRNA
miRBase Accession #: MIMAT0017020
miRBase ID: mmu-miR-26a-1-3p
Mature miRNA Sequence: CCUAUUCUUGGUUACUUGCACG
Chromosome Location: Chr. 9 - 119031796 - 119031885 [+] on Build GRCm38
miRBase Accession #: MIMAT0000533
miRBase ID: mmu-miR-26a-5p
Mature miRNA Sequence: UUCAAGUAAUCCAGGAUAGGCU
Chromosome Location: Chr. 9 - 119031796 - 119031885 [+] on Build GRCm38
Gene Family ID
MIPF0000043,
mir-26
Distance to Amplicon
165 bases
Back To Top
More Information
Related Products
Ambion
®
Anti-miR
™
miRNA Inhibitor :
AM12758
Ambion
®
Pre-miR
™
miRNA Precursor :
PM12758
TaqMan
™
MicroRNA Assay :
002443
mir
Vana
®
miRNA inhibitor :
MH12758
mir
Vana
®
miRNA mimic :
MC12758
TaqMan
™
Advanced miRNA Assay :
478787_mir
TaqMan
™
Advanced miRNA Assay :
mmu481661_mir
Ambion
®
Anti-miR
™
miRNA Inhibitor :
AM10249
Ambion
®
Pre-miR
™
miRNA Precursor :
PM10249
TaqMan
™
MicroRNA Assay :
000405
mir
Vana
®
miRNA inhibitor :
MH10249
mir
Vana
®
miRNA mimic :
MC10249
TaqMan
™
Advanced miRNA Assay :
mmu481013_mir
TaqMan
™
Advanced miRNA Assay :
477995_mir
TaqMan
™
Advanced miRNA Assay :
rno481013_mir
Back To Top
Related Products
High Capacity RNA-to-cDNA Kit
SuperScript
®
IV VILO
™
Master Mix
TaqMan
®
Universal PCR Master Mix
TaqMan
®
Fast Advanced Master Mix
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-67655d8755-zsnhf:80/100.66.75.14:80.
git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.46.0-Offline