Search
Search

| Assay Name: | rno-miR-411-5p |
|---|---|
| Stem-loop Accession Number: | MI0003675 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | UAGUAGACCGUAUAGCGUACG |
| Species: | Human, Mouse, Rat, Callithrix jacchus, Dasypus novemcinctus, Guinea pig, Horse, Microcebus murinus, Otolemur garnettii, Pan paniscus, Pan troglodytes, Papio hamadryas, Pongo pygmaeus, Rabbit, Rhesus monkey, Tupaia chinensis |
| Product Type: | TaqMan™ Advanced miRNA Assay |
Gene Family ID: | MIPF0000126, mir-379 |
Mature miRNA Sequence: | UAGUAGACCGUAUAGCGUACG |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-411-5p | MIMAT0003329 |
| Mouse | mmu-miR-411-5p | MIMAT0004747 |
| Rat | rno-miR-411-5p | MIMAT0005312 |
| Callithrix jacchus | cja-miR-411 | MIMAT0039445 |
| Dasypus novemcinctus | dno-miR-411-5p | MIMAT0047874 |
| Guinea pig | cpo-miR-411-5p | MIMAT0047237 |
| View More Rows | ||
| Horse | eca-miR-411 | MIMAT0013154 |
| Microcebus murinus | mmr-miR-411a | MIMAT0049194 |
| Otolemur garnettii | oga-miR-411a | MIMAT0049676 |
| Pan paniscus | ppa-miR-411a | MIMAT0049046 |
| Pan troglodytes | ptr-miR-411 | MIMAT0008138 |
| Papio hamadryas | pha-miR-411a | MIMAT0049579 |
| Pongo pygmaeus | ppy-miR-411 | MIMAT0015878 |
| Rabbit | ocu-miR-411-5p | MIMAT0048437 |
| Rhesus monkey | mml-miR-411-5p | MIMAT0006317 |
| Tupaia chinensis | tch-miR-411-5p | MIMAT0036490 |