Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
New Products
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
Home
›
Search Tool
›
Search Results
› Rn03464919_pri
Assay Name:
rno-mir-411
Stem-loop ID:
rno-mir-411
Mapped to miRBase Version:
v22.1
Chromosome Location:
Chr.6: 133858849 - 133858924 [+] on Build Rnor_6.0
Stem-loop Location:
Chr.6: 133858849 - 133858924 [+] on Build Rnor_6.0
Mature miRNA ID:
rno-miR-411-3p
rno-miR-411-5p
Species:
Rat
Product Type:
TaqMan
™
Pri-miRNA Assay
Assay ID
Rn03464919_pri
Size
S: 360 rxns; 20X
M: 750 rxns; 20X
L: 2,900 rxns; 60X
Availability
Made To Order
Catalog #
4427012
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
Assay Details
More Information
Genomic Map
LOADING...
×
Back To Top
Assay Details
Amplicon Length:
68
Chromosome Location:
Chr. 6 - 133858701 - 133858701 [+] on 133858701 Build Rnor_6.0
Species:
Rat
miRBase Details:
Species
miRBase ID
miRBase Accession Number
Rat
rno-miR-411-3p
MIMAT0017304
Rat
rno-miR-411-5p
MIMAT0005312
Stem-loop Details
Rat
Stem-loop ID
rno-mir-411
Stem-loop Accession #
MI0006144
Stem-loop Sequence
CUUGGAGAGAUAGUAGACCGUAUAGCGUACGCUUUAUCUGUGACGUAUGUAACACGGUCCACUAACCCUCAGUAUC
Chromosome Location
Chr. 6 - 133858849 - 133858924 [+] on Build Rnor_6.0
Mature MicroRNA
miRBase Accession #: MIMAT0017304
miRBase ID: rno-miR-411-3p
Mature miRNA Sequence: UAUGUAACACGGUCCACUAA
Chromosome Location: Chr. 6 - 133858849 - 133858924 [+] on Build Rnor_6.0
miRBase Accession #: MIMAT0005312
miRBase ID: rno-miR-411-5p
Mature miRNA Sequence: UAGUAGACCGUAUAGCGUACG
Chromosome Location: Chr. 6 - 133858849 - 133858924 [+] on Build Rnor_6.0
Gene Family ID
MIPF0000126,
mir-379
Distance to Amplicon
117 bases
Back To Top
More Information
Related Products
Ambion
®
Anti-miR
™
miRNA Inhibitor :
AM19703
Ambion
®
Pre-miR
™
miRNA Precursor :
PM19703
TaqMan
™
MicroRNA Assay :
464124_mat
mir
Vana
®
miRNA inhibitor :
MH19703
mir
Vana
®
miRNA mimic :
MC19703
TaqMan
™
Advanced miRNA Assay :
rno481480_mir
Ambion
®
Anti-miR
™
miRNA Inhibitor :
AM11409
Ambion
®
Pre-miR
™
miRNA Precursor :
PM11409
TaqMan
™
MicroRNA Assay :
001610
mir
Vana
®
miRNA inhibitor :
MH11409
mir
Vana
®
miRNA mimic :
MC11409
TaqMan
™
Advanced miRNA Assay :
478086_mir
TaqMan
™
Advanced miRNA Assay :
rno481157_mir
TaqMan
™
Advanced miRNA Assay :
mmu481157_mir
Back To Top
Related Products
High Capacity RNA-to-cDNA Kit
SuperScript
®
IV VILO
™
Master Mix
TaqMan
®
Universal PCR Master Mix
TaqMan
®
Fast Advanced Master Mix
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-5db85fbf84-q86qf:80/100.66.76.64:80.
git-commit: 64d6f4084a597c0a860dbc264f43a6f389f0d7ac
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.60.1-2026-secondary-format-patch