Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Clinical Genomics
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • New Products
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › 478086_mir

          Specificity testing was performed using human anti-targets.

          Assay Name: hsa-miR-411-5p
          Stem-loop Accession Number: MI0003675
          miRBase Version: v22.1
          Chromosome Location: Chr.14: 101023325 - 101023420 [+] on Build GRCh38
          Mature miRNA Sequence: UAGUAGACCGUAUAGCGUACG
          Species: Human, Mouse, Rat, Callithrix jacchus, Dasypus novemcinctus, Guinea pig, Horse, Microcebus murinus, Otolemur garnettii, Pan paniscus, Pan troglodytes, Papio hamadryas, Pongo pygmaeus, Rabbit, Rhesus monkey, Tupaia chinensis
          Product Type: TaqMan™ Advanced miRNA Assay
          Assay ID 478086_mir
          Size S: 250 rxns
          Availability Inventoried
          Catalog # A25576
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Family ID:

          MIPF0000126, mir-379

          Mature miRNA Sequence:

          UAGUAGACCGUAUAGCGUACG

          miRBase Details:



          Species miRBase ID miRBase Accession Number
          Human hsa-miR-411-5p MIMAT0003329
          Mouse mmu-miR-411-5p MIMAT0004747
          Rat rno-miR-411-5p MIMAT0005312
          Callithrix jacchus cja-miR-411 MIMAT0039445
          Dasypus novemcinctus dno-miR-411-5p MIMAT0047874
          Guinea pig cpo-miR-411-5p MIMAT0047237
          View More Rows
          Horse eca-miR-411 MIMAT0013154
          Microcebus murinus mmr-miR-411a MIMAT0049194
          Otolemur garnettii oga-miR-411a MIMAT0049676
          Pan paniscus ppa-miR-411a MIMAT0049046
          Pan troglodytes ptr-miR-411 MIMAT0008138
          Papio hamadryas pha-miR-411a MIMAT0049579
          Pongo pygmaeus ppy-miR-411 MIMAT0015878
          Rabbit ocu-miR-411-5p MIMAT0048437
          Rhesus monkey mml-miR-411-5p MIMAT0006317
          Tupaia chinensis tch-miR-411-5p MIMAT0036490

          Stem-loop Details



          Human

          Stem-loop ID hsa-mir-411
          Stem-loop Accession # MI0003675
          Stem-loop Sequence
          UGGUACUUGGAGAGAUAGUAGACCGUAUAGCGUACGCUUUAUCUGUGACGUAUGUAACACGGUCCACUAACCCUCAGUAUCAAAUCCAUCCCCGAG
          Chromosome Location Chr. 14 - 101023325 - 101023420 [+] on Build GRCh38
          Mature MicroRNA miRBase Accession #: MIMAT0003329
          miRBase ID: hsa-miR-411-5p
          Mature miRNA Sequence: UAGUAGACCGUAUAGCGUACG
          Chromosome Location: Chr. 14 - 101023325 - 101023420 [+] on Build GRCh38

          Mouse

          Stem-loop ID mmu-mir-411
          Stem-loop Accession # MI0001163
          Stem-loop Sequence
          UGGUACUUGGAGAGAUAGUAGACCGUAUAGCGUACGCUUUAUCUGUGACGUAUGUAACACGGUCCACUAACCCUCAGUAUCA
          Chromosome Location Chr. 12 - 109710175 - 109710256 [+] on Build GRCm38
          Mature MicroRNA miRBase Accession #: MIMAT0004747
          miRBase ID: mmu-miR-411-5p
          Mature miRNA Sequence: UAGUAGACCGUAUAGCGUACG
          Chromosome Location: Chr. 12 - 109710175 - 109710256 [+] on Build GRCm38

          Rat

          Stem-loop ID rno-mir-411
          Stem-loop Accession # MI0006144
          Stem-loop Sequence
          CUUGGAGAGAUAGUAGACCGUAUAGCGUACGCUUUAUCUGUGACGUAUGUAACACGGUCCACUAACCCUCAGUAUC
          Chromosome Location Chr. 6 - 133858849 - 133858924 [+] on Build Rnor_6.0
          Mature MicroRNA miRBase Accession #: MIMAT0005312
          miRBase ID: rno-miR-411-5p
          Mature miRNA Sequence: UAGUAGACCGUAUAGCGUACG
          Chromosome Location: Chr. 6 - 133858849 - 133858924 [+] on Build Rnor_6.0

          Callithrix jacchus

          Stem-loop ID cja-mir-411
          Stem-loop Accession # MI0031968
          Stem-loop Sequence
          UGGUACUUAGAGAGAUAGUAGACCGUAUAGCGUACGCUUUAUCUGUGACGUAUGUAACACGGUCCACUAACCCUCAGUAUCA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0039445
          miRBase ID: cja-miR-411
          Mature miRNA Sequence: UAGUAGACCGUAUAGCGUACG
          Chromosome Location: NA

          Dasypus novemcinctus

          Stem-loop ID dno-mir-411
          Stem-loop Accession # MI0039102
          Stem-loop Sequence
          UAGUAGACCGUAUAGCGUACGCUUUAUCUGUGACGUAUGUAACACGGUCCACUAAC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0047874
          miRBase ID: dno-miR-411-5p
          Mature miRNA Sequence: UAGUAGACCGUAUAGCGUACG
          Chromosome Location: NA

          Guinea pig

          Stem-loop ID cpo-mir-411
          Stem-loop Accession # MI0038767
          Stem-loop Sequence
          UAGUAGACCGUAUAGCGUACGCUUUAUCUGUGACGUAUGUAACACGGUCCACUAAC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0047237
          miRBase ID: cpo-miR-411-5p
          Mature miRNA Sequence: UAGUAGACCGUAUAGCGUACG
          Chromosome Location: NA

          Horse

          Stem-loop ID eca-mir-411
          Stem-loop Accession # MI0012898
          Stem-loop Sequence
          UGGUACUUGGAGAGAUAGUAGACCGUAUAGCGUACGCUUUAUCUGUGACGUAUGUAACACGGUCCACUAACCCUCAGUAUCA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0013154
          miRBase ID: eca-miR-411
          Mature miRNA Sequence: UAGUAGACCGUAUAGCGUACG
          Chromosome Location: NA

          Microcebus murinus

          Stem-loop ID mmr-mir-411a
          Stem-loop Accession # MI0039913
          Stem-loop Sequence
          UAGUAGACCGUAUAGCGUACGCUUUAUCUGUGACGUAUGUAACACGGUCCACUAAC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0049194
          miRBase ID: mmr-miR-411a
          Mature miRNA Sequence: UAGUAGACCGUAUAGCGUACG
          Chromosome Location: NA

          Otolemur garnettii

          Stem-loop ID oga-mir-411a
          Stem-loop Accession # MI0040459
          Stem-loop Sequence
          UAGUAGACCGUAUAGCGUACGCUUUAUCUGUGACGUAUGUAACACGGUCCACUAAC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0049676
          miRBase ID: oga-miR-411a
          Mature miRNA Sequence: UAGUAGACCGUAUAGCGUACG
          Chromosome Location: NA

          Pan paniscus

          Stem-loop ID ppa-mir-411a
          Stem-loop Accession # MI0039754
          Stem-loop Sequence
          UAGUAGACCGUAUAGCGUACGCUUUAUCUGUGACGUAUGUAACACAGUCCACUAAC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0049046
          miRBase ID: ppa-miR-411a
          Mature miRNA Sequence: UAGUAGACCGUAUAGCGUACG
          Chromosome Location: NA

          Pan troglodytes

          Stem-loop ID ptr-mir-411
          Stem-loop Accession # MI0008659
          Stem-loop Sequence
          GGUACUUGGAGAGAUAGUAGACCGUAUAGCGUACGCUUUAUCUGUGACGUAUGUAACACGGUCCACUAACCCUCAGUAUCAAAUCCAUCCCCGAG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0008138
          miRBase ID: ptr-miR-411
          Mature miRNA Sequence: UAGUAGACCGUAUAGCGUACG
          Chromosome Location: NA

          Papio hamadryas

          Stem-loop ID pha-mir-411a
          Stem-loop Accession # MI0040354
          Stem-loop Sequence
          UAGUAGACCGUAUAGCGUACGCUUUAUCUGUGACGUAUGUAACACGGUCCACUAAC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0049579
          miRBase ID: pha-miR-411a
          Mature miRNA Sequence: UAGUAGACCGUAUAGCGUACG
          Chromosome Location: NA

          Pongo pygmaeus

          Stem-loop ID ppy-mir-411
          Stem-loop Accession # MI0014935
          Stem-loop Sequence
          UGGUACUUGGAGAGAUAGUAGACCGUAUAGCGUACGCUUUAUCUGUGACGUAUGUAACACGGUCCACUAACCCUCAGUAUCAAAUCCAUCCCCGAG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0015878
          miRBase ID: ppy-miR-411
          Mature miRNA Sequence: UAGUAGACCGUAUAGCGUACG
          Chromosome Location: NA

          Rabbit

          Stem-loop ID ocu-mir-411
          Stem-loop Accession # MI0039400
          Stem-loop Sequence
          UAGUAGACCGUAUAGCGUACGCUUUAUCUGUGACGUAUGUAACACGGUCCACUAAC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0048437
          miRBase ID: ocu-miR-411-5p
          Mature miRNA Sequence: UAGUAGACCGUAUAGCGUACG
          Chromosome Location: NA

          Rhesus monkey

          Stem-loop ID mml-mir-411
          Stem-loop Accession # MI0007735
          Stem-loop Sequence
          UGGUACUUGGAGAGAUAGUAGACCGUAUAGCGUACGCUUUAUCUGUGACGUAUGUAACACGGUCCACUAACCCUCAGUAUCA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0006317
          miRBase ID: mml-miR-411-5p
          Mature miRNA Sequence: UAGUAGACCGUAUAGCGUACG
          Chromosome Location: NA

          Tupaia chinensis

          Stem-loop ID tch-mir-411
          Stem-loop Accession # MI0031148
          Stem-loop Sequence
          UAGUAGACCGUAUAGCGUACGCUUUAUCUGUGACGUAUGUAACACGGUCCACUAACC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0036490
          miRBase ID: tch-miR-411-5p
          Mature miRNA Sequence: UAGUAGACCGUAUAGCGUACG
          Chromosome Location: NA


          Back To Top

          More Information




          Related Products

          TaqMan™ Pri-miRNA Assay : Mm03307452_pri
          TaqMan™ Pri-miRNA Assay : Hs03303403_pri
          TaqMan™ Pri-miRNA Assay : Rn03464919_pri
          Ambion® Anti-miR™ miRNA Inhibitor : AM11409
          Ambion® Pre-miR™ miRNA Precursor : PM11409
          TaqMan™ MicroRNA Assay : 001610
          mirVana® miRNA inhibitor : MH11409
          mirVana® miRNA mimic : MC11409
          TaqMan™ Advanced miRNA Assay : rno481157_mir
          TaqMan™ Advanced miRNA Assay : mmu481157_mir
          MIRNA : TaqMan® Advanced miRNA Human A and B 96-well Plates, Fast
          MIRNA : TaqMan® OpenArray® Human Advanced MicroRNA Panel
          MIRNA : TaqMan® Advanced miRNA Human A and B 96-well Plates, Standard
          FGMIR : TaqManTM Advanced miRNA Human A Card
          MIRNA : TaqMan® Advanced miRNA Human A and B Cards
          MIRNA : TaqMan® Advanced miRNA Human A Card
          FGMIR : TaqManTM Advanced miRNA Human A 96-well Plate 3
          FGMIR : QS TaqmanTM OpenArray Human Advanced miRNA Panel
          MIRNA : TaqMan® Advanced miRNA Human A 96-well Plates, Fast
          MIRNA : TaqMan® Advanced miRNA Human A 96-well Plates, Standard
          FGMIR : TaqManTM Advanced miRNA Human A 96-well Plate 3

          Back To Top

          Related Products

          • TaqMan® Advanced miRNA cDNA Synthesis Kit
          • TaqMan® Fast Advanced Master Mix
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Hong Kong flag icon
          Hong Kong

          Your items have has been added!


          Host server : magellan-search-blue-7fb9f846c9-xq5gc:80/100.66.76.64:80.
          git-commit: 8207b5afa5c305bd40e0a4f0b9b53fd46f2a8829
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.59.1-2026.09.91-1.0