Search
Search

| Assay Name: | rno-miR-410-5p |
|---|---|
| Stem-loop Accession Number: | MI0002465 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | AGGUUGUCUGUGAUGAGUUCG |
| Species: | Human, Mouse, Rat, Cricetulus griseus, Guinea pig, Pteropus alecto, Rabbit, Rhesus monkey |
| Product Type: | TaqMan™ Advanced miRNA Assay |
Gene Family ID: | MIPF0000018, mir-154 |
Mature miRNA Sequence: | AGGUUGUCUGUGAUGAGUUCG |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-410-5p | MIMAT0026558 |
| Mouse | mmu-miR-410-5p | MIMAT0017172 |
| Rat | rno-miR-410-5p | MIMAT0017303 |
| Cricetulus griseus | cgr-miR-410-5p | MIMAT0023952 |
| Guinea pig | cpo-miR-410-5p | MIMAT0047235 |
| Pteropus alecto | pal-miR-410-5p | MIMAT0040151 |
| View More Rows | ||
| Rabbit | ocu-miR-410-5p | MIMAT0048435 |
| Rhesus monkey | mml-miR-410-5p | MIMAT0026869 |