Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm03307442_pri
          Assay Name:
          mmu-mir-410
          Stem-loop ID:
          mmu-mir-410
          Mapped to miRBase Version:
          v22.1
          Chromosome Location:
          Chr.12: 109743715 - 109743795 [+] on Build GRCm38
          Stem-loop Location:
          Chr.12: 109743715 - 109743795 [+] on Build GRCm38
          Mature miRNA ID:
          mmu-miR-410-3p mmu-miR-410-5p
          Species:
          Mouse
          Product Type:
          TaqMan™ Pri-miRNA Assay
          Assay ID Mm03307442_pri
          Size
          Availability Made To Order
          Catalog # 4427012
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Amplicon Length:

          58

          Chromosome Location:

          Chr. 12 - 109744012 - 109744012 [+] on 109744012 Build GRCm38

          Species:

          Mouse

          miRBase Details:



          Species miRBase ID miRBase Accession Number
          Mouse mmu-miR-410-3p MIMAT0001091
          Mouse mmu-miR-410-5p MIMAT0017172

          Stem-loop Details



          Mouse

          Stem-loop ID mmu-mir-410
          Stem-loop Accession # MI0001161
          Stem-loop Sequence
          GGGUACUUGAGGAGAGGUUGUCUGUGAUGAGUUCGCUUUAUUAAUGACGAAUAUAACACAGAUGGCCUGUUUUCAAUACCA
          Chromosome Location Chr. 12 - 109743715 - 109743795 [+] on Build GRCm38
          Mature MicroRNA miRBase Accession #: MIMAT0001091
          miRBase ID: mmu-miR-410-3p
          Mature miRNA Sequence: AAUAUAACACAGAUGGCCUGU
          Chromosome Location: Chr. 12 - 109743715 - 109743795 [+] on Build GRCm38

          miRBase Accession #: MIMAT0017172
          miRBase ID: mmu-miR-410-5p
          Mature miRNA Sequence: AGGUUGUCUGUGAUGAGUUCG
          Chromosome Location: Chr. 12 - 109743715 - 109743795 [+] on Build GRCm38

          Gene Family ID MIPF0000018, mir-154
          Distance to Amplicon 189 bases

          Back To Top

          More Information




          Related Products

          Ambion® Anti-miR™ miRNA Inhibitor : AM11119
          Ambion® Pre-miR™ miRNA Precursor : PM11119
          TaqMan™ MicroRNA Assay : 001274
          mirVana® miRNA inhibitor : MH11119
          mirVana® miRNA mimic : MC11119
          TaqMan™ Advanced miRNA Assay : rno481156_mir
          TaqMan™ Advanced miRNA Assay : 478085_mir
          TaqMan™ Advanced miRNA Assay : mmu481156_mir
          Ambion® Anti-miR™ miRNA Inhibitor : AM19960
          Ambion® Pre-miR™ miRNA Precursor : PM19960
          TaqMan™ MicroRNA Assay : 464693_mat
          mirVana® miRNA inhibitor : MH19960
          mirVana® miRNA mimic : MC19960
          TaqMan™ Advanced miRNA Assay : rno481391_mir
          TaqMan™ Advanced miRNA Assay : mmu482760_mir
          TaqMan™ Advanced miRNA Assay : 478873_mir

          Back To Top

          Related Products

          • High Capacity RNA-to-cDNA Kit
          • SuperScript® IV VILO™ Master Mix
          • TaqMan® Universal PCR Master Mix
          • TaqMan® Fast Advanced Master Mix
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-b2k9d:80/100.66.79.173:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline