Search
Search

| Assay Name: | mmu-miR-410-3p |
|---|---|
| Stem-loop Accession Number: | MI0002465 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | AAUAUAACACAGAUGGCCUGU |
| Species: | Human, Mouse, Rat, Bovine, Callithrix jacchus, Cricetulus griseus, Dasypus novemcinctus, Dog, Goat, Guinea pig, Horse, Microcebus murinus, Nomascus leucogenys, Otolemur garnettii, Pan paniscus, Pan troglodytes, Pongo pygmaeus, Pteropus alecto, Rabbit, Rhesus monkey, Sheep |
| Product Type: | TaqMan™ Advanced miRNA Assay |
Gene Family ID: | MIPF0000018, mir-154 |
Mature miRNA Sequence: | AAUAUAACACAGAUGGCCUGU |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-410-3p | MIMAT0002171 |
| Mouse | mmu-miR-410-3p | MIMAT0001091 |
| Rat | rno-miR-410-3p | MIMAT0005311 |
| Bovine | bta-miR-410 | MIMAT0009311 |
| Callithrix jacchus | cja-miR-410 | MIMAT0039354 |
| Cricetulus griseus | cgr-miR-410-3p | MIMAT0023953 |
| View More Rows | ||
| Dasypus novemcinctus | dno-miR-410-3p | MIMAT0047873 |
| Dog | cfa-miR-410 | MIMAT0006728 |
| Goat | chi-miR-410-3p | MIMAT0036212 |
| Guinea pig | cpo-miR-410-3p | MIMAT0047236 |
| Horse | eca-miR-410 | MIMAT0013153 |
| Microcebus murinus | mmr-miR-410 | MIMAT0049189 |
| Nomascus leucogenys | nle-miR-410 | MIMAT0049369 |
| Otolemur garnettii | oga-miR-410 | MIMAT0049673 |
| Pan paniscus | ppa-miR-410 | MIMAT0049033 |
| Pan troglodytes | ptr-miR-410 | MIMAT0008137 |
| Pongo pygmaeus | ppy-miR-410 | MIMAT0015877 |
| Pteropus alecto | pal-miR-410-3p | MIMAT0040152 |
| Rabbit | ocu-miR-410-3p | MIMAT0048436 |
| Rhesus monkey | mml-miR-410-3p | MIMAT0006316 |
| Sheep | oar-miR-410-3p | MIMAT0019334 |