Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › 001274
          Assay Name: hsa-miR-410
          miRBase Accession Number: MI0002465
          miRBase Version: v22.1
          Chromosome Location: Chr.14: 101065912 - 101065991 [+] on Build GRCh38
          Mature miRNA Sequence: AAUAUAACACAGAUGGCCUGU
          Species: Human, Mouse, Rat, Bovine, Callithrix jacchus, Cricetulus griseus, Dasypus novemcinctus, Dog, Goat, Guinea pig, Horse, Microcebus murinus, Nomascus leucogenys, Otolemur garnettii, Pan paniscus, Pan troglodytes, Pongo pygmaeus, Pteropus alecto, Rabbit, Rhesus monkey, Sheep
          Product Type: TaqMan™ MicroRNA Assay
          Assay ID 001274
          Size
          Availability Inventoried
          Catalog # 4427975
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Family ID:

          MIPF0000018, mir-154

          Mature miRNA Sequence:

          AAUAUAACACAGAUGGCCUGU

          Mature miRNA Details:



          Species miRBase ID miRBase Accession Number
          Human hsa-miR-410-3p MIMAT0002171
          Mouse mmu-miR-410-3p MIMAT0001091
          Rat rno-miR-410-3p MIMAT0005311
          Bovine bta-miR-410 MIMAT0009311
          Callithrix jacchus cja-miR-410 MIMAT0039354
          Cricetulus griseus cgr-miR-410-3p MIMAT0023953
          View More Rows
          Dasypus novemcinctus dno-miR-410-3p MIMAT0047873
          Dog cfa-miR-410 MIMAT0006728
          Goat chi-miR-410-3p MIMAT0036212
          Guinea pig cpo-miR-410-3p MIMAT0047236
          Horse eca-miR-410 MIMAT0013153
          Microcebus murinus mmr-miR-410 MIMAT0049189
          Nomascus leucogenys nle-miR-410 MIMAT0049369
          Otolemur garnettii oga-miR-410 MIMAT0049673
          Pan paniscus ppa-miR-410 MIMAT0049033
          Pan troglodytes ptr-miR-410 MIMAT0008137
          Pongo pygmaeus ppy-miR-410 MIMAT0015877
          Pteropus alecto pal-miR-410-3p MIMAT0040152
          Rabbit ocu-miR-410-3p MIMAT0048436
          Rhesus monkey mml-miR-410-3p MIMAT0006316
          Sheep oar-miR-410-3p MIMAT0019334

          Stem-loop Details



          Human

          Stem-loop ID hsa-mir-410
          Stem-loop Accession # MI0002465
          Stem-loop Sequence
          GGUACCUGAGAAGAGGUUGUCUGUGAUGAGUUCGCUUUUAUUAAUGACGAAUAUAACACAGAUGGCCUGUUUUCAGUACC
          Chromosome Location Chr. 14 - 101065912 - 101065991 [+] on Build GRCh38
          Mature MicroRNA miRBase Accession #: MIMAT0002171
          miRBase ID: hsa-miR-410-3p
          Mature miRNA Sequence: AAUAUAACACAGAUGGCCUGU
          Chromosome Location: Chr. 14 - 101065912 - 101065991 [+] on Build GRCh38

          Mouse

          Stem-loop ID mmu-mir-410
          Stem-loop Accession # MI0001161
          Stem-loop Sequence
          GGGUACUUGAGGAGAGGUUGUCUGUGAUGAGUUCGCUUUAUUAAUGACGAAUAUAACACAGAUGGCCUGUUUUCAAUACCA
          Chromosome Location Chr. 12 - 109743715 - 109743795 [+] on Build GRCm38
          Mature MicroRNA miRBase Accession #: MIMAT0001091
          miRBase ID: mmu-miR-410-3p
          Mature miRNA Sequence: AAUAUAACACAGAUGGCCUGU
          Chromosome Location: Chr. 12 - 109743715 - 109743795 [+] on Build GRCm38

          Rat

          Stem-loop ID rno-mir-410
          Stem-loop Accession # MI0006143
          Stem-loop Sequence
          ACUUGAGGAGAGGUUGUCUGUGAUGAGUUCGCUUUAUUAAUGACGAAUAUAACACAGAUGGCCUGUUUUCAAUACC
          Chromosome Location Chr. 6 - 133893997 - 133894072 [+] on Build Rnor_6.0
          Mature MicroRNA miRBase Accession #: MIMAT0005311
          miRBase ID: rno-miR-410-3p
          Mature miRNA Sequence: AAUAUAACACAGAUGGCCUGU
          Chromosome Location: Chr. 6 - 133893997 - 133894072 [+] on Build Rnor_6.0

          Bovine

          Stem-loop ID bta-mir-410
          Stem-loop Accession # MI0009825
          Stem-loop Sequence
          GGGUACUUGAGGAGAGGUUGUCUGUGAUGAGUUCGCUUUAUUAAUGACGAAUAUAACACAGAUGGCCUGUUUUCAGUACCA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0009311
          miRBase ID: bta-miR-410
          Mature miRNA Sequence: AAUAUAACACAGAUGGCCUGU
          Chromosome Location: NA

          Callithrix jacchus

          Stem-loop ID cja-mir-410
          Stem-loop Accession # MI0031868
          Stem-loop Sequence
          GGUACCUGAGGAGAGGUUGUCUGUGAUGAGUUCGCUUUUAUUAAUGACGAAUAUAACACAGAUGGCCUGUUUUCAGUACC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0039354
          miRBase ID: cja-miR-410
          Mature miRNA Sequence: AAUAUAACACAGAUGGCCUGU
          Chromosome Location: NA

          Cricetulus griseus

          Stem-loop ID cgr-mir-410
          Stem-loop Accession # MI0020521
          Stem-loop Sequence
          CUUGAGGAGAGGUUGUCUGUGAUGAGUUCGCUUUAUUAAUGACGAAUAUAACACAGAUGGCCUGUUUUCAAUACC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0023953
          miRBase ID: cgr-miR-410-3p
          Mature miRNA Sequence: AAUAUAACACAGAUGGCCUGU
          Chromosome Location: NA

          Dasypus novemcinctus

          Stem-loop ID dno-mir-410
          Stem-loop Accession # MI0039101
          Stem-loop Sequence
          AGGCUGUCUGUGAUGAGUUCGCUUUAUUAAUGACGAAUAUAACACAGAUGGCCUGU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0047873
          miRBase ID: dno-miR-410-3p
          Mature miRNA Sequence: AAUAUAACACAGAUGGCCUGU
          Chromosome Location: NA

          Dog

          Stem-loop ID cfa-mir-410
          Stem-loop Accession # MI0008149
          Stem-loop Sequence
          AGGUUGUCUGUGAUGAGUUCGCUUUAUUAAUGCCGAAUAUAACACAGAUGGCCUGU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0006728
          miRBase ID: cfa-miR-410
          Mature miRNA Sequence: AAUAUAACACAGAUGGCCUGU
          Chromosome Location: NA

          Goat

          Stem-loop ID chi-mir-410
          Stem-loop Accession # MI0030783
          Stem-loop Sequence
          CACCUGCGGUCUGUGGAGUAGGCGCCACUUUGGGUACUUGAGGAGAGGUUGUCUGUGAUGAGUUCGCUUUAUUAAUGACGAAUAUAACACAGAUGGCCUGUUUUCAGUACCACCCACCCCUCGUG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0036212
          miRBase ID: chi-miR-410-3p
          Mature miRNA Sequence: AAUAUAACACAGAUGGCCUGU
          Chromosome Location: NA

          Guinea pig

          Stem-loop ID cpo-mir-410
          Stem-loop Accession # MI0038766
          Stem-loop Sequence
          AGGUUGUCUGUGAUGAGUUCGCUUUAUUAAUGACGAAUAUAACACAGAUGGCCUGU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0047236
          miRBase ID: cpo-miR-410-3p
          Mature miRNA Sequence: AAUAUAACACAGAUGGCCUGU
          Chromosome Location: NA

          Horse

          Stem-loop ID eca-mir-410
          Stem-loop Accession # MI0012897
          Stem-loop Sequence
          UGAGGAGAGGUUGUCUGUGAUGAGUUCGCUUUAUUAAUGACGAAUAUAACACAGAUGGCCUGUCUUCAGUACCA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0013153
          miRBase ID: eca-miR-410
          Mature miRNA Sequence: AAUAUAACACAGAUGGCCUGU
          Chromosome Location: NA

          Microcebus murinus

          Stem-loop ID mmr-mir-410
          Stem-loop Accession # MI0039908
          Stem-loop Sequence
          AGGUUGUCUGUGAUGAGUUCGCUUUAUUAAUGACGAAUAUAACACAGAUGGCCUGU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0049189
          miRBase ID: mmr-miR-410
          Mature miRNA Sequence: AAUAUAACACAGAUGGCCUGU
          Chromosome Location: NA

          Nomascus leucogenys

          Stem-loop ID nle-mir-410
          Stem-loop Accession # MI0040106
          Stem-loop Sequence
          AGGUUGUCUGUGAUGAGUUCGCUUUUAUUAAUGACGAAUAUAACACAGAUGGCCUGU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0049369
          miRBase ID: nle-miR-410
          Mature miRNA Sequence: AAUAUAACACAGAUGGCCUGU
          Chromosome Location: NA

          Otolemur garnettii

          Stem-loop ID oga-mir-410
          Stem-loop Accession # MI0040456
          Stem-loop Sequence
          AGGUUGUCUGUGAUGAGUUCGCUUUAUUAAUGACGAAUAUAACACAGAUGGCCUGU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0049673
          miRBase ID: oga-miR-410
          Mature miRNA Sequence: AAUAUAACACAGAUGGCCUGU
          Chromosome Location: NA

          Pan paniscus

          Stem-loop ID ppa-mir-410
          Stem-loop Accession # MI0039741
          Stem-loop Sequence
          AGGUUGUCUGUGAUGAGUUCGCUUUUAUUAAUGACGAAUAUAACACAGAUGGCCUGU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0049033
          miRBase ID: ppa-miR-410
          Mature miRNA Sequence: AAUAUAACACAGAUGGCCUGU
          Chromosome Location: NA

          Pan troglodytes

          Stem-loop ID ptr-mir-410
          Stem-loop Accession # MI0008658
          Stem-loop Sequence
          GUACCUGAGAAGAGGUUGUCUGUGAUGAGUUCGCUUUUAUUAAUGACGAAUAUAACACAGAUGGCCUGUUUUCAGUACC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0008137
          miRBase ID: ptr-miR-410
          Mature miRNA Sequence: AAUAUAACACAGAUGGCCUGU
          Chromosome Location: NA

          Pongo pygmaeus

          Stem-loop ID ppy-mir-410
          Stem-loop Accession # MI0014934
          Stem-loop Sequence
          GGUACCUGAGAAGAGGUUGUCUGUGAUGAGUUCGCUUUUAUUAAUGACGAAUAUAACACAGAUGGCCUGUUUUCAGUACC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0015877
          miRBase ID: ppy-miR-410
          Mature miRNA Sequence: AAUAUAACACAGAUGGCCUGU
          Chromosome Location: NA

          Pteropus alecto

          Stem-loop ID pal-mir-410
          Stem-loop Accession # MI0032520
          Stem-loop Sequence
          AGGUUGUCUGUGAUGAGUUCGCUUUAUUAAUGACGAAUAUAACACAGAUGGCCUGU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0040152
          miRBase ID: pal-miR-410-3p
          Mature miRNA Sequence: AAUAUAACACAGAUGGCCUGU
          Chromosome Location: NA

          Rabbit

          Stem-loop ID ocu-mir-410
          Stem-loop Accession # MI0039399
          Stem-loop Sequence
          AGGUUGUCUGUGAUGAGUUCGCUGUAUUAAAGCCGAAUAUAACACAGAUGGCCUGU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0048436
          miRBase ID: ocu-miR-410-3p
          Mature miRNA Sequence: AAUAUAACACAGAUGGCCUGU
          Chromosome Location: NA

          Rhesus monkey

          Stem-loop ID mml-mir-410
          Stem-loop Accession # MI0007734
          Stem-loop Sequence
          GGUACCUGAGGAGAGGUUGUCUGUGAUGAGUUCGCUUUUAUUAAUGACGAAUAUAACACAGAUGGCCUGUUUUCAGUACC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0006316
          miRBase ID: mml-miR-410-3p
          Mature miRNA Sequence: AAUAUAACACAGAUGGCCUGU
          Chromosome Location: NA

          Sheep

          Stem-loop ID oar-mir-410
          Stem-loop Accession # MI0016961
          Stem-loop Sequence
          CACCUGCGGUCUGUGGAGUAGGCGCCACUUUGGGUACUUGAGGAGAGGUUGUCUGUGAUGAGUUCGCUUUAUUAAUGACGAAUAUAACACAGAUGGCCUGUUUUCAGUACCACCCACCCCUCGUGC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0019334
          miRBase ID: oar-miR-410-3p
          Mature miRNA Sequence: AAUAUAACACAGAUGGCCUGU
          Chromosome Location: NA


          Back To Top

          More Information


          Associated Megaplex™ RT Primer Pools:

          Megaplex™ RT Primers, Human Pool A
          Megaplex™ RT Primers, Rodent Pool A




          Related Products

          TaqMan™ Pri-miRNA Assay : Mm03307442_pri
          TaqMan™ Pri-miRNA Assay : Hs03303788_pri
          TaqMan™ Pri-miRNA Assay : Rn03465409_pri
          Ambion® Anti-miR™ miRNA Inhibitor : AM11119
          Ambion® Pre-miR™ miRNA Precursor : PM11119
          mirVana® miRNA inhibitor : MH11119
          mirVana® miRNA mimic : MC11119
          TaqMan™ Advanced miRNA Assay : rno481156_mir
          TaqMan™ Advanced miRNA Assay : 478085_mir
          TaqMan™ Advanced miRNA Assay : mmu481156_mir
          MIRNA : TaqMan® OpenArray Human MicroRNA Panel
          miRNA Card : TaqMan® Rodent MicroRNA A Cards v2.0
          FGMIR : TaqManTM Human MicroRNA Array A
          MIRNA : TaqMan® Array Rodent MicroRNA A+B Cards Set v3.0
          FGMIR : QS TaqMan OpenArray Rod miRNA Plate
          MIRNA : TaqMan® Array Rodent MicroRNA A Card v2.0
          MIRNA : TaqMan® Array Human MicroRNA A Cards v2.0
          MIRNA : TaqMan® Array Human MicroRNA A+B Cards Set v3.0
          FGMIR : QS TaqMan OpenArray Hu miRNA Plate
          miRNA Pool : Megaplex™ RT Primers, Human Pool A
          miRNA Card : TaqMan® Human MicroRNA A Cards v2.0
          MIRNA : TaqMan® OpenArray Rodent MicroRNA Panel
          FGMIR : TaqManTM Rodent MicroRNA Array A
          miRNA Pool : Megaplex™ RT Primers, Rodent Pool A


          miRBase Alias:

          Human: hsa-miR-410 (v19)
          Mouse: mmu-miR-410 (v17)
          Rat: rno-miR-410 (v18)
          Macaca mulatta: mml-miR-410 (v19)

          Back To Top

          Related Products

          • TaqMan® Universal Master Mix II, with UNG
          • TaqMan® Universal Master Mix II, no UNG
          • TaqMan® MicroRNA Reverse Transcription Kit
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-vzwvz:80/100.66.79.173:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline