Hamburger Menu Button
Thermo Fisher Scientific Logo
登入
還沒有帳戶嗎 ? 立即註冊
  • 產品
    • Antibodies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Greener Products
    • 查看所有產品類別
  • 應用
    • 生物製程
    • 細胞培養和轉染
    • 食品和飲料
    • 實驗室解決方案
    • 製藥和生物製藥
    • 即時定量PCR
    • 半導體分析
    • 臨床和診斷
    • 數位解決方案
    • 分子光譜、同位素與元素分析
    • 查看所有應用與技術
  • 服務
    • 360° CDMO 和 CRO 服務
    • CDMO 服务
    • 临床试验 CRO 服务
    • 客製化服務
    • 金融與租賃服務
    • 儀器服務
    • 實驗室資訊學
    • 授權與商業供應
    • 培訓服務
    • 查看所有服務
  • 支援
    • 線上訂購協助
    • 數位解決方案
    • 產品支持
    • 技術相關資訊
    • 培訓與教育
    • 查看所有協助與支援
  • 熱門商品
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • 我們的服務對象
    • 生物技術
    • 生物製藥
    • CDMO
    • 實驗室診斷
    • 工業與應用科學
  • 特價商品
  • 聯絡我們
  • 快速訂購
  • 訂單狀態與貨物追蹤
  • 文件和證書
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • 訂單查詢
          • 快速訂購
          • 登入
            登入
            還沒有帳戶嗎 ? 立即註冊
            • 帳戶
            • 訂單
            • Connect: Lab, Data, Apps
            • 客製化產品與項目
            • 儀器管理
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › MC11119
          Assay Name: hsa-miR-410-3p
          miRBase Accession Number: MI0002465
          miRBase Version: v22.1
          Chromosome Location: Chr.14: 101065912 - 101065991 [+] on Build GRCh38
          Mature miRNA Sequence: AAUAUAACACAGAUGGCCUGU
          Species: Human, Mouse, Rat, Bovine, Callithrix jacchus, Cricetulus griseus, Dasypus novemcinctus, Dog, Goat, Guinea pig, Horse, Microcebus murinus, Nomascus leucogenys, Otolemur garnettii, Pan paniscus, Pan troglodytes, Pongo pygmaeus, Pteropus alecto, Rabbit, Rhesus monkey, Sheep
          Product Type: mirVana® miRNA mimic
          Assay ID MC11119

          Purification
          Size
          Availability Made To Order
          Catalog # 4464066
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Family ID:

          MIPF0000018, mir-154

          Mature miRNA Sequence:

          AAUAUAACACAGAUGGCCUGU

          miRBase Details:



          Species miRBase ID miRBase Accession Number
          Human hsa-miR-410-3p MIMAT0002171
          Mouse mmu-miR-410-3p MIMAT0001091
          Rat rno-miR-410-3p MIMAT0005311
          Bovine bta-miR-410 MIMAT0009311
          Callithrix jacchus cja-miR-410 MIMAT0039354
          Cricetulus griseus cgr-miR-410-3p MIMAT0023953
          View More Rows
          Dasypus novemcinctus dno-miR-410-3p MIMAT0047873
          Dog cfa-miR-410 MIMAT0006728
          Goat chi-miR-410-3p MIMAT0036212
          Guinea pig cpo-miR-410-3p MIMAT0047236
          Horse eca-miR-410 MIMAT0013153
          Microcebus murinus mmr-miR-410 MIMAT0049189
          Nomascus leucogenys nle-miR-410 MIMAT0049369
          Otolemur garnettii oga-miR-410 MIMAT0049673
          Pan paniscus ppa-miR-410 MIMAT0049033
          Pan troglodytes ptr-miR-410 MIMAT0008137
          Pongo pygmaeus ppy-miR-410 MIMAT0015877
          Pteropus alecto pal-miR-410-3p MIMAT0040152
          Rabbit ocu-miR-410-3p MIMAT0048436
          Rhesus monkey mml-miR-410-3p MIMAT0006316
          Sheep oar-miR-410-3p MIMAT0019334

          Stem-loop Details



          Human

          Stem-loop ID hsa-mir-410
          Stem-loop Accession # MI0002465
          Stem-loop Sequence
          GGUACCUGAGAAGAGGUUGUCUGUGAUGAGUUCGCUUUUAUUAAUGACGAAUAUAACACAGAUGGCCUGUUUUCAGUACC
          Chromosome Location Chr. 14 - 101065912 - 101065991 [+] on Build GRCh38
          Mature MicroRNA miRBase Accession #: MIMAT0002171
          miRBase ID: hsa-miR-410-3p
          Mature miRNA Sequence: AAUAUAACACAGAUGGCCUGU
          Chromosome Location: Chr. 14 - 101065912 - 101065991 [+] on Build GRCh38

          Mouse

          Stem-loop ID mmu-mir-410
          Stem-loop Accession # MI0001161
          Stem-loop Sequence
          GGGUACUUGAGGAGAGGUUGUCUGUGAUGAGUUCGCUUUAUUAAUGACGAAUAUAACACAGAUGGCCUGUUUUCAAUACCA
          Chromosome Location Chr. 12 - 109743715 - 109743795 [+] on Build GRCm38
          Mature MicroRNA miRBase Accession #: MIMAT0001091
          miRBase ID: mmu-miR-410-3p
          Mature miRNA Sequence: AAUAUAACACAGAUGGCCUGU
          Chromosome Location: Chr. 12 - 109743715 - 109743795 [+] on Build GRCm38

          Rat

          Stem-loop ID rno-mir-410
          Stem-loop Accession # MI0006143
          Stem-loop Sequence
          ACUUGAGGAGAGGUUGUCUGUGAUGAGUUCGCUUUAUUAAUGACGAAUAUAACACAGAUGGCCUGUUUUCAAUACC
          Chromosome Location Chr. 6 - 133893997 - 133894072 [+] on Build Rnor_6.0
          Mature MicroRNA miRBase Accession #: MIMAT0005311
          miRBase ID: rno-miR-410-3p
          Mature miRNA Sequence: AAUAUAACACAGAUGGCCUGU
          Chromosome Location: Chr. 6 - 133893997 - 133894072 [+] on Build Rnor_6.0

          Bovine

          Stem-loop ID bta-mir-410
          Stem-loop Accession # MI0009825
          Stem-loop Sequence
          GGGUACUUGAGGAGAGGUUGUCUGUGAUGAGUUCGCUUUAUUAAUGACGAAUAUAACACAGAUGGCCUGUUUUCAGUACCA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0009311
          miRBase ID: bta-miR-410
          Mature miRNA Sequence: AAUAUAACACAGAUGGCCUGU
          Chromosome Location: NA

          Callithrix jacchus

          Stem-loop ID cja-mir-410
          Stem-loop Accession # MI0031868
          Stem-loop Sequence
          GGUACCUGAGGAGAGGUUGUCUGUGAUGAGUUCGCUUUUAUUAAUGACGAAUAUAACACAGAUGGCCUGUUUUCAGUACC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0039354
          miRBase ID: cja-miR-410
          Mature miRNA Sequence: AAUAUAACACAGAUGGCCUGU
          Chromosome Location: NA

          Cricetulus griseus

          Stem-loop ID cgr-mir-410
          Stem-loop Accession # MI0020521
          Stem-loop Sequence
          CUUGAGGAGAGGUUGUCUGUGAUGAGUUCGCUUUAUUAAUGACGAAUAUAACACAGAUGGCCUGUUUUCAAUACC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0023953
          miRBase ID: cgr-miR-410-3p
          Mature miRNA Sequence: AAUAUAACACAGAUGGCCUGU
          Chromosome Location: NA

          Dasypus novemcinctus

          Stem-loop ID dno-mir-410
          Stem-loop Accession # MI0039101
          Stem-loop Sequence
          AGGCUGUCUGUGAUGAGUUCGCUUUAUUAAUGACGAAUAUAACACAGAUGGCCUGU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0047873
          miRBase ID: dno-miR-410-3p
          Mature miRNA Sequence: AAUAUAACACAGAUGGCCUGU
          Chromosome Location: NA

          Dog

          Stem-loop ID cfa-mir-410
          Stem-loop Accession # MI0008149
          Stem-loop Sequence
          AGGUUGUCUGUGAUGAGUUCGCUUUAUUAAUGCCGAAUAUAACACAGAUGGCCUGU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0006728
          miRBase ID: cfa-miR-410
          Mature miRNA Sequence: AAUAUAACACAGAUGGCCUGU
          Chromosome Location: NA

          Goat

          Stem-loop ID chi-mir-410
          Stem-loop Accession # MI0030783
          Stem-loop Sequence
          CACCUGCGGUCUGUGGAGUAGGCGCCACUUUGGGUACUUGAGGAGAGGUUGUCUGUGAUGAGUUCGCUUUAUUAAUGACGAAUAUAACACAGAUGGCCUGUUUUCAGUACCACCCACCCCUCGUG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0036212
          miRBase ID: chi-miR-410-3p
          Mature miRNA Sequence: AAUAUAACACAGAUGGCCUGU
          Chromosome Location: NA

          Guinea pig

          Stem-loop ID cpo-mir-410
          Stem-loop Accession # MI0038766
          Stem-loop Sequence
          AGGUUGUCUGUGAUGAGUUCGCUUUAUUAAUGACGAAUAUAACACAGAUGGCCUGU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0047236
          miRBase ID: cpo-miR-410-3p
          Mature miRNA Sequence: AAUAUAACACAGAUGGCCUGU
          Chromosome Location: NA

          Horse

          Stem-loop ID eca-mir-410
          Stem-loop Accession # MI0012897
          Stem-loop Sequence
          UGAGGAGAGGUUGUCUGUGAUGAGUUCGCUUUAUUAAUGACGAAUAUAACACAGAUGGCCUGUCUUCAGUACCA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0013153
          miRBase ID: eca-miR-410
          Mature miRNA Sequence: AAUAUAACACAGAUGGCCUGU
          Chromosome Location: NA

          Microcebus murinus

          Stem-loop ID mmr-mir-410
          Stem-loop Accession # MI0039908
          Stem-loop Sequence
          AGGUUGUCUGUGAUGAGUUCGCUUUAUUAAUGACGAAUAUAACACAGAUGGCCUGU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0049189
          miRBase ID: mmr-miR-410
          Mature miRNA Sequence: AAUAUAACACAGAUGGCCUGU
          Chromosome Location: NA

          Nomascus leucogenys

          Stem-loop ID nle-mir-410
          Stem-loop Accession # MI0040106
          Stem-loop Sequence
          AGGUUGUCUGUGAUGAGUUCGCUUUUAUUAAUGACGAAUAUAACACAGAUGGCCUGU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0049369
          miRBase ID: nle-miR-410
          Mature miRNA Sequence: AAUAUAACACAGAUGGCCUGU
          Chromosome Location: NA

          Otolemur garnettii

          Stem-loop ID oga-mir-410
          Stem-loop Accession # MI0040456
          Stem-loop Sequence
          AGGUUGUCUGUGAUGAGUUCGCUUUAUUAAUGACGAAUAUAACACAGAUGGCCUGU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0049673
          miRBase ID: oga-miR-410
          Mature miRNA Sequence: AAUAUAACACAGAUGGCCUGU
          Chromosome Location: NA

          Pan paniscus

          Stem-loop ID ppa-mir-410
          Stem-loop Accession # MI0039741
          Stem-loop Sequence
          AGGUUGUCUGUGAUGAGUUCGCUUUUAUUAAUGACGAAUAUAACACAGAUGGCCUGU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0049033
          miRBase ID: ppa-miR-410
          Mature miRNA Sequence: AAUAUAACACAGAUGGCCUGU
          Chromosome Location: NA

          Pan troglodytes

          Stem-loop ID ptr-mir-410
          Stem-loop Accession # MI0008658
          Stem-loop Sequence
          GUACCUGAGAAGAGGUUGUCUGUGAUGAGUUCGCUUUUAUUAAUGACGAAUAUAACACAGAUGGCCUGUUUUCAGUACC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0008137
          miRBase ID: ptr-miR-410
          Mature miRNA Sequence: AAUAUAACACAGAUGGCCUGU
          Chromosome Location: NA

          Pongo pygmaeus

          Stem-loop ID ppy-mir-410
          Stem-loop Accession # MI0014934
          Stem-loop Sequence
          GGUACCUGAGAAGAGGUUGUCUGUGAUGAGUUCGCUUUUAUUAAUGACGAAUAUAACACAGAUGGCCUGUUUUCAGUACC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0015877
          miRBase ID: ppy-miR-410
          Mature miRNA Sequence: AAUAUAACACAGAUGGCCUGU
          Chromosome Location: NA

          Pteropus alecto

          Stem-loop ID pal-mir-410
          Stem-loop Accession # MI0032520
          Stem-loop Sequence
          AGGUUGUCUGUGAUGAGUUCGCUUUAUUAAUGACGAAUAUAACACAGAUGGCCUGU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0040152
          miRBase ID: pal-miR-410-3p
          Mature miRNA Sequence: AAUAUAACACAGAUGGCCUGU
          Chromosome Location: NA

          Rabbit

          Stem-loop ID ocu-mir-410
          Stem-loop Accession # MI0039399
          Stem-loop Sequence
          AGGUUGUCUGUGAUGAGUUCGCUGUAUUAAAGCCGAAUAUAACACAGAUGGCCUGU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0048436
          miRBase ID: ocu-miR-410-3p
          Mature miRNA Sequence: AAUAUAACACAGAUGGCCUGU
          Chromosome Location: NA

          Rhesus monkey

          Stem-loop ID mml-mir-410
          Stem-loop Accession # MI0007734
          Stem-loop Sequence
          GGUACCUGAGGAGAGGUUGUCUGUGAUGAGUUCGCUUUUAUUAAUGACGAAUAUAACACAGAUGGCCUGUUUUCAGUACC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0006316
          miRBase ID: mml-miR-410-3p
          Mature miRNA Sequence: AAUAUAACACAGAUGGCCUGU
          Chromosome Location: NA

          Sheep

          Stem-loop ID oar-mir-410
          Stem-loop Accession # MI0016961
          Stem-loop Sequence
          CACCUGCGGUCUGUGGAGUAGGCGCCACUUUGGGUACUUGAGGAGAGGUUGUCUGUGAUGAGUUCGCUUUAUUAAUGACGAAUAUAACACAGAUGGCCUGUUUUCAGUACCACCCACCCCUCGUGC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0019334
          miRBase ID: oar-miR-410-3p
          Mature miRNA Sequence: AAUAUAACACAGAUGGCCUGU
          Chromosome Location: NA


          Back To Top

          More Information




          Related Products

          TaqMan™ Pri-miRNA Assay : Mm03307442_pri
          TaqMan™ Pri-miRNA Assay : Hs03303788_pri
          TaqMan™ Pri-miRNA Assay : Rn03465409_pri
          Ambion® Anti-miR™ miRNA Inhibitor : AM11119
          Ambion® Pre-miR™ miRNA Precursor : PM11119
          TaqMan™ MicroRNA Assay : 001274
          mirVana® miRNA inhibitor : MH11119
          TaqMan™ Advanced miRNA Assay : rno481156_mir
          TaqMan™ Advanced miRNA Assay : 478085_mir
          TaqMan™ Advanced miRNA Assay : mmu481156_mir


          miRBase Alias:

          Human: hsa-miR-410 (v19)
          Mouse: mmu-miR-410 (v17)
          Rat: rno-miR-410 (v18)
          Macaca mulatta: mml-miR-410 (v19)

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Neon™ Transfection System
          產品訂購 Plus Icon Minus Icon
          • 訂單狀態
          • 訂購協助
          • 快速訂購
          • 現貨供應中心
          • 電子採購
          支持 Plus Icon Minus Icon
          • 協助與支持
          • 聯絡我們
          • 技術支持中心
          • 搜尋文件和證書
          • 回報網站問題
          資源 Plus Icon Minus Icon
          • 學習中心
          • 促銷活動
          • 活動和網路研討會
          • 社群媒體
          關於 Thermo Fisher Plus Icon Minus Icon
          • 關於我們 關於我們
          • 人才招募 人才招募
          • 投資者資訊 投資者資訊
          • 新聞 新聞
          • 企業社會責任 企業社會責任
          • 商標
          • 政策和聲明
          品牌 Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-b2k9d:80/100.66.79.173:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline