Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › MC19960
          Assay Name: hsa-miR-410-5p
          miRBase Accession Number: MI0002465
          miRBase Version: v22.1
          Chromosome Location: Chr.14: 101065912 - 101065991 [+] on Build GRCh38
          Mature miRNA Sequence: AGGUUGUCUGUGAUGAGUUCG
          Species: Human, Mouse, Rat, Cricetulus griseus, Guinea pig, Pteropus alecto, Rabbit, Rhesus monkey
          Product Type: mirVana® miRNA mimic
          Assay ID MC19960

          Purification
          Size
          Availability Made To Order
          Catalog # 4464066
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Family ID:

          MIPF0000018, mir-154

          Mature miRNA Sequence:

          AGGUUGUCUGUGAUGAGUUCG

          miRBase Details:



          Species miRBase ID miRBase Accession Number
          Human hsa-miR-410-5p MIMAT0026558
          Mouse mmu-miR-410-5p MIMAT0017172
          Rat rno-miR-410-5p MIMAT0017303
          Cricetulus griseus cgr-miR-410-5p MIMAT0023952
          Guinea pig cpo-miR-410-5p MIMAT0047235
          Pteropus alecto pal-miR-410-5p MIMAT0040151
          View More Rows
          Rabbit ocu-miR-410-5p MIMAT0048435
          Rhesus monkey mml-miR-410-5p MIMAT0026869

          Stem-loop Details



          Human

          Stem-loop ID hsa-mir-410
          Stem-loop Accession # MI0002465
          Stem-loop Sequence
          GGUACCUGAGAAGAGGUUGUCUGUGAUGAGUUCGCUUUUAUUAAUGACGAAUAUAACACAGAUGGCCUGUUUUCAGUACC
          Chromosome Location Chr. 14 - 101065912 - 101065991 [+] on Build GRCh38
          Mature MicroRNA miRBase Accession #: MIMAT0026558
          miRBase ID: hsa-miR-410-5p
          Mature miRNA Sequence: AGGUUGUCUGUGAUGAGUUCG
          Chromosome Location: Chr. 14 - 101065912 - 101065991 [+] on Build GRCh38

          Mouse

          Stem-loop ID mmu-mir-410
          Stem-loop Accession # MI0001161
          Stem-loop Sequence
          GGGUACUUGAGGAGAGGUUGUCUGUGAUGAGUUCGCUUUAUUAAUGACGAAUAUAACACAGAUGGCCUGUUUUCAAUACCA
          Chromosome Location Chr. 12 - 109743715 - 109743795 [+] on Build GRCm38
          Mature MicroRNA miRBase Accession #: MIMAT0017172
          miRBase ID: mmu-miR-410-5p
          Mature miRNA Sequence: AGGUUGUCUGUGAUGAGUUCG
          Chromosome Location: Chr. 12 - 109743715 - 109743795 [+] on Build GRCm38

          Rat

          Stem-loop ID rno-mir-410
          Stem-loop Accession # MI0006143
          Stem-loop Sequence
          ACUUGAGGAGAGGUUGUCUGUGAUGAGUUCGCUUUAUUAAUGACGAAUAUAACACAGAUGGCCUGUUUUCAAUACC
          Chromosome Location Chr. 6 - 133893997 - 133894072 [+] on Build Rnor_6.0
          Mature MicroRNA miRBase Accession #: MIMAT0017303
          miRBase ID: rno-miR-410-5p
          Mature miRNA Sequence: AGGUUGUCUGUGAUGAGUUCG
          Chromosome Location: Chr. 6 - 133893997 - 133894072 [+] on Build Rnor_6.0

          Cricetulus griseus

          Stem-loop ID cgr-mir-410
          Stem-loop Accession # MI0020521
          Stem-loop Sequence
          CUUGAGGAGAGGUUGUCUGUGAUGAGUUCGCUUUAUUAAUGACGAAUAUAACACAGAUGGCCUGUUUUCAAUACC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0023952
          miRBase ID: cgr-miR-410-5p
          Mature miRNA Sequence: AGGUUGUCUGUGAUGAGUUCG
          Chromosome Location: NA

          Guinea pig

          Stem-loop ID cpo-mir-410
          Stem-loop Accession # MI0038766
          Stem-loop Sequence
          AGGUUGUCUGUGAUGAGUUCGCUUUAUUAAUGACGAAUAUAACACAGAUGGCCUGU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0047235
          miRBase ID: cpo-miR-410-5p
          Mature miRNA Sequence: AGGUUGUCUGUGAUGAGUUCG
          Chromosome Location: NA

          Pteropus alecto

          Stem-loop ID pal-mir-410
          Stem-loop Accession # MI0032520
          Stem-loop Sequence
          AGGUUGUCUGUGAUGAGUUCGCUUUAUUAAUGACGAAUAUAACACAGAUGGCCUGU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0040151
          miRBase ID: pal-miR-410-5p
          Mature miRNA Sequence: AGGUUGUCUGUGAUGAGUUCG
          Chromosome Location: NA

          Rabbit

          Stem-loop ID ocu-mir-410
          Stem-loop Accession # MI0039399
          Stem-loop Sequence
          AGGUUGUCUGUGAUGAGUUCGCUGUAUUAAAGCCGAAUAUAACACAGAUGGCCUGU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0048435
          miRBase ID: ocu-miR-410-5p
          Mature miRNA Sequence: AGGUUGUCUGUGAUGAGUUCG
          Chromosome Location: NA

          Rhesus monkey

          Stem-loop ID mml-mir-410
          Stem-loop Accession # MI0007734
          Stem-loop Sequence
          GGUACCUGAGGAGAGGUUGUCUGUGAUGAGUUCGCUUUUAUUAAUGACGAAUAUAACACAGAUGGCCUGUUUUCAGUACC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0026869
          miRBase ID: mml-miR-410-5p
          Mature miRNA Sequence: AGGUUGUCUGUGAUGAGUUCG
          Chromosome Location: NA


          Back To Top

          More Information




          Related Products

          TaqMan™ Pri-miRNA Assay : Mm03307442_pri
          TaqMan™ Pri-miRNA Assay : Hs03303788_pri
          TaqMan™ Pri-miRNA Assay : Rn03465409_pri
          Ambion® Anti-miR™ miRNA Inhibitor : AM19960
          Ambion® Pre-miR™ miRNA Precursor : PM19960
          TaqMan™ MicroRNA Assay : 464693_mat
          mirVana® miRNA inhibitor : MH19960
          TaqMan™ Advanced miRNA Assay : rno481391_mir
          TaqMan™ Advanced miRNA Assay : mmu482760_mir
          TaqMan™ Advanced miRNA Assay : 478873_mir


          miRBase Alias:

          Mouse: mmu-miR-410* (v17)
          Rat: rno-miR-410* (v18)

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Spain flag icon
          Spain

          TEST

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-zsnhf:80/100.66.75.14:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline