Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Clinical Genomics
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
Gene:
YAP1
Transcript:
Assay Name:
Mutation genome location:
chr.11 102110355-102110855 on build GRCh38
Assay ID
Hs00111251_CE
Purification
DS
HPLC
Size
25 nmol
25 nmol
50 nmol
50 nmol
25 nmol
25 nmol
50 nmol
50 nmol
25 nmol
25 nmol
50 nmol
50 nmol
25 nmol
25 nmol
50 nmol
50 nmol
Availability
Made To Order
Catalog #
A15629
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
Assay Details
More Information
Genomic Map
LOADING...
×
Back To Top
Assay Details
Back To Top
More Information
Set Membership:
Between 400-700 bp
Panther Classification:
Molecular Function -
transcription cofactor
Gene Ontology Categories:
Function(s)
Process(es)
cell morphogenesis
vasculogenesis
embryonic heart tube morphogenesis
transcription initiation from RNA polymerase II promoter
cellular response to DNA damage stimulus
cell proliferation
positive regulation of cell proliferation
regulation of keratinocyte proliferation
keratinocyte differentiation
negative regulation of epithelial cell differentiation
notochord development
somatic stem cell population maintenance
hippo signaling
positive regulation of transcription from RNA polymerase II promoter
positive regulation of organ growth
paraxial mesoderm development
lateral mesoderm development
regulation of neurogenesis
contact inhibition
bud elongation involved in lung branching
lung epithelial cell differentiation
cellular response to retinoic acid
cellular response to gamma radiation
regulation of stem cell proliferation
regulation of metanephric nephron tubule epithelial cell differentiation
positive regulation of canonical Wnt signaling pathway
positive regulation of stem cell population maintenance
negative regulation of nucleic acid-templated transcription
negative regulation of stem cell differentiation
negative regulation of extrinsic apoptotic signaling pathway
RNA polymerase II core promoter sequence-specific DNA binding
transcription factor activity, RNA polymerase II transcription factor binding
chromatin binding
transcription coactivator activity
transcription corepressor activity
protein binding
protein C-terminus binding
transcription regulatory region DNA binding
proline-rich region binding
Back To Top
Related Products
POP-6
™
Polymer
POP-7
™
Polymer
BigDye
®
Terminator v1.1 Cycle Sequencing Kit
BigDye
®
Terminator v3.1 Cycle Sequencing Kit
BigDye
®
Direct Cycle Sequencing Kit
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-b49b87d85-8dsbr:80/100.66.76.150:80.
git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.47.0-Offline