Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture and Transfection
Chemicals
Chromatography
Electron Microscopes
Lab Plasticware and Supplies
Lab Centrifuges
Lab Solutions
Mass Spectrometers
Next Generation Sequencers
See all product categories
Applications
Brands
Industrial and Applied Sciences
Food and Beverage
Forensics
Lab Solutions
Life Sciences
Pharma and Biopharma
Biotechnology
Clinical and Diagnostics
Digital Solutions
See all applications
Services
Lab Instrument and Equipment Services
Custom Services
Training Services
Financial and Leasing Services
Enterprise Level Lab Informatics
Partnering and Licensing Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Cell Biology Services
Food Safety Inspection Services
See all services
Help and Support
Contact Us
Product Documentation
Knowledge Base and Product FAQs
Learning Centers
Supply Center
eProcurement Solutions
Lab Instrument and Equipment Support
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Special Offers
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
Gene:
ATM
Transcript:
NM_000051.3
Assay Name:
Mutation genome location:
chr.11 108304405-108304937 on build GRCh38
Assay ID
Hs00111959_CE
Purification
DS
HPLC
Size
25 nmol
25 nmol
50 nmol
50 nmol
25 nmol
25 nmol
50 nmol
50 nmol
25 nmol
25 nmol
50 nmol
50 nmol
25 nmol
25 nmol
50 nmol
50 nmol
Availability
Made To Order
Catalog #
A15629
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
Assay Details
More Information
Genomic Map
LOADING...
×
Back To Top
Assay Details
Back To Top
More Information
Set Membership:
Between 400-700 bp
Panther Classification:
Molecular Function -
non-receptor serine/threonine protein kinase
Gene Ontology Categories:
Function(s)
Process(es)
double-strand break repair via homologous recombination
DNA double-strand break processing
DNA synthesis involved in DNA repair
strand displacement
response to hypoxia
somitogenesis
pre-B cell allelic exclusion
DNA replication
DNA repair
double-strand break repair via nonhomologous end joining
protein phosphorylation
cellular response to DNA damage stimulus
DNA damage induced protein phosphorylation
DNA damage response, signal transduction by p53 class mediator resulting in cell cycle arrest
telomere maintenance via telomerase
cell cycle arrest
mitotic spindle assembly checkpoint
reciprocal meiotic recombination
signal transduction
brain development
heart development
determination of adult lifespan
intrinsic apoptotic signaling pathway in response to DNA damage
response to ionizing radiation
regulation of autophagy
histone phosphorylation
peptidyl-serine phosphorylation
negative regulation of B cell proliferation
positive regulation of telomere maintenance via telomerase
positive regulation of histone phosphorylation
V(D)J recombination
phosphatidylinositol-3-phosphate biosynthetic process
peptidyl-serine autophosphorylation
lipoprotein catabolic process
regulation of apoptotic process
positive regulation of apoptotic process
positive regulation of DNA damage response, signal transduction by p53 class mediator
positive regulation of neuron apoptotic process
meiotic telomere clustering
protein autophosphorylation
oocyte development
neuron apoptotic process
regulation of telomerase activity
histone mRNA catabolic process
cellular response to gamma radiation
cellular response to X-ray
cellular response to nitrosative stress
signal transduction involved in mitotic G2 DNA damage checkpoint
replicative senescence
establishment of RNA localization to telomere
establishment of macromolecular complex localization to telomere
regulation of cellular response to heat
regulation of signal transduction by p53 class mediator
negative regulation of TORC1 signaling
negative regulation of telomere capping
positive regulation of telomere maintenance via telomere lengthening
positive regulation of telomerase catalytic core complex assembly
DNA binding
protein serine/threonine kinase activity
DNA-dependent protein kinase activity
protein binding
ATP binding
1-phosphatidylinositol-3-kinase activity
protein complex binding
protein dimerization activity
protein N-terminus binding
Back To Top
Related Products
POP-6
™
Polymer
POP-7
™
Polymer
BigDye
®
Terminator v1.1 Cycle Sequencing Kit
BigDye
®
Terminator v3.1 Cycle Sequencing Kit
BigDye
®
Direct Cycle Sequencing Kit
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-67655d8755-zsnhf:80/100.66.75.14:80.
git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.46.0-Offline