Hamburger Menu Button
Thermo Fisher Scientific Logo
登入
還沒有帳戶嗎 ?
立即註冊
產品
Antibodies
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Cell Culture Media
Chemicals
Western Blot Products
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Greener Products
查看所有產品類別
應用
生物製程
細胞培養和轉染
食品和飲料
實驗室解決方案
製藥和生物製藥
即時定量PCR
半導體分析
臨床和診斷
數位解決方案
分子光譜、同位素與元素分析
查看所有應用與技術
服務
360° CDMO 和 CRO 服務
CDMO 服务
临床试验 CRO 服务
客製化服務
金融與租賃服務
儀器服務
實驗室資訊學
授權與商業供應
培訓服務
查看所有服務
支援
線上訂購協助
數位解決方案
產品支持
技術相關資訊
培訓與教育
查看所有協助與支援
熱門商品
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
我們的服務對象
生物技術
生物製藥
CDMO
實驗室診斷
工業與應用科學
特價商品
聯絡我們
快速訂購
訂單狀態與貨物追蹤
文件和證書
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
訂單查詢
快速訂購
登入
登入
還沒有帳戶嗎 ?
立即註冊
帳戶
訂單
Connect: Lab, Data, Apps
客製化產品與項目
儀器管理
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
Gene:
ATM
Transcript:
NM_000051.3
Assay Name:
Mutation genome location:
chr.11 108332646-108333180 on build GRCh38
Gene:
C11orf65
Transcript:
NM_000051.3
Assay Name:
Mutation genome location:
chr.11 108332646-108333180 on build GRCh38
Assay ID
Hs00111977_CE
Purification
DS
HPLC
Size
25 nmol
25 nmol
50 nmol
50 nmol
25 nmol
25 nmol
50 nmol
50 nmol
25 nmol
25 nmol
50 nmol
50 nmol
25 nmol
25 nmol
50 nmol
50 nmol
Availability
Made To Order
Catalog #
A15629
Price
Your Price
Online offer:
Check your price ›
新增到購物車
Genomic Map
Assay Details
More Information
Genomic Map
LOADING...
×
Back To Top
Assay Details
Back To Top
More Information
Set Membership:
Between 400-700 bp
Panther Classification:
Molecular Function -
non-receptor serine/threonine protein kinase
dehydrogenase
Gene Ontology Categories:
Function(s)
Process(es)
double-strand break repair via homologous recombination
DNA double-strand break processing
DNA synthesis involved in DNA repair
strand displacement
response to hypoxia
somitogenesis
pre-B cell allelic exclusion
DNA replication
DNA repair
double-strand break repair via nonhomologous end joining
protein phosphorylation
cellular response to DNA damage stimulus
DNA damage induced protein phosphorylation
DNA damage response, signal transduction by p53 class mediator resulting in cell cycle arrest
telomere maintenance via telomerase
cell cycle arrest
mitotic spindle assembly checkpoint
reciprocal meiotic recombination
signal transduction
brain development
heart development
determination of adult lifespan
intrinsic apoptotic signaling pathway in response to DNA damage
response to ionizing radiation
regulation of autophagy
histone phosphorylation
peptidyl-serine phosphorylation
negative regulation of B cell proliferation
positive regulation of telomere maintenance via telomerase
positive regulation of histone phosphorylation
V(D)J recombination
phosphatidylinositol-3-phosphate biosynthetic process
peptidyl-serine autophosphorylation
lipoprotein catabolic process
regulation of apoptotic process
positive regulation of apoptotic process
positive regulation of DNA damage response, signal transduction by p53 class mediator
positive regulation of neuron apoptotic process
meiotic telomere clustering
protein autophosphorylation
oocyte development
neuron apoptotic process
regulation of telomerase activity
histone mRNA catabolic process
cellular response to gamma radiation
cellular response to X-ray
cellular response to nitrosative stress
signal transduction involved in mitotic G2 DNA damage checkpoint
replicative senescence
establishment of RNA localization to telomere
establishment of macromolecular complex localization to telomere
regulation of cellular response to heat
regulation of signal transduction by p53 class mediator
negative regulation of TORC1 signaling
negative regulation of telomere capping
positive regulation of telomere maintenance via telomere lengthening
positive regulation of telomerase catalytic core complex assembly
DNA binding
protein serine/threonine kinase activity
DNA-dependent protein kinase activity
protein binding
ATP binding
1-phosphatidylinositol-3-kinase activity
protein complex binding
protein dimerization activity
protein N-terminus binding
Back To Top
Related Products
POP-6
™
Polymer
POP-7
™
Polymer
BigDye
®
Terminator v1.1 Cycle Sequencing Kit
BigDye
®
Terminator v3.1 Cycle Sequencing Kit
BigDye
®
Direct Cycle Sequencing Kit
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-b49b87d85-n9r8p:80/100.66.76.150:80.
git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.47.0-Offline