Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Cell Analysis
Antibodies
Mass Spectrometry
Cell Culture
Laboratory Instruments
Clinical and Diagnostics
Chromatography
Laboratory Equipment
Laboratory Supplies
Molecular Biology and Nucleic Acid Analysis
Sequence-Specific Nucleic Acid Products
See all product categories
Applications
Cell Culture and Transfection
Flow Cytometry
Cancer Research
Chromatography
Sequencing
PCR
Lab Solutions
Allergy Diagnostics
See all applications and techniques
Services
Cell Biology Services
Custom Services
Training Services
Lab Informatics Services
Financial and Leasing Services
Partnering and Licensing Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Food Safety Inspection Services
See all services
Help and Support
How to Order
Promotions and Online Offers
Contact Us
Change Location
Create a New Account
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Special Offers
Contact Us
Quick Order
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Contact Us
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
Gene:
DRD2
Transcript:
Assay Name:
Mutation genome location:
chr.11 113409895-113410397 on build GRCh38
Assay ID
Hs00112551_CE
Purification
DS
HPLC
Size
25 nmol
25 nmol
50 nmol
50 nmol
25 nmol
25 nmol
50 nmol
50 nmol
25 nmol
25 nmol
50 nmol
50 nmol
25 nmol
25 nmol
50 nmol
50 nmol
Availability
Made To Order
Catalog #
A15629
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
Assay Details
More Information
Genomic Map
LOADING...
×
Back To Top
Assay Details
Back To Top
More Information
Set Membership:
Between 400-700 bp
Panther Classification:
Molecular Function -
G-protein coupled receptor
Gene Ontology Categories:
Function(s)
Process(es)
temperature homeostasis
response to hypoxia
synaptic transmission, dopaminergic
response to amphetamine
neurological system process involved in regulation of systemic arterial blood pressure
regulation of heart rate
regulation of sodium ion transport
G-protein coupled receptor internalization
positive regulation of neuroblast proliferation
positive regulation of receptor internalization
negative regulation of adenylate cyclase activity
adenylate cyclase-inhibiting dopamine receptor signaling pathway
neuron-neuron synaptic transmission
axonogenesis
synapse assembly
sensory perception of smell
long-term memory
grooming behavior
locomotory behavior
adult walking behavior
feeding behavior
protein localization
negative regulation of cell proliferation
associative learning
visual learning
response to light stimulus
response to toxic substance
response to iron ion
regulation of dopamine secretion
response to inactivity
Wnt signaling pathway
striatum development
orbitofrontal cortex development
cerebral cortex GABAergic interneuron migration
adenohypophysis development
negative regulation of cell migration
peristalsis
regulation of cAMP metabolic process
auditory behavior
activation of protein kinase activity
regulation of synaptic transmission, GABAergic
positive regulation of cytokinesis
circadian regulation of gene expression
negative regulation of dopamine secretion
response to histamine
response to nicotine
positive regulation of urine volume
positive regulation of renal sodium excretion
positive regulation of multicellular organism growth
response to cocaine
negative regulation of circadian sleep/wake cycle, sleep
dopamine metabolic process
response to drug
regulation of potassium ion transport
response to morphine
pigmentation
regulation of phosphoprotein phosphatase activity
positive regulation of G-protein coupled receptor protein signaling pathway
negative regulation of blood pressure
negative regulation of innate immune response
positive regulation of transcription from RNA polymerase II promoter
phosphatidylinositol metabolic process
negative regulation of insulin secretion
acid secretion
behavioral response to cocaine
behavioral response to ethanol
regulation of long-term neuronal synaptic plasticity
response to axon injury
branching morphogenesis of a nerve
arachidonic acid secretion
negative regulation of protein secretion
release of sequestered calcium ion into cytosol
negative regulation of cytosolic calcium ion concentration
regulation of dopamine uptake involved in synaptic transmission
positive regulation of dopamine uptake involved in synaptic transmission
regulation of synapse structural plasticity
negative regulation of protein kinase B signaling
negative regulation of synaptic transmission, glutamatergic
positive regulation of growth hormone secretion
prepulse inhibition
phospholipase C-activating dopamine receptor signaling pathway
negative regulation of dopamine receptor signaling pathway
positive regulation of ERK1 and ERK2 cascade
adenylate cyclase-activating adrenergic receptor signaling pathway
regulation of locomotion involved in locomotory behavior
chemical synaptic transmission, postsynaptic
positive regulation of glial cell-derived neurotrophic factor secretion
positive regulation of long-term synaptic potentiation
negative regulation of voltage-gated calcium channel activity
dopamine neurotransmitter receptor activity, coupled via Gi/Go
adrenergic receptor activity
protein binding
drug binding
dopamine binding
ionotropic glutamate receptor binding
identical protein binding
protein homodimerization activity
protein heterodimerization activity
Back To Top
Related Products
POP-6
™
Polymer
POP-7
™
Polymer
BigDye
®
Terminator v1.1 Cycle Sequencing Kit
BigDye
®
Terminator v3.1 Cycle Sequencing Kit
BigDye
®
Direct Cycle Sequencing Kit
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-b49b87d85-5cvsx:80/100.66.79.163:80.
git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.47.0-Offline