Hamburger Menu Button
Thermo Fisher Scientific Logo
登入
Don't have an account ?
Create Account
Products
Antibodies
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Tubes
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Clinical Genomics
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO 和 CRO 服務
CDMO 服務
臨床試驗 CRO 服務
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
Register for an Account
How to Order
Instrument Support
Technical Support Centers
Learning Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
快速訂購
登入
登入
Don't have an account ?
Create Account
帳戶
訂單
訂製產品與專案
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
Gene:
LOC105369748
Transcript:
NM_006105.5
Assay Name:
Mutation genome location:
chr.12 47738552-47739050 on build GRCh38
Gene:
RAPGEF3
Transcript:
NM_006105.5
Assay Name:
Mutation genome location:
chr.12 47738552-47739050 on build GRCh38
Assay ID
Hs00118557_CE
Purification
DS
HPLC
Size
25 nmol
25 nmol
50 nmol
50 nmol
25 nmol
25 nmol
50 nmol
50 nmol
25 nmol
25 nmol
50 nmol
50 nmol
25 nmol
25 nmol
50 nmol
50 nmol
Availability
Made To Order
Catalog #
A15629
Price
Your Price
Online offer:
Check your price ›
新增到購物車
Genomic Map
Assay Details
More Information
Genomic Map
LOADING...
×
Back To Top
Assay Details
Back To Top
More Information
Set Membership:
Between 400-700 bp
Panther Classification:
Molecular Function -
guanyl-nucleotide exchange factor
Gene Ontology Categories:
Function(s)
Process(es)
angiogenesis
signal transduction
brain development
cell proliferation
negative regulation of cardiac muscle cell apoptotic process
positive regulation of smooth muscle cell migration
cAMP-mediated signaling
spinal cord development
positive regulation of cAMP catabolic process
Rap protein signal transduction
negative regulation of collagen biosynthetic process
positive regulation of peptidyl-serine phosphorylation
negative regulation of syncytium formation by plasma membrane fusion
response to nicotine
cellular response to drug
positive regulation of GTPase activity
positive regulation of angiogenesis
positive regulation of cell size
positive regulation of transcription from RNA polymerase II promoter
positive regulation of protein export from nucleus
regulation of insulin secretion
positive regulation of stress fiber assembly
positive regulation of syncytium formation by plasma membrane fusion
artery development
establishment of endothelial barrier
cellular response to cAMP
cellular response to transforming growth factor beta stimulus
liver regeneration
positive regulation of protein acetylation
positive regulation of GTP binding
positive regulation of calcium ion transmembrane transport
positive regulation of hydrogen:potassium-exchanging ATPase activity
cellular response to L-dopa
dorsal root ganglion development
regulation of actin cytoskeleton reorganization
positive regulation of actin cytoskeleton reorganization
regulation of histone H3-K9 acetylation
guanyl-nucleotide exchange factor activity
protein binding
Rap guanyl-nucleotide exchange factor activity
protein domain specific binding
cAMP binding
ion channel binding
Back To Top
Related Products
POP-6
™
Polymer
POP-7
™
Polymer
BigDye
®
Terminator v1.1 Cycle Sequencing Kit
BigDye
®
Terminator v3.1 Cycle Sequencing Kit
BigDye
®
Direct Cycle Sequencing Kit
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-b49b87d85-9fm2r:80/100.66.79.29:80.
git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.47.0-Offline