Hamburger Menu Button
Thermo Fisher Scientific Logo
登入
Don't have an account ?
Create Account
Products
Antibodies
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Tubes
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Clinical Genomics
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO 和 CRO 服務
CDMO 服務
臨床試驗 CRO 服務
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
Register for an Account
How to Order
Instrument Support
Technical Support Centers
Learning Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
快速訂購
登入
登入
Don't have an account ?
Create Account
帳戶
訂單
訂製產品與專案
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
Gene:
ATP2A2
Transcript:
NM_170665.3
Assay Name:
Mutation genome location:
chr.12 110291620-110292152 on build GRCh38
Assay ID
Hs00127579_CE
Purification
DS
HPLC
Size
25 nmol
25 nmol
50 nmol
50 nmol
25 nmol
25 nmol
50 nmol
50 nmol
25 nmol
25 nmol
50 nmol
50 nmol
25 nmol
25 nmol
50 nmol
50 nmol
Availability
Made To Order
Catalog #
A15629
Price
Your Price
Online offer:
Check your price ›
新增到購物車
Genomic Map
Assay Details
More Information
Genomic Map
LOADING...
×
Back To Top
Assay Details
Back To Top
More Information
Set Membership:
Between 400-700 bp
Panther Classification:
Molecular Function -
primary active transporter
Gene Ontology Categories:
Function(s)
Process(es)
regulation of the force of heart contraction
ER-nucleus signaling pathway
organelle organization
cell adhesion
epidermis development
positive regulation of heart rate
regulation of cardiac muscle contraction by calcium ion signaling
transition between fast and slow fiber
endoplasmic reticulum calcium ion homeostasis
positive regulation of endoplasmic reticulum calcium ion concentration
T-tubule organization
ion transmembrane transport
cellular response to oxidative stress
response to endoplasmic reticulum stress
response to peptide hormone
negative regulation of heart contraction
relaxation of cardiac muscle
sarcoplasmic reticulum calcium ion transport
calcium ion transmembrane transport
regulation of cardiac muscle cell membrane potential
regulation of cardiac muscle cell action potential involved in regulation of contraction
regulation of calcium ion-dependent exocytosis of neurotransmitter
calcium ion transport from cytosol to endoplasmic reticulum
regulation of cardiac conduction
calcium ion import into sarcoplasmic reticulum
calcium-transporting ATPase activity
calcium ion binding
protein binding
ATP binding
protein C-terminus binding
enzyme binding
lutropin-choriogonadotropic hormone receptor binding
S100 protein binding
calcium-transporting ATPase activity involved in regulation of cardiac muscle cell membrane potential
Back To Top
Related Products
POP-6
™
Polymer
POP-7
™
Polymer
BigDye
®
Terminator v1.1 Cycle Sequencing Kit
BigDye
®
Terminator v3.1 Cycle Sequencing Kit
BigDye
®
Direct Cycle Sequencing Kit
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-b49b87d85-n9r8p:80/100.66.76.150:80.
git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.47.0-Offline