Hamburger Menu Button
Thermo Fisher Scientific Logo
登入
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Tubes
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Clinical Genomics
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO 和 CRO 服務
    • CDMO 服務
    • 臨床試驗 CRO 服務
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • Register for an Account
    • How to Order
    • Instrument Support
    • Technical Support Centers
    • Learning Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • 快速訂購
          • 登入
            登入
            Don't have an account ? Create Account
            • 帳戶
            • 訂單
            • 訂製產品與專案
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          Gene:
          HMGB1
          Transcript:
          NM_001313893.1
          Assay Name:
          Mutation genome location:
          chr.13 30463453-30463967 on build GRCh38
          Assay ID Hs00130701_CE

          Purification
          Size
          Availability Made To Order
          Catalog # A15629
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details


          Back To Top

          More Information


          Set Membership:

          Between 400-700 bp

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of transcription from RNA polymerase II promoter
          eye development
          myeloid dendritic cell activation
          endothelial cell proliferation
          activation of innate immune response
          plasmacytoid dendritic cell activation
          macrophage activation involved in immune response
          dendritic cell chemotaxis
          inflammatory response to antigenic stimulus
          regulation of tolerance induction
          regulation of T cell mediated immune response to tumor cell
          DNA topological change
          base-excision repair
          apoptotic DNA fragmentation
          DNA recombination
          regulation of transcription from RNA polymerase II promoter
          autophagy
          inflammatory response
          positive regulation of cytosolic calcium ion concentration
          regulation of autophagy
          negative regulation of RNA polymerase II transcriptional preinitiation complex assembly
          lung development
          neuron projection development
          chromatin assembly
          regulation of restriction endodeoxyribonuclease activity
          activation of protein kinase activity
          DNA geometric change
          positive regulation of mismatch repair
          negative regulation of interferon-gamma production
          positive regulation of interferon-alpha production
          positive regulation of interferon-beta production
          positive regulation of interleukin-10 production
          positive regulation of interleukin-12 production
          positive regulation of tumor necrosis factor production
          V(D)J recombination
          positive regulation of toll-like receptor 2 signaling pathway
          positive regulation of toll-like receptor 4 signaling pathway
          positive regulation of toll-like receptor 9 signaling pathway
          T-helper 1 cell activation
          endothelial cell chemotaxis
          positive regulation of activated T cell proliferation
          positive regulation of apoptotic process
          apoptotic cell clearance
          positive regulation of cysteine-type endopeptidase activity involved in apoptotic process
          negative regulation of CD4-positive, alpha-beta T cell differentiation
          positive regulation of DNA binding
          positive regulation of MAPK cascade
          negative regulation of blood vessel endothelial cell migration
          T-helper 1 cell differentiation
          innate immune response
          positive regulation of myeloid cell differentiation
          positive regulation of glycogen catabolic process
          positive regulation of transcription from RNA polymerase II promoter
          positive regulation of JNK cascade
          positive regulation of interleukin-1 secretion
          positive regulation of interleukin-1 beta secretion
          positive chemotaxis
          DNA ligation involved in DNA repair
          positive regulation of DNA ligation
          response to glucocorticoid
          positive regulation of ERK1 and ERK2 cascade
          positive regulation of monocyte chemotaxis
          positive regulation of wound healing
          neutrophil clearance
          positive regulation of NIK/NF-kappaB signaling
          positive regulation of sprouting angiogenesis
          tumor necrosis factor secretion
          negative regulation of apoptotic cell clearance
          positive regulation of interleukin-6 secretion
          regulation of nucleotide-excision repair
          positive regulation of dendritic cell differentiation
          four-way junction DNA binding
          bubble DNA binding
          lipopolysaccharide binding
          phosphatidylserine binding
          damaged DNA binding
          double-stranded DNA binding
          single-stranded DNA binding
          transcription factor activity, sequence-specific DNA binding
          double-stranded RNA binding
          single-stranded RNA binding
          cytokine activity
          protein binding
          transcription factor binding
          DNA binding, bending
          calcium-dependent protein kinase regulator activity
          lyase activity
          C-X-C chemokine binding
          protein kinase activator activity
          chemoattractant activity
          poly(A) RNA binding
          RAGE receptor binding
          DNA polymerase binding
          repressing transcription factor binding
          supercoiled DNA binding

          Back To Top

          Related Products

          • POP-6™ Polymer
          • POP-7™ Polymer
          • BigDye® Terminator v1.1 Cycle Sequencing Kit
          • BigDye® Terminator v3.1 Cycle Sequencing Kit
          • BigDye® Direct Cycle Sequencing Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Hong Kong flag icon
          Hong Kong

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-8dsbr:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline