Hamburger Menu Button
Thermo Fisher Scientific Logo
登入
Don't have an account ?
Create Account
Products
Antibodies
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Tubes
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Clinical Genomics
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO 和 CRO 服務
CDMO 服務
臨床試驗 CRO 服務
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
Register for an Account
How to Order
Instrument Support
Technical Support Centers
Learning Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
快速訂購
登入
登入
Don't have an account ?
Create Account
帳戶
訂單
訂製產品與專案
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
Gene:
IRS2
Transcript:
NM_003749.2
Assay Name:
Mutation genome location:
chr.13 109781886-109782374 on build GRCh38
Assay ID
Hs00134525_CE
Purification
DS
HPLC
Size
25 nmol
25 nmol
50 nmol
50 nmol
25 nmol
25 nmol
50 nmol
50 nmol
25 nmol
25 nmol
50 nmol
50 nmol
25 nmol
25 nmol
50 nmol
50 nmol
Availability
Made To Order
Catalog #
A15629
Price
Your Price
Online offer:
Check your price ›
新增到購物車
Genomic Map
Assay Details
More Information
Genomic Map
LOADING...
×
Back To Top
Assay Details
Back To Top
More Information
Set Membership:
Between 400-700 bp
Gene Ontology Categories:
Function(s)
Process(es)
MAPK cascade
positive regulation of mesenchymal cell proliferation
negative regulation of B cell apoptotic process
glucose metabolic process
signal transduction
brain development
cell proliferation
positive regulation of cell proliferation
insulin receptor signaling pathway
response to glucose
negative regulation of plasma membrane long-chain fatty acid transport
positive regulation of glucose metabolic process
regulation of phosphatidylinositol 3-kinase signaling
regulation of lipid metabolic process
positive regulation of cell migration
mammary gland development
positive regulation of B cell proliferation
positive regulation of fatty acid beta-oxidation
positive regulation of insulin secretion
cellular response to insulin stimulus
negative regulation of kinase activity
phosphatidylinositol-3-phosphate biosynthetic process
positive regulation of GTPase activity
positive regulation of glycogen biosynthetic process
positive regulation of glucose import
phosphatidylinositol phosphorylation
phosphatidylinositol-mediated signaling
lipid homeostasis
cellular response to glucose stimulus
signal transducer activity
Ras guanyl-nucleotide exchange factor activity
insulin receptor binding
protein binding
1-phosphatidylinositol-3-kinase activity
protein kinase binding
protein phosphatase binding
protein domain specific binding
phosphatidylinositol 3-kinase binding
phosphatidylinositol-4,5-bisphosphate 3-kinase activity
14-3-3 protein binding
Back To Top
Related Products
POP-6
™
Polymer
POP-7
™
Polymer
BigDye
®
Terminator v1.1 Cycle Sequencing Kit
BigDye
®
Terminator v3.1 Cycle Sequencing Kit
BigDye
®
Direct Cycle Sequencing Kit
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-b49b87d85-5cvsx:80/100.66.79.163:80.
git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.47.0-Offline