Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
Gene:
ESR2
Transcript:
NM_001291712.1
Assay Name:
Mutation genome location:
chr.14 64282914-64283422 on build GRCh38
Assay ID
Hs00138895_CE
Purification
DS
HPLC
Size
25 nmol
25 nmol
50 nmol
50 nmol
25 nmol
25 nmol
50 nmol
50 nmol
25 nmol
25 nmol
50 nmol
50 nmol
25 nmol
25 nmol
50 nmol
50 nmol
Availability
Made To Order
Catalog #
A15629
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
Assay Details
More Information
Genomic Map
LOADING...
×
Back To Top
Assay Details
Back To Top
More Information
Set Membership:
Between 400-700 bp
Gene Ontology Categories:
Function(s)
Process(es)
negative regulation of transcription from RNA polymerase II promoter
ovarian follicle development
behavioral fear response
regulation of transcription, DNA-templated
transcription initiation from RNA polymerase II promoter
signal transduction
cell-cell signaling
learning or memory
response to water deprivation
response to activity
response to insecticide
cerebellum development
amygdala development
hypothalamus development
negative regulation of cell growth
intracellular estrogen receptor signaling pathway
prostate gland development
response to nutrient levels
response to testosterone
response to genistein
cellular response to drug
positive regulation of apoptotic process
regulation of neuron apoptotic process
response to estrogen
response to human chorionic gonadotropin
estrous cycle
response to ethanol
positive regulation of epidermal growth factor receptor signaling pathway
positive regulation of transcription, DNA-templated
positive regulation of vasodilation
positive regulation of transcription from RNA polymerase II promoter
negative regulation of smooth muscle cell proliferation
negative regulation of epithelial cell proliferation
positive regulation of sequence-specific DNA binding transcription factor activity
Sertoli cell development
Sertoli cell proliferation
positive regulation of ERK1 and ERK2 cascade
cellular response to magnetism
cellular response to estradiol stimulus
response to dexamethasone
extracellular negative regulation of signal transduction
negative regulation of neuron death
response to salt
response to bisphenol A
negative regulation of feeding behavior
negative regulation of reactive oxygen species metabolic process
core promoter sequence-specific DNA binding
DNA binding
transcription factor activity, sequence-specific DNA binding
steroid hormone receptor activity
transcription coactivator activity
RNA polymerase II transcription factor activity, ligand-activated sequence-specific DNA binding
steroid binding
protein binding
drug binding
zinc ion binding
enzyme binding
estrogen receptor activity
estrogen response element binding
RNA polymerase II transcription factor activity, estrogen-activated sequence-specific DNA binding
peroxisome proliferator activated receptor binding
receptor antagonist activity
estradiol binding
steroid hormone binding
Back To Top
Related Products
POP-6
™
Polymer
POP-7
™
Polymer
BigDye
®
Terminator v1.1 Cycle Sequencing Kit
BigDye
®
Terminator v3.1 Cycle Sequencing Kit
BigDye
®
Direct Cycle Sequencing Kit
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-b49b87d85-n9r8p:80/100.66.76.150:80.
git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.47.0-Offline