Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture and Transfection
Chemicals
Chromatography
Electron Microscopes
Lab Plasticware and Supplies
Lab Centrifuges
Lab Solutions
Mass Spectrometers
Next Generation Sequencers
See all product categories
Applications
Brands
Industrial and Applied Sciences
Food and Beverage
Forensics
Lab Solutions
Life Sciences
Pharma and Biopharma
Biotechnology
Clinical and Diagnostics
Digital Solutions
See all applications
Services
Lab Instrument and Equipment Services
Custom Services
Training Services
Financial and Leasing Services
Enterprise Level Lab Informatics
Partnering and Licensing Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Cell Biology Services
Food Safety Inspection Services
See all services
Help and Support
Contact Us
Product Documentation
Knowledge Base and Product FAQs
Learning Centers
Supply Center
eProcurement Solutions
Lab Instrument and Equipment Support
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Special Offers
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
Gene:
RORA
Transcript:
Assay Name:
Mutation genome location:
chr.15 60494296-60494806 on build GRCh38
Gene:
RORA-AS1
Transcript:
Assay Name:
Mutation genome location:
chr.15 60494296-60494806 on build GRCh38
Assay ID
Hs00147379_CE
Purification
DS
HPLC
Size
25 nmol
25 nmol
50 nmol
50 nmol
25 nmol
25 nmol
50 nmol
50 nmol
25 nmol
25 nmol
50 nmol
50 nmol
25 nmol
25 nmol
50 nmol
50 nmol
Availability
Made To Order
Catalog #
A15629
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
Assay Details
More Information
Genomic Map
LOADING...
×
Back To Top
Assay Details
Back To Top
More Information
Set Membership:
Between 400-700 bp
Panther Classification:
Molecular Function -
C4 zinc finger nuclear receptor
Gene Ontology Categories:
Function(s)
Process(es)
angiogenesis
regulation of transcription, DNA-templated
transcription initiation from RNA polymerase II promoter
xenobiotic metabolic process
nitric oxide biosynthetic process
regulation of smoothened signaling pathway
positive regulation of vascular endothelial growth factor production
regulation of glucose metabolic process
regulation of steroid metabolic process
cerebellar Purkinje cell differentiation
cerebellar granule cell precursor proliferation
intracellular receptor signaling pathway
circadian regulation of gene expression
cellular response to sterol
muscle cell differentiation
positive regulation of circadian rhythm
regulation of macrophage activation
negative regulation of I-kappaB kinase/NF-kappaB signaling
steroid hormone mediated signaling pathway
negative regulation of fat cell differentiation
positive regulation of transcription, DNA-templated
positive regulation of transcription from RNA polymerase II promoter
cGMP metabolic process
negative regulation of inflammatory response
regulation of transcription involved in cell fate commitment
triglyceride homeostasis
cellular response to interleukin-1
cellular response to tumor necrosis factor
cellular response to hypoxia
T-helper 17 cell differentiation
regulation of cholesterol homeostasis
RNA polymerase II regulatory region sequence-specific DNA binding
core promoter sequence-specific DNA binding
transcription corepressor binding
transcription coactivator binding
DNA binding
transcription factor activity, sequence-specific DNA binding
steroid hormone receptor activity
RNA polymerase II transcription factor activity, ligand-activated sequence-specific DNA binding
protein binding
beta-catenin binding
transcription factor binding
oxysterol binding
zinc ion binding
sequence-specific DNA binding
transcription factor activity, direct ligand regulated sequence-specific DNA binding
Back To Top
Related Products
POP-6
™
Polymer
POP-7
™
Polymer
BigDye
®
Terminator v1.1 Cycle Sequencing Kit
BigDye
®
Terminator v3.1 Cycle Sequencing Kit
BigDye
®
Direct Cycle Sequencing Kit
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-b49b87d85-5cvsx:80/100.66.79.163:80.
git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.47.0-Offline