Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture and Transfection
Chemicals
Chromatography
Electron Microscopes
Lab Plasticware and Supplies
Lab Centrifuges
Lab Solutions
Mass Spectrometers
Next Generation Sequencers
See all product categories
Applications
Brands
Industrial and Applied Sciences
Food and Beverage
Forensics
Lab Solutions
Life Sciences
Pharma and Biopharma
Biotechnology
Clinical and Diagnostics
Digital Solutions
See all applications
Services
Lab Instrument and Equipment Services
Custom Services
Training Services
Financial and Leasing Services
Enterprise Level Lab Informatics
Partnering and Licensing Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Cell Biology Services
Food Safety Inspection Services
See all services
Help and Support
Contact Us
Product Documentation
Knowledge Base and Product FAQs
Learning Centers
Supply Center
eProcurement Solutions
Lab Instrument and Equipment Support
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Special Offers
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
Gene:
COL1A1
Transcript:
Assay Name:
Mutation genome location:
chr.17 50184692-50185235 on build GRCh38
Assay ID
Hs00168710_CE
Purification
DS
HPLC
Size
25 nmol
25 nmol
50 nmol
50 nmol
25 nmol
25 nmol
50 nmol
50 nmol
25 nmol
25 nmol
50 nmol
50 nmol
25 nmol
25 nmol
50 nmol
50 nmol
Availability
Made To Order
Catalog #
A15629
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
Assay Details
More Information
Genomic Map
LOADING...
×
Back To Top
Assay Details
Back To Top
More Information
Set Membership:
Between 400-700 bp
Panther Classification:
Molecular Function -
extracellular matrix structural protein
Gene Ontology Categories:
Function(s)
Process(es)
skeletal system development
blood vessel development
osteoblast differentiation
intramembranous ossification
endochondral ossification
blood coagulation
visual perception
sensory perception of sound
positive regulation of epithelial to mesenchymal transition
negative regulation of cell-substrate adhesion
protein transport
platelet activation
extracellular matrix organization
collagen fibril organization
positive regulation of cell migration
collagen catabolic process
response to corticosteroid
response to estradiol
collagen biosynthetic process
protein localization to nucleus
tooth mineralization
response to drug
response to hydrogen peroxide
response to peptide hormone
skin morphogenesis
cellular response to fibroblast growth factor stimulus
tooth eruption
positive regulation of transcription, DNA-templated
embryonic skeletal system development
regulation of immune response
leukocyte migration
response to cAMP
response to hyperoxia
face morphogenesis
bone trabecula formation
cartilage development involved in endochondral bone morphogenesis
protein heterotrimerization
cellular response to amino acid stimulus
cellular response to mechanical stimulus
cellular response to retinoic acid
cellular response to vitamin E
cellular response to tumor necrosis factor
cellular response to epidermal growth factor stimulus
cellular response to transforming growth factor beta stimulus
positive regulation of canonical Wnt signaling pathway
cellular response to fluoride
extracellular matrix structural constituent
protein binding
identical protein binding
metal ion binding
platelet-derived growth factor binding
Back To Top
Related Products
POP-6
™
Polymer
POP-7
™
Polymer
BigDye
®
Terminator v1.1 Cycle Sequencing Kit
BigDye
®
Terminator v3.1 Cycle Sequencing Kit
BigDye
®
Direct Cycle Sequencing Kit
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-b49b87d85-5cvsx:80/100.66.79.163:80.
git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.47.0-Offline