Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
Gene:
ACOX1
Transcript:
NM_007292.5
Assay Name:
Mutation genome location:
chr.17 75979030-75979519 on build GRCh38
Assay transcription exon location:
Overlaps - Exon 1
Gene:
TEN1
Transcript:
NM_007292.5
Assay Name:
Mutation genome location:
chr.17 75979030-75979519 on build GRCh38
Assay transcription exon location:
Overlaps - Exon 1
Gene:
TEN1-CDK3
Transcript:
NM_007292.5
Assay Name:
Mutation genome location:
chr.17 75979030-75979519 on build GRCh38
Assay transcription exon location:
Overlaps - Exon 1
Assay ID
Hs00169774_CE
Purification
DS
HPLC
Size
25 nmol
25 nmol
50 nmol
50 nmol
25 nmol
25 nmol
50 nmol
50 nmol
25 nmol
25 nmol
50 nmol
50 nmol
25 nmol
25 nmol
50 nmol
50 nmol
Availability
Made To Order
Catalog #
A15629
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
Assay Details
More Information
Genomic Map
LOADING...
×
Back To Top
Assay Details
Back To Top
More Information
Set Membership:
Between 400-700 bp
Panther Classification:
Molecular Function -
oxidoreductase
Gene Ontology Categories:
Function(s)
Process(es)
very long-chain fatty acid metabolic process
generation of precursor metabolites and energy
lipid metabolic process
prostaglandin metabolic process
spermatogenesis
fatty acid oxidation
fatty acid beta-oxidation using acyl-CoA dehydrogenase
fatty acid beta-oxidation using acyl-CoA oxidase
alpha-linolenic acid metabolic process
negative regulation of telomerase activity
regulation of telomere maintenance via telomere lengthening
fatty-acyl-CoA binding
acyl-CoA dehydrogenase activity
acyl-CoA oxidase activity
receptor binding
electron carrier activity
PDZ domain binding
protein N-terminus binding
flavin adenine dinucleotide binding
oxidoreductase activity, acting on the CH-CH group of donors, with a flavin as acceptor
single-stranded DNA binding
protein binding
telomerase inhibitor activity
telomeric DNA binding
Back To Top
Related Products
POP-6
™
Polymer
POP-7
™
Polymer
BigDye
®
Terminator v1.1 Cycle Sequencing Kit
BigDye
®
Terminator v3.1 Cycle Sequencing Kit
BigDye
®
Direct Cycle Sequencing Kit
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-8jdkq:80/100.66.75.107:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0