Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Cell Analysis
Antibodies
Mass Spectrometry
Cell Culture
Laboratory Instruments
Clinical and Diagnostics
Chromatography
Laboratory Equipment
Laboratory Supplies
Molecular Biology and Nucleic Acid Analysis
Sequence-Specific Nucleic Acid Products
See all product categories
Applications
Cell Culture and Transfection
Flow Cytometry
Cancer Research
Chromatography
Sequencing
PCR
Lab Solutions
Allergy Diagnostics
See all applications and techniques
Services
Instrument Services
Cell Biology Services
Custom Services
Training Services
Lab Informatics Services
Financial and Leasing Services
Partnering and Licensing Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
See all services
Help and Support
How to Order
Promotions and Online Offers
Contact Us
Change Location
Create a New Account
See all help and support topics
Popular
Our Instagram
Our Facebook
Blog Behind the Bench
Customer Experience Center (CEC)
Ecommerce Exclusives
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Special Offers
Contact Us
Quick Order
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Contact Us
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
Gene:
PLCB1
Transcript:
NM_182734.2
Assay Name:
Mutation genome location:
chr.20 8658401-8658901 on build GRCh38
Assay ID
Hs00211464_CE
Purification
DS
HPLC
Size
25 nmol
25 nmol
50 nmol
50 nmol
25 nmol
25 nmol
50 nmol
50 nmol
25 nmol
25 nmol
50 nmol
50 nmol
25 nmol
25 nmol
50 nmol
50 nmol
Availability
Made To Order
Catalog #
A15629
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
Assay Details
More Information
Genomic Map
LOADING...
×
Back To Top
Assay Details
Back To Top
More Information
Set Membership:
Between 400-700 bp
Panther Classification:
Molecular Function -
phospholipase
Gene Ontology Categories:
Function(s)
Process(es)
G2/M transition of mitotic cell cycle
signal transduction
G-protein coupled acetylcholine receptor signaling pathway
glutamate receptor signaling pathway
Wnt signaling pathway, calcium modulating pathway
memory
regulation of G-protein coupled receptor protein signaling pathway
lipid catabolic process
cerebral cortex development
positive regulation of interleukin-12 production
intracellular signal transduction
interleukin-12-mediated signaling pathway
interleukin-15-mediated signaling pathway
positive regulation of embryonic development
positive regulation of GTPase activity
inositol phosphate metabolic process
fat cell differentiation
positive regulation of myoblast differentiation
negative regulation of transcription, DNA-templated
positive regulation of transcription, DNA-templated
positive regulation of JNK cascade
phosphatidylinositol metabolic process
insulin-like growth factor receptor signaling pathway
positive regulation of developmental growth
activation of meiosis involved in egg activation
interleukin-1-mediated signaling pathway
regulation of fertilization
positive regulation of G1/S transition of mitotic cell cycle
positive regulation of acrosome reaction
negative regulation of monocyte extravasation
positive regulation of CD24 biosynthetic process
phosphatidylinositol phospholipase C activity
signal transducer activity
GTPase activator activity
calcium ion binding
protein binding
calmodulin binding
lamin binding
phosphatidylinositol-4,5-bisphosphate binding
enzyme binding
protein homodimerization activity
Back To Top
Related Products
POP-6
™
Polymer
POP-7
™
Polymer
BigDye
®
Terminator v1.1 Cycle Sequencing Kit
BigDye
®
Terminator v3.1 Cycle Sequencing Kit
BigDye
®
Direct Cycle Sequencing Kit
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-b49b87d85-8dsbr:80/100.66.76.150:80.
git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.47.0-Offline