Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Clinical Genomics
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
Gene:
SOD1
Transcript:
NM_000454.4
Assay Name:
Mutation genome location:
chr.21 31667140-31667683 on build GRCh38
Assay ID
Hs00219174_CE
Purification
DS
HPLC
Size
25 nmol
25 nmol
50 nmol
50 nmol
25 nmol
25 nmol
50 nmol
50 nmol
25 nmol
25 nmol
50 nmol
50 nmol
25 nmol
25 nmol
50 nmol
50 nmol
Availability
Made To Order
Catalog #
A15629
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
Assay Details
More Information
Genomic Map
LOADING...
×
Back To Top
Assay Details
Back To Top
More Information
Set Membership:
Between 400-700 bp
Panther Classification:
Molecular Function -
oxidoreductase
Gene Ontology Categories:
Function(s)
Process(es)
activation of MAPK activity
response to reactive oxygen species
response to superoxide
ovarian follicle development
positive regulation of cytokine production
placenta development
retina homeostasis
response to amphetamine
myeloid cell homeostasis
platelet degranulation
glutathione metabolic process
superoxide metabolic process
cellular iron ion homeostasis
spermatogenesis
embryo implantation
cell aging
sensory perception of sound
locomotory behavior
anterograde axonal transport
retrograde axonal transport
regulation of blood pressure
response to heat
response to organic substance
transmission of nerve impulse
removal of superoxide radicals
response to nutrient levels
peripheral nervous system myelin maintenance
positive regulation of superoxide anion generation
regulation of T cell differentiation in thymus
response to carbon monoxide
cellular response to potassium ion
regulation of multicellular organism growth
response to drug
response to hydrogen peroxide
superoxide anion generation
positive regulation of apoptotic process
positive regulation of catalytic activity
regulation of GTPase activity
negative regulation of neuron apoptotic process
response to ethanol
negative regulation of cholesterol biosynthetic process
regulation of protein kinase activity
regulation of organ growth
response to antibiotic
response to copper ion
muscle cell cellular homeostasis
thymus development
response to axon injury
hydrogen peroxide biosynthetic process
regulation of mitochondrial membrane potential
oxidation-reduction process
heart contraction
neurofilament cytoskeleton organization
relaxation of vascular smooth muscle
auditory receptor cell stereocilium organization
cellular response to cadmium ion
cellular response to ATP
reactive oxygen species metabolic process
response to antipsychotic drug
positive regulation of oxidative stress-induced intrinsic apoptotic signaling pathway
superoxide dismutase activity
copper ion binding
protein binding
zinc ion binding
protein phosphatase 2B binding
identical protein binding
protein homodimerization activity
Rac GTPase binding
chaperone binding
Back To Top
Related Products
POP-6
™
Polymer
POP-7
™
Polymer
BigDye
®
Terminator v1.1 Cycle Sequencing Kit
BigDye
®
Terminator v3.1 Cycle Sequencing Kit
BigDye
®
Direct Cycle Sequencing Kit
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-67655d8755-zsnhf:80/100.66.75.14:80.
git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.46.0-Offline