Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Western Blot Products
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Food and Beverage
Lab Solutions
Pharma and Biopharma
Real-Time PCR
Semiconductor Analysis
Clinical and Diagnostics
Digital Solutions
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
See all services
Help and Support
Order Help
Digital Solutions
Product Support
Technical Information
Training and Education
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Special Offers
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
Gene:
CBS
Transcript:
XM_017028209.1
Assay Name:
Mutation genome location:
chr.21 6457782-6458319 on build GRCh38
Gene:
CBSL
Transcript:
XM_017028209.1
Assay Name:
Mutation genome location:
chr.21 6457782-6458319 on build GRCh38
Gene:
LOC102724843
Transcript:
XM_017028209.1
Assay Name:
Mutation genome location:
chr.21 6457782-6458319 on build GRCh38
Assay ID
Hs00220696_CE
Purification
DS
HPLC
Size
25 nmol
25 nmol
50 nmol
50 nmol
25 nmol
25 nmol
50 nmol
50 nmol
25 nmol
25 nmol
50 nmol
50 nmol
25 nmol
25 nmol
50 nmol
50 nmol
Availability
Made To Order
Catalog #
A15629
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
Assay Details
More Information
Genomic Map
LOADING...
×
Back To Top
Assay Details
Back To Top
More Information
Set Membership:
Between 400-700 bp
Panther Classification:
Molecular Function -
lyase
metabolite interconversion enzyme
Gene Ontology Categories:
Function(s)
Process(es)
cysteine biosynthetic process from serine
L-serine metabolic process
L-serine catabolic process
cellular amino acid biosynthetic process
cysteine biosynthetic process via cystathionine
cysteine biosynthetic process
transsulfuration
L-cysteine catabolic process
DNA protection
homocysteine catabolic process
homocysteine metabolic process
oxidation-reduction process
hydrogen sulfide biosynthetic process
cystathionine beta-synthase activity
cysteine synthase activity
protein binding
oxygen binding
enzyme binding
heme binding
pyridoxal phosphate binding
ubiquitin protein ligase binding
identical protein binding
protein homodimerization activity
metal ion binding
nitrite reductase (NO-forming) activity
carbon monoxide binding
nitric oxide binding
modified amino acid binding
S-adenosyl-L-methionine binding
Back To Top
Related Products
POP-6
™
Polymer
POP-7
™
Polymer
BigDye
®
Terminator v1.1 Cycle Sequencing Kit
BigDye
®
Terminator v3.1 Cycle Sequencing Kit
BigDye
®
Direct Cycle Sequencing Kit
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-b49b87d85-pgj68:80/100.66.79.163:80.
git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.47.0-Offline