Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Pipettes and Pipette Tips
Lab Centrifuges
Ultra-Low Temperature Freezers
Spectroscopy
Beakers
PCR Equipment and Supplies
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
See all product categories
Applications
Cell Analysis
Lab Equipment
Real-Time PCR
PCR
Chromatography
Cell Culture and Transfection
DNA and RNA Extraction and Analysis
Protein Biology
Flow Cytometry
Chemicals
See all applications and techniques
Services
Custom Services
Lab Informatics
Enterprise Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Instrument Services
Training Services
See all services
Help and Support
Customer Center
Contact Us
Certificates of Analysis and Conformance
Safety Data Sheets (SDS)
Manuals
How to Cite Our Products in a Paper
Instrument Support
Knowledge Base and Product FAQs
Learning Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
Gene:
TLR9
Transcript:
NM_017442.3
Assay Name:
Mutation genome location:
chr.3 52223690-52224189 on build GRCh38
Assay transcription exon location:
Within Exon 2
Assay ID
Hs00233012_CE
Purification
DS
HPLC
Size
25 nmol
25 nmol
50 nmol
50 nmol
25 nmol
25 nmol
50 nmol
50 nmol
25 nmol
25 nmol
50 nmol
50 nmol
25 nmol
25 nmol
50 nmol
50 nmol
Availability
Made To Order
Catalog #
A15629
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
Assay Details
More Information
Genomic Map
LOADING...
×
Back To Top
Assay Details
Back To Top
More Information
Set Membership:
Between 400-700 bp
Gene Ontology Categories:
Function(s)
Process(es)
toll-like receptor signaling pathway
response to molecule of bacterial origin
MyD88-dependent toll-like receptor signaling pathway
inflammatory response
I-kappaB phosphorylation
axonogenesis
positive regulation of gene expression
maintenance of gastrointestinal epithelium
negative regulation of NF-kappaB transcription factor activity
tumor necrosis factor production
negative regulation of interleukin-6 production
negative regulation of interleukin-8 production
positive regulation of chemokine production
positive regulation of granulocyte macrophage colony-stimulating factor production
positive regulation of interferon-beta production
positive regulation of interleukin-10 production
positive regulation of interleukin-12 production
positive regulation of interleukin-18 production
positive regulation of interleukin-6 production
positive regulation of interleukin-8 production
positive regulation of tumor necrosis factor production
negative regulation of toll-like receptor signaling pathway
positive regulation of toll-like receptor signaling pathway
toll-like receptor 9 signaling pathway
phosphatidylinositol-3-phosphate biosynthetic process
positive regulation of NF-kappaB import into nucleus
defense response to bacterium
positive regulation of I-kappaB kinase/NF-kappaB signaling
positive regulation of JUN kinase activity
positive regulation of interferon-gamma biosynthetic process
innate immune response
positive regulation of interferon-alpha biosynthetic process
positive regulation of interferon-beta biosynthetic process
positive regulation of transcription from RNA polymerase II promoter
positive regulation of JNK cascade
regulation of cytokine secretion
positive regulation of inflammatory response
defense response to Gram-negative bacterium
positive regulation of NF-kappaB transcription factor activity
positive regulation of nitric-oxide synthase biosynthetic process
negative regulation of calcium-transporting ATPase activity
transmembrane signaling receptor activity
interleukin-1 receptor binding
1-phosphatidylinositol-3-kinase activity
siRNA binding
Back To Top
Related Products
POP-6
™
Polymer
POP-7
™
Polymer
BigDye
®
Terminator v1.1 Cycle Sequencing Kit
BigDye
®
Terminator v3.1 Cycle Sequencing Kit
BigDye
®
Direct Cycle Sequencing Kit
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-zbw5q:80/100.66.77.212:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0