Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture and Transfection
Chemicals
Chromatography
Electron Microscopes
Lab Plasticware and Supplies
Lab Centrifuges
Lab Solutions
Mass Spectrometers
Next Generation Sequencers
See all product categories
Applications
Brands
Industrial and Applied Sciences
Food and Beverage
Forensics
Lab Solutions
Life Sciences
Pharma and Biopharma
Biotechnology
Clinical and Diagnostics
Digital Solutions
See all applications
Services
Lab Instrument and Equipment Services
Custom Services
Training Services
Financial and Leasing Services
Enterprise Level Lab Informatics
Partnering and Licensing Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Cell Biology Services
Food Safety Inspection Services
See all services
Help and Support
Contact Us
Product Documentation
Knowledge Base and Product FAQs
Learning Centers
Supply Center
eProcurement Solutions
Lab Instrument and Equipment Support
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Special Offers
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
Gene:
GSK3B
Transcript:
Assay Name:
Mutation genome location:
chr.3 119824114-119824635 on build GRCh38
Assay ID
Hs00237753_CE
Purification
DS
HPLC
Size
25 nmol
25 nmol
50 nmol
50 nmol
25 nmol
25 nmol
50 nmol
50 nmol
25 nmol
25 nmol
50 nmol
50 nmol
25 nmol
25 nmol
50 nmol
50 nmol
Availability
Made To Order
Catalog #
A15629
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
Assay Details
More Information
Genomic Map
LOADING...
×
Back To Top
Assay Details
Back To Top
More Information
Set Membership:
Between 400-700 bp
Panther Classification:
Molecular Function -
non-receptor serine/threonine protein kinase
Gene Ontology Categories:
Function(s)
Process(es)
re-entry into mitotic cell cycle
epithelial to mesenchymal transition
positive regulation of cell-matrix adhesion
glycogen metabolic process
regulation of gene expression by genetic imprinting
protein phosphorylation
protein export from nucleus
ER overload response
dopamine receptor signaling pathway
axonogenesis
myoblast fusion
aging
circadian rhythm
organ morphogenesis
negative regulation of cardiac muscle hypertrophy
positive regulation of peptidyl-threonine phosphorylation
positive regulation of mitochondrial membrane potential
negative regulation of neuron maturation
response to activity
Wnt signaling pathway
cell migration
peptidyl-serine phosphorylation
peptidyl-threonine phosphorylation
hippocampus development
establishment of cell polarity
negative regulation of protein complex assembly
positive regulation of protein complex assembly
negative regulation of protein binding
positive regulation of protein binding
response to estradiol
positive regulation of proteasomal ubiquitin-dependent protein catabolic process
response to insulin
regulation of microtubule-based process
positive regulation of peptidyl-serine phosphorylation
negative regulation of smooth muscle cell apoptotic process
protein localization to microtubule
intracellular signal transduction
cellular response to hepatocyte growth factor stimulus
cellular response to interleukin-3
cellular response to erythropoietin
response to drug
negative regulation of apoptotic process
proteasome-mediated ubiquitin-dependent protein catabolic process
negative regulation of MAP kinase activity
positive regulation of neuron apoptotic process
positive regulation of GTPase activity
hypermethylation of CpG island
canonical Wnt signaling pathway involved in positive regulation of apoptotic process
fat cell differentiation
negative regulation of glycogen biosynthetic process
positive regulation of protein catabolic process
positive regulation of axon extension
positive regulation of transcription from RNA polymerase II promoter
protein autophosphorylation
positive regulation of protein export from nucleus
regulation of neuronal synaptic plasticity
positive regulation of smooth muscle cell proliferation
negative regulation of dendrite morphogenesis
negative regulation of nitric-oxide synthase activity
negative regulation of NFAT protein import into nucleus
canonical Wnt signaling pathway
intrinsic apoptotic signaling pathway in response to endoplasmic reticulum stress
cellular response to hydrogen peroxide
superior temporal gyrus development
cellular response to mechanical stimulus
cellular response to iron(II) ion
cellular response to lithium ion
response to epinephrine
negative regulation of canonical Wnt signaling pathway
extrinsic apoptotic signaling pathway in absence of ligand
chemical synaptic transmission, postsynaptic
regulation of cellular response to heat
negative regulation of protein localization to nucleus
positive regulation of mitochondrial outer membrane permeabilization involved in apoptotic signaling pathway
positive regulation of neuron death
response to L-glutamate
cellular response to dopamine
negative regulation of dopaminergic neuron differentiation
beta-catenin destruction complex assembly
beta-catenin destruction complex disassembly
response to insulin-like growth factor stimulus
response to ultrasound
response to angiotensin
negative regulation of type B pancreatic cell development
positive regulation of excitatory postsynaptic potential
negative regulation of glycogen (starch) synthase activity
positive regulation of DNA biosynthetic process
positive regulation of stem cell differentiation
negative regulation of neuron migration
RNA polymerase II transcription factor binding
p53 binding
protein kinase activity
protein serine/threonine kinase activity
integrin binding
protein binding
ATP binding
beta-catenin binding
kinase activity
protein kinase binding
ubiquitin protein ligase binding
protein kinase A catalytic subunit binding
ionotropic glutamate receptor binding
tau protein binding
tau-protein kinase activity
NF-kappaB binding
Back To Top
Related Products
POP-6
™
Polymer
POP-7
™
Polymer
BigDye
®
Terminator v1.1 Cycle Sequencing Kit
BigDye
®
Terminator v3.1 Cycle Sequencing Kit
BigDye
®
Direct Cycle Sequencing Kit
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-b49b87d85-n9r8p:80/100.66.76.150:80.
git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.47.0-Offline