Hamburger Menu Button
Thermo Fisher Scientific Logo
登入
還沒有帳戶嗎 ?
立即註冊
產品
Antibodies
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Cell Culture Media
Chemicals
Western Blot Products
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Greener Products
查看所有產品類別
應用
生物製程
細胞培養和轉染
食品和飲料
實驗室解決方案
製藥和生物製藥
即時定量PCR
半導體分析
臨床和診斷
數位解決方案
分子光譜、同位素與元素分析
查看所有應用與技術
服務
360° CDMO 和 CRO 服務
CDMO 服务
临床试验 CRO 服务
客製化服務
金融與租賃服務
儀器服務
實驗室資訊學
授權與商業供應
培訓服務
查看所有服務
支援
線上訂購協助
數位解決方案
產品支持
技術相關資訊
培訓與教育
查看所有協助與支援
熱門商品
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
我們的服務對象
生物技術
生物製藥
CDMO
實驗室診斷
工業與應用科學
特價商品
聯絡我們
快速訂購
訂單狀態與貨物追蹤
文件和證書
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
訂單查詢
快速訂購
登入
登入
還沒有帳戶嗎 ?
立即註冊
帳戶
訂單
Connect: Lab, Data, Apps
客製化產品與項目
儀器管理
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
Gene:
GATA2
Transcript:
Assay Name:
Mutation genome location:
chr.3 128492803-128493338 on build GRCh38
Gene:
GATA2-AS1
Transcript:
Assay Name:
Mutation genome location:
chr.3 128492803-128493338 on build GRCh38
Assay ID
Hs00239113_CE
Purification
DS
HPLC
Size
25 nmol
25 nmol
50 nmol
50 nmol
25 nmol
25 nmol
50 nmol
50 nmol
25 nmol
25 nmol
50 nmol
50 nmol
25 nmol
25 nmol
50 nmol
50 nmol
Availability
Made To Order
Catalog #
A15629
Price
Your Price
Online offer:
Check your price ›
新增到購物車
Genomic Map
Assay Details
More Information
Genomic Map
LOADING...
×
Back To Top
Assay Details
Back To Top
More Information
Set Membership:
Between 400-700 bp
Panther Classification:
Molecular Function -
DNA-binding transcription factor
Gene Ontology Categories:
Function(s)
Process(es)
negative regulation of transcription from RNA polymerase II promoter
urogenital system development
cell fate determination
neuron migration
embryonic placenta development
transcription, DNA-templated
transcription from RNA polymerase II promoter
phagocytosis
positive regulation of cytosolic calcium ion concentration
heart development
blood coagulation
organ morphogenesis
tissue development
regulation of primitive erythrocyte differentiation
ventral spinal cord interneuron differentiation
cell differentiation in hindbrain
commitment of neuronal cell to specific neuron type in forebrain
central nervous system neuron development
pituitary gland development
response to lipid
somatic stem cell population maintenance
regulation of histone acetylation
eosinophil fate commitment
inner ear morphogenesis
mast cell degranulation
negative regulation of fat cell differentiation
positive regulation of erythrocyte differentiation
negative regulation of macrophage differentiation
positive regulation of megakaryocyte differentiation
positive regulation of neuron differentiation
negative regulation of Notch signaling pathway
positive regulation of angiogenesis
positive regulation of transcription from RNA polymerase II promoter
cell maturation
digestive tract development
homeostasis of number of cells within a tissue
positive regulation of phagocytosis
positive regulation of phagocytosis, engulfment
definitive hemopoiesis
semicircular canal development
negative regulation of fat cell proliferation
GABAergic neuron differentiation
negative regulation of neural precursor cell proliferation
regulation of forebrain neuron differentiation
RNA polymerase II regulatory region sequence-specific DNA binding
RNA polymerase II core promoter sequence-specific DNA binding
RNA polymerase II distal enhancer sequence-specific DNA binding
transcriptional activator activity, RNA polymerase II core promoter proximal region sequence-specific binding
RNA polymerase II transcription factor binding
enhancer sequence-specific DNA binding
transcriptional activator activity, RNA polymerase II transcription regulatory region sequence-specific binding
chromatin binding
transcription factor activity, sequence-specific DNA binding
protein binding
transcription factor binding
zinc ion binding
C2H2 zinc finger domain binding
Back To Top
Related Products
POP-6
™
Polymer
POP-7
™
Polymer
BigDye
®
Terminator v1.1 Cycle Sequencing Kit
BigDye
®
Terminator v3.1 Cycle Sequencing Kit
BigDye
®
Direct Cycle Sequencing Kit
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-b49b87d85-9fm2r:80/100.66.79.29:80.
git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.47.0-Offline