Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
Gene:
RBPJ
Transcript:
NM_005349.3
Assay Name:
Mutation genome location:
chr.4 26428589-26429108 on build GRCh38
Assay ID
Hs00248575_CE
Purification
DS
HPLC
Size
25 nmol
25 nmol
50 nmol
50 nmol
25 nmol
25 nmol
50 nmol
50 nmol
25 nmol
25 nmol
50 nmol
50 nmol
25 nmol
25 nmol
50 nmol
50 nmol
Availability
Made To Order
Catalog #
A15629
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
Assay Details
More Information
Genomic Map
LOADING...
×
Back To Top
Assay Details
Back To Top
More Information
Set Membership:
Between 400-700 bp
Panther Classification:
Molecular Function -
DNA-binding transcription factor
Gene Ontology Categories:
Function(s)
Process(es)
negative regulation of transcription from RNA polymerase II promoter
angiogenesis
somitogenesis
epithelial to mesenchymal transition
blood vessel remodeling
inflammatory response to antigenic stimulus
secondary heart field specification
outflow tract morphogenesis
endocardium morphogenesis
epithelial to mesenchymal transition involved in endocardial cushion formation
cardiac left ventricle morphogenesis
ventricular trabecula myocardium morphogenesis
regulation of transcription from RNA polymerase II promoter involved in myocardial precursor cell differentiation
DNA recombination
transcription initiation from RNA polymerase II promoter
humoral immune response
Notch signaling pathway
positive regulation of transcription of Notch receptor target
negative regulation of cell proliferation
auditory receptor cell fate commitment
epidermal cell fate specification
pituitary gland development
B cell differentiation
keratinocyte differentiation
negative regulation of ossification
positive regulation of BMP signaling pathway
somatic stem cell population maintenance
dorsal aorta morphogenesis
atrioventricular canal development
defense response to bacterium
myeloid dendritic cell differentiation
negative regulation of cell differentiation
negative regulation of transcription, DNA-templated
positive regulation of transcription from RNA polymerase II promoter
regulation of timing of cell differentiation
sebaceous gland development
hair follicle maturation
positive regulation of cardiac muscle cell proliferation
Clara cell differentiation
labyrinthine layer blood vessel development
arterial endothelial cell fate commitment
Notch signaling involved in heart development
positive regulation of transcription from RNA polymerase II promoter in response to hypoxia
blood vessel lumenization
interleukin-4 secretion
blood vessel endothelial cell fate specification
positive regulation of ERBB signaling pathway
positive regulation of ephrin receptor signaling pathway
positive regulation of canonical Wnt signaling pathway involved in cardiac muscle cell fate commitment
positive regulation of cell proliferation involved in heart morphogenesis
recombinase activity
RNA polymerase II core promoter proximal region sequence-specific DNA binding
RNA polymerase II core promoter sequence-specific DNA binding
transcriptional activator activity, RNA polymerase II core promoter proximal region sequence-specific binding
RNA polymerase II repressing transcription factor binding
DNA binding
chromatin binding
transcription factor activity, sequence-specific DNA binding
protein binding
transcription factor binding
sequence-specific DNA binding
protein N-terminus binding
Back To Top
Related Products
POP-6
™
Polymer
POP-7
™
Polymer
BigDye
®
Terminator v1.1 Cycle Sequencing Kit
BigDye
®
Terminator v3.1 Cycle Sequencing Kit
BigDye
®
Direct Cycle Sequencing Kit
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-ctdjb:80/100.66.76.55:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0