Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
Gene:
APC
Transcript:
Assay Name:
Mutation genome location:
chr.5 112840120-112840619 on build GRCh38
Assay ID
Hs00263616_CE
Purification
DS
HPLC
Size
25 nmol
25 nmol
50 nmol
50 nmol
25 nmol
25 nmol
50 nmol
50 nmol
25 nmol
25 nmol
50 nmol
50 nmol
25 nmol
25 nmol
50 nmol
50 nmol
Availability
Made To Order
Catalog #
A15629
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
Assay Details
More Information
Genomic Map
LOADING...
×
Back To Top
Assay Details
Back To Top
More Information
Set Membership:
Between 400-700 bp
Gene Ontology Categories:
Function(s)
Process(es)
mitotic cytokinesis
cell fate specification
protein complex assembly
cellular component disassembly involved in execution phase of apoptosis
cellular response to DNA damage stimulus
negative regulation of microtubule depolymerization
cell cycle arrest
mitotic spindle assembly checkpoint
cell adhesion
establishment or maintenance of cell polarity
pattern specification process
negative regulation of cell proliferation
Wnt signaling pathway
cell migration
positive regulation of cell migration
pancreas development
neuron projection development
positive regulation of pseudopodium assembly
regulation of microtubule-based process
response to drug
positive regulation of apoptotic process
proteasome-mediated ubiquitin-dependent protein catabolic process
regulation of cell differentiation
positive regulation of protein catabolic process
negative regulation of cyclin-dependent protein serine/threonine kinase activity
regulation of attachment of spindle microtubules to kinetochore
canonical Wnt signaling pathway
bicellular tight junction assembly
negative regulation of canonical Wnt signaling pathway
response to alcohol
beta-catenin destruction complex assembly
beta-catenin destruction complex disassembly
cellular response to nerve growth factor stimulus
regulation of glutamate metabolic process
protein binding
beta-catenin binding
microtubule binding
protein kinase regulator activity
protein kinase binding
protein complex binding
gamma-catenin binding
cadherin binding
microtubule plus-end binding
Back To Top
Related Products
POP-6
™
Polymer
POP-7
™
Polymer
BigDye
®
Terminator v1.1 Cycle Sequencing Kit
BigDye
®
Terminator v3.1 Cycle Sequencing Kit
BigDye
®
Direct Cycle Sequencing Kit
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-67655d8755-vzwvz:80/100.66.79.173:80.
git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.46.0-Offline