Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture and Transfection
Chemicals
Chromatography
Electron Microscopes
Lab Plasticware and Supplies
Lab Centrifuges
Lab Solutions
Mass Spectrometers
Next Generation Sequencers
See all product categories
Applications
Brands
Industrial and Applied Sciences
Food and Beverage
Forensics
Lab Solutions
Life Sciences
Pharma and Biopharma
Biotechnology
Clinical and Diagnostics
Digital Solutions
See all applications
Services
Lab Instrument and Equipment Services
Custom Services
Training Services
Financial and Leasing Services
Enterprise Level Lab Informatics
Partnering and Licensing Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Cell Biology Services
Food Safety Inspection Services
See all services
Help and Support
Contact Us
Product Documentation
Knowledge Base and Product FAQs
Learning Centers
Supply Center
eProcurement Solutions
Lab Instrument and Equipment Support
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Special Offers
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
Gene:
CD74
Transcript:
Assay Name:
Mutation genome location:
chr.5 150412778-150413219 on build GRCh38
Assay ID
Hs00267943_CE
Purification
DS
HPLC
Size
25 nmol
25 nmol
50 nmol
50 nmol
25 nmol
25 nmol
50 nmol
50 nmol
25 nmol
25 nmol
50 nmol
50 nmol
25 nmol
25 nmol
50 nmol
50 nmol
Availability
Made To Order
Catalog #
A15629
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
Assay Details
More Information
Genomic Map
LOADING...
×
Back To Top
Assay Details
Back To Top
More Information
Set Membership:
Between 400-700 bp
Panther Classification:
Molecular Function -
scaffold/adaptor protein
Gene Ontology Categories:
Function(s)
Process(es)
activation of MAPK activity
prostaglandin biosynthetic process
positive regulation of cytokine-mediated signaling pathway
positive regulation of dendritic cell antigen processing and presentation
negative regulation of peptide secretion
positive regulation of type 2 immune response
negative regulation of mature B cell apoptotic process
protein complex assembly
intracellular protein transport
defense response
signal transduction
cell proliferation
immunoglobulin mediated immune response
antigen processing and presentation of endogenous antigen
antigen processing and presentation of exogenous peptide antigen via MHC class II
positive regulation of B cell proliferation
macrophage migration inhibitory factor signaling pathway
regulation of macrophage activation
negative regulation of apoptotic process
negative regulation of DNA damage response, signal transduction by p53 class mediator
T cell selection
positive thymic T cell selection
negative thymic T cell selection
negative regulation of T cell differentiation
positive regulation of T cell differentiation
positive regulation of fibroblast proliferation
positive regulation of peptidyl-tyrosine phosphorylation
leukocyte migration
chaperone mediated protein folding requiring cofactor
positive regulation of macrophage cytokine production
positive regulation of ERK1 and ERK2 cascade
positive regulation of neutrophil chemotaxis
negative regulation of intrinsic apoptotic signaling pathway in response to DNA damage by p53 class mediator
positive regulation of chemokine (C-X-C motif) ligand 2 production
beta-amyloid binding
cytokine receptor activity
protein binding
cytokine binding
MHC class II protein complex binding
macrophage migration inhibitory factor binding
MHC class II protein binding
MHC class II protein binding, via antigen binding groove
identical protein binding
protein binding involved in protein folding
nitric-oxide synthase binding
Back To Top
Related Products
POP-6
™
Polymer
POP-7
™
Polymer
BigDye
®
Terminator v1.1 Cycle Sequencing Kit
BigDye
®
Terminator v3.1 Cycle Sequencing Kit
BigDye
®
Direct Cycle Sequencing Kit
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-b49b87d85-pgj68:80/100.66.79.163:80.
git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.47.0-Offline