Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Cell Analysis
Antibodies
Mass Spectrometry
Cell Culture
Laboratory Instruments
Clinical and Diagnostics
Chromatography
Laboratory Equipment
Laboratory Supplies
Molecular Biology and Nucleic Acid Analysis
Sequence-Specific Nucleic Acid Products
See all product categories
Applications
Cell Culture and Transfection
Flow Cytometry
Cancer Research
Chromatography
Sequencing
PCR
Lab Solutions
Allergy Diagnostics
See all applications and techniques
Services
Instrument Services
Cell Biology Services
Custom Services
Training Services
Lab Informatics Services
Financial and Leasing Services
Partnering and Licensing Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
See all services
Help and Support
How to Order
Promotions and Online Offers
Contact Us
Change Location
Create a New Account
See all help and support topics
Popular
Our Instagram
Our Facebook
Blog Behind the Bench
Customer Experience Center (CEC)
Ecommerce Exclusives
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Special Offers
Contact Us
Quick Order
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Contact Us
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
Gene:
HAVCR2
Transcript:
NM_032782.4
Assay Name:
Mutation genome location:
chr.5 157106536-157107038 on build GRCh38
Assay ID
Hs00268611_CE
Purification
DS
HPLC
Size
25 nmol
25 nmol
50 nmol
50 nmol
25 nmol
25 nmol
50 nmol
50 nmol
25 nmol
25 nmol
50 nmol
50 nmol
25 nmol
25 nmol
50 nmol
50 nmol
Availability
Made To Order
Catalog #
A15629
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
Assay Details
More Information
Genomic Map
LOADING...
×
Back To Top
Assay Details
Back To Top
More Information
Set Membership:
Between 400-700 bp
Panther Classification:
Molecular Function -
immunoglobulin receptor superfamily
Gene Ontology Categories:
Function(s)
Process(es)
adaptive immune response
macrophage activation involved in immune response
natural killer cell tolerance induction
regulation of tolerance induction dependent upon immune response
negative regulation of T-helper 1 type immune response
negative regulation of natural killer cell mediated cytotoxicity directed against tumor cell target
inflammatory response
negative regulation of gene expression
negative regulation of myeloid dendritic cell activation
negative regulation of NF-kappaB transcription factor activity
negative regulation of interferon-alpha production
negative regulation of interferon-gamma production
negative regulation of interleukin-2 production
negative regulation of interleukin-3 production
negative regulation of interleukin-6 production
negative regulation of tumor necrosis factor production
positive regulation of chemokine production
positive regulation of interferon-gamma production
positive regulation of interleukin-1 production
positive regulation of interleukin-4 production
negative regulation of natural killer cell activation
toll-like receptor 3 signaling pathway
toll-like receptor 7 signaling pathway
toll-like receptor 9 signaling pathway
positive regulation of T cell proliferation
negative regulation of T cell proliferation
positive regulation of macrophage activation
innate immune response
defense response to Gram-positive bacterium
maternal process involved in female pregnancy
positive regulation of ERK1 and ERK2 cascade
cellular response to lipopolysaccharide
negative regulation of granulocyte colony-stimulating factor production
negative regulation of defense response to bacterium
positive regulation of defense response to bacterium
positive regulation of NIK/NF-kappaB signaling
positive regulation of tumor necrosis factor secretion
negative regulation of immunological synapse formation
negative regulation of T cell activation via T cell receptor contact with antigen bound to MHC molecule on antigen presenting cell
protein binding
metal ion binding
Back To Top
Related Products
POP-6
™
Polymer
POP-7
™
Polymer
BigDye
®
Terminator v1.1 Cycle Sequencing Kit
BigDye
®
Terminator v3.1 Cycle Sequencing Kit
BigDye
®
Direct Cycle Sequencing Kit
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-b49b87d85-lnstr:80/100.66.74.145:80.
git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.47.0-Offline