Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Western Blot Products
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Food and Beverage
Lab Solutions
Pharma and Biopharma
Real-Time PCR
Semiconductor Analysis
Clinical and Diagnostics
Digital Solutions
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
See all services
Help and Support
Order Help
Digital Solutions
Product Support
Technical Information
Training and Education
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Special Offers
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
Gene:
PDGFA
Transcript:
Assay Name:
Mutation genome location:
chr.7 497552-498057 on build GRCh38
Assay ID
Hs00284354_CE
Purification
DS
HPLC
Size
25 nmol
25 nmol
50 nmol
50 nmol
25 nmol
25 nmol
50 nmol
50 nmol
25 nmol
25 nmol
50 nmol
50 nmol
25 nmol
25 nmol
50 nmol
50 nmol
Availability
Made To Order
Catalog #
A15629
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
Assay Details
More Information
Genomic Map
LOADING...
×
Back To Top
Assay Details
Back To Top
More Information
Set Membership:
Between 400-700 bp
Panther Classification:
Molecular Function -
growth factor
Gene Ontology Categories:
Function(s)
Process(es)
MAPK cascade
angiogenesis
response to hypoxia
cell activation
hair follicle development
positive regulation of mesenchymal cell proliferation
platelet degranulation
transforming growth factor beta receptor signaling pathway
cell-cell signaling
positive regulation of cell proliferation
response to wounding
organ morphogenesis
response to inorganic substance
negative regulation of phosphatidylinositol biosynthetic process
negative regulation of platelet activation
regulation of phosphatidylinositol 3-kinase signaling
positive regulation of phosphatidylinositol 3-kinase signaling
regulation of smooth muscle cell migration
cell projection assembly
actin cytoskeleton organization
extracellular matrix organization
positive regulation of cell migration
positive regulation of protein autophosphorylation
response to estradiol
response to retinoic acid
regulation of actin cytoskeleton organization
positive regulation of metanephric mesenchymal cell migration by platelet-derived growth factor receptor-beta signaling pathway
wound healing
response to drug
positive regulation of MAP kinase activity
positive regulation of MAPK cascade
positive regulation of GTPase activity
skin development
positive regulation of DNA replication
phosphatidylinositol phosphorylation
platelet-derived growth factor receptor signaling pathway
phosphatidylinositol-mediated signaling
positive regulation of fibroblast proliferation
lung alveolus development
inner ear development
regulation of peptidyl-tyrosine phosphorylation
negative chemotaxis
positive regulation of cell division
positive regulation of protein kinase B signaling
regulation of branching involved in salivary gland morphogenesis by epithelial-mesenchymal signaling
positive regulation of ERK1 and ERK2 cascade
regulation of glomerular mesangial cell proliferation
embryonic lung development
regulation of DNA biosynthetic process
Ras guanyl-nucleotide exchange factor activity
platelet-derived growth factor receptor binding
protein binding
collagen binding
growth factor activity
protein homodimerization activity
phosphatidylinositol-4,5-bisphosphate 3-kinase activity
protein heterodimerization activity
platelet-derived growth factor binding
Back To Top
Related Products
POP-6
™
Polymer
POP-7
™
Polymer
BigDye
®
Terminator v1.1 Cycle Sequencing Kit
BigDye
®
Terminator v3.1 Cycle Sequencing Kit
BigDye
®
Direct Cycle Sequencing Kit
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-b49b87d85-pgj68:80/100.66.79.163:80.
git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.47.0-Offline