Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Cell Analysis
Antibodies
Mass Spectrometry
Cell Culture
Laboratory Instruments
Clinical and Diagnostics
Chromatography
Laboratory Equipment
Laboratory Supplies
Molecular Biology and Nucleic Acid Analysis
Sequence-Specific Nucleic Acid Products
See all product categories
Applications
Cell Culture and Transfection
Flow Cytometry
Cancer Research
Chromatography
Sequencing
PCR
Lab Solutions
Allergy Diagnostics
See all applications and techniques
Services
Instrument Services
Cell Biology Services
Custom Services
Training Services
Lab Informatics Services
Financial and Leasing Services
Partnering and Licensing Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
See all services
Help and Support
How to Order
Promotions and Online Offers
Contact Us
Change Location
Create a New Account
See all help and support topics
Popular
Our Instagram
Our Facebook
Blog Behind the Bench
Customer Experience Center (CEC)
Ecommerce Exclusives
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Special Offers
Contact Us
Quick Order
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Contact Us
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
Gene:
AQP1
Transcript:
NM_198098.2
Assay Name:
Mutation genome location:
chr.7 30921741-30922234 on build GRCh38
Assay ID
Hs00286268_CE
Purification
DS
HPLC
Size
25 nmol
25 nmol
50 nmol
50 nmol
25 nmol
25 nmol
50 nmol
50 nmol
25 nmol
25 nmol
50 nmol
50 nmol
25 nmol
25 nmol
50 nmol
50 nmol
Availability
Made To Order
Catalog #
A15629
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
Assay Details
More Information
Genomic Map
LOADING...
×
Back To Top
Assay Details
Back To Top
More Information
Set Membership:
Between 400-700 bp
Panther Classification:
Molecular Function -
transporter
Gene Ontology Categories:
Function(s)
Process(es)
renal water homeostasis
renal water transport
cGMP biosynthetic process
potassium ion transport
water transport
cell volume homeostasis
cellular water homeostasis
carbon dioxide transport
ammonium transport
bicarbonate transport
glycerol transport
cellular homeostasis
lateral ventricle development
pancreatic juice secretion
nitric oxide transport
establishment or maintenance of actin cytoskeleton polarity
cerebrospinal fluid secretion
cellular response to stress
cellular response to UV
transepithelial water transport
carbon dioxide transmembrane transport
odontogenesis
response to drug
negative regulation of apoptotic process
negative regulation of cysteine-type endopeptidase activity involved in apoptotic process
positive regulation of angiogenesis
positive regulation of saliva secretion
positive regulation of fibroblast proliferation
multicellular organismal water homeostasis
cellular response to hydrogen peroxide
cellular response to inorganic substance
cellular response to mechanical stimulus
cellular response to copper ion
cellular response to mercury ion
cellular response to retinoic acid
cellular response to cAMP
cellular response to hypoxia
cellular response to salt stress
cellular hyperosmotic response
cellular response to dexamethasone stimulus
cellular response to nitric oxide
potassium ion transmembrane transport
ammonium transmembrane transport
maintenance of symbiont-containing vacuole by host
intracellular cGMP activated cation channel activity
potassium channel activity
water transmembrane transporter activity
protein binding
ammonium transmembrane transporter activity
potassium ion transmembrane transporter activity
glycerol transmembrane transporter activity
water channel activity
glycerol channel activity
transmembrane transporter activity
nitric oxide transmembrane transporter activity
carbon dioxide transmembrane transporter activity
Back To Top
Related Products
POP-6
™
Polymer
POP-7
™
Polymer
BigDye
®
Terminator v1.1 Cycle Sequencing Kit
BigDye
®
Terminator v3.1 Cycle Sequencing Kit
BigDye
®
Direct Cycle Sequencing Kit
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-b49b87d85-8dsbr:80/100.66.76.150:80.
git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.47.0-Offline