Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Cell Analysis
Antibodies
Mass Spectrometry
Cell Culture
Laboratory Instruments
Clinical and Diagnostics
Chromatography
Laboratory Equipment
Laboratory Supplies
Molecular Biology and Nucleic Acid Analysis
Sequence-Specific Nucleic Acid Products
See all product categories
Applications
Cell Culture and Transfection
Flow Cytometry
Cancer Research
Chromatography
Sequencing
PCR
Lab Solutions
Allergy Diagnostics
See all applications and techniques
Services
Instrument Services
Cell Biology Services
Custom Services
Training Services
Lab Informatics Services
Financial and Leasing Services
Partnering and Licensing Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
See all services
Help and Support
How to Order
Promotions and Online Offers
Contact Us
Change Location
Create a New Account
See all help and support topics
Popular
Our Instagram
Our Facebook
Blog Behind the Bench
Customer Experience Center (CEC)
Ecommerce Exclusives
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Special Offers
Contact Us
Quick Order
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Contact Us
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
Gene:
TNFSF4
Transcript:
NM_003326.4
Assay Name:
Mutation genome location:
chr.1 173186069-173186578 on build GRCh38
Assay transcription exon location:
Within Exon 3
Assay ID
Hs00295493_CE
Purification
DS
HPLC
Size
25 nmol
25 nmol
50 nmol
50 nmol
25 nmol
25 nmol
50 nmol
50 nmol
25 nmol
25 nmol
50 nmol
50 nmol
25 nmol
25 nmol
50 nmol
50 nmol
Availability
Made To Order
Catalog #
A15629
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
Assay Details
More Information
Genomic Map
LOADING...
×
Back To Top
Assay Details
Back To Top
More Information
Set Membership:
Between 400-700 bp
Gene Ontology Categories:
Function(s)
Process(es)
defense response to nematode
acute inflammatory response
positive regulation of T cell cytokine production
regulation of adaptive immune response
positive regulation of type 2 immune response
positive regulation of immunoglobulin mediated immune response
immune response
signal transduction
cholesterol metabolic process
positive regulation of cell proliferation
response to virus
negative regulation of interferon-gamma production
negative regulation of interleukin-17 production
positive regulation of interferon-gamma production
positive regulation of interleukin-10 production
positive regulation of interleukin-12 production
positive regulation of interleukin-13 production
positive regulation of interleukin-2 production
positive regulation of interleukin-4 production
positive regulation of interleukin-6 production
tumor necrosis factor-mediated signaling pathway
memory T cell activation
T-helper 2 cell activation
response to nitrogen dioxide
cellular response to nitrogen dioxide
CD4-positive, alpha-beta T cell costimulation
T cell proliferation
positive regulation of T cell proliferation
positive regulation of activated T cell proliferation
positive regulation of CD4-positive, alpha-beta T cell differentiation
positive regulation of memory T cell differentiation
negative regulation of sequence-specific DNA binding transcription factor activity
negative regulation of regulatory T cell differentiation
negative regulation of T-helper 1 cell differentiation
positive regulation of T-helper 2 cell differentiation
negative regulation of transcription, DNA-templated
positive regulation of alpha-beta T cell proliferation
negative regulation of cytokine secretion
regulation of inflammatory response
positive regulation of inflammatory response
positive regulation of B cell activation
positive regulation of immunoglobulin secretion
cellular response to lipopolysaccharide
cellular response to prostaglandin E stimulus
chemokine (C-C motif) ligand 11 production
positive regulation of CD4-positive, alpha-beta T cell costimulation
positive regulation of memory T cell activation
positive regulation of T-helper 2 cell activation
positive regulation of interleukin-4-dependent isotype switching to IgE isotypes
receptor binding
cytokine activity
tumor necrosis factor receptor binding
tumor necrosis factor receptor superfamily binding
Back To Top
Related Products
POP-6
™
Polymer
POP-7
™
Polymer
BigDye
®
Terminator v1.1 Cycle Sequencing Kit
BigDye
®
Terminator v3.1 Cycle Sequencing Kit
BigDye
®
Direct Cycle Sequencing Kit
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-b49b87d85-8dsbr:80/100.66.76.150:80.
git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.47.0-Offline