Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Clinical Genomics
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
Gene:
TEK
Transcript:
NM_000459.4
Assay Name:
Mutation genome location:
chr.9 27173029-27173568 on build GRCh38
Assay ID
Hs00300059_CE
Purification
DS
HPLC
Size
25 nmol
25 nmol
50 nmol
50 nmol
25 nmol
25 nmol
50 nmol
50 nmol
25 nmol
25 nmol
50 nmol
50 nmol
25 nmol
25 nmol
50 nmol
50 nmol
Availability
Made To Order
Catalog #
A15629
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
Assay Details
More Information
Genomic Map
LOADING...
×
Back To Top
Assay Details
Back To Top
More Information
Set Membership:
Between 400-700 bp
Panther Classification:
Molecular Function -
transmembrane signal receptor
Gene Ontology Categories:
Function(s)
Process(es)
MAPK cascade
angiogenesis
response to hypoxia
positive regulation of protein phosphorylation
endothelial cell proliferation
positive regulation of endothelial cell proliferation
endochondral ossification
sprouting angiogenesis
signal transduction
transmembrane receptor protein tyrosine kinase signaling pathway
cell-cell signaling
heart development
positive regulation of endothelial cell migration
positive regulation of phosphatidylinositol 3-kinase signaling
negative regulation of angiogenesis
peptidyl-tyrosine phosphorylation
organ regeneration
regulation of establishment or maintenance of cell polarity
substrate adhesion-dependent cell spreading
negative regulation of apoptotic process
regulation of vascular permeability
response to peptide hormone
positive regulation of GTPase activity
positive regulation of phosphatidylinositol 3-kinase activity
response to estrogen
positive regulation of angiogenesis
protein autophosphorylation
Tie signaling pathway
negative regulation of inflammatory response
leukocyte migration
protein oligomerization
response to cAMP
positive regulation of focal adhesion assembly
positive regulation of protein kinase B signaling
definitive hemopoiesis
heart trabecula formation
positive regulation of ERK1 and ERK2 cascade
positive regulation of intracellular signal transduction
positive regulation of actin cytoskeleton reorganization
regulation of endothelial cell apoptotic process
negative regulation of endothelial cell apoptotic process
protein kinase activity
protein tyrosine kinase activity
transmembrane receptor protein tyrosine kinase activity
receptor activity
Ras guanyl-nucleotide exchange factor activity
protein binding
ATP binding
growth factor binding
Back To Top
Related Products
POP-6
™
Polymer
POP-7
™
Polymer
BigDye
®
Terminator v1.1 Cycle Sequencing Kit
BigDye
®
Terminator v3.1 Cycle Sequencing Kit
BigDye
®
Direct Cycle Sequencing Kit
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-b49b87d85-n9r8p:80/100.66.76.150:80.
git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.47.0-Offline