Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture and Transfection
Chemicals
Chromatography
Electron Microscopes
Lab Plasticware and Supplies
Lab Centrifuges
Lab Solutions
Mass Spectrometers
Next Generation Sequencers
See all product categories
Applications
Brands
Industrial and Applied Sciences
Food and Beverage
Forensics
Lab Solutions
Life Sciences
Pharma and Biopharma
Biotechnology
Clinical and Diagnostics
Digital Solutions
See all applications
Services
Lab Instrument and Equipment Services
Custom Services
Training Services
Financial and Leasing Services
Enterprise Level Lab Informatics
Partnering and Licensing Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Cell Biology Services
Food Safety Inspection Services
See all services
Help and Support
Contact Us
Product Documentation
Knowledge Base and Product FAQs
Learning Centers
Supply Center
eProcurement Solutions
Lab Instrument and Equipment Support
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Special Offers
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
Gene:
RGN
Transcript:
Assay Name:
Mutation genome location:
chr.X 47091974-47092489 on build GRCh38
Assay ID
Hs00305160_CE
Purification
DS
HPLC
Size
25 nmol
25 nmol
50 nmol
50 nmol
25 nmol
25 nmol
50 nmol
50 nmol
25 nmol
25 nmol
50 nmol
50 nmol
25 nmol
25 nmol
50 nmol
50 nmol
Availability
Made To Order
Catalog #
A15629
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
Assay Details
More Information
Genomic Map
LOADING...
×
Back To Top
Assay Details
Back To Top
More Information
Set Membership:
Between 400-700 bp
Panther Classification:
Molecular Function -
esterase
Gene Ontology Categories:
Function(s)
Process(es)
kidney development
negative regulation of protein kinase activity
cellular calcium ion homeostasis
spermatogenesis
aging
positive regulation of triglyceride biosynthetic process
positive regulation of glucose metabolic process
positive regulation of phosphatase activity
L-ascorbic acid biosynthetic process
negative regulation of phosphoprotein phosphatase activity
positive regulation of ATPase activity
negative regulation of GTPase activity
negative regulation of apoptotic process
positive regulation of GTPase activity
negative regulation of nitric oxide biosynthetic process
positive regulation of fatty acid biosynthetic process
negative regulation of epithelial cell proliferation
regulation of calcium-mediated signaling
negative regulation of cyclic-nucleotide phosphodiesterase activity
liver regeneration
negative regulation of sperm motility
positive regulation of superoxide dismutase activity
positive regulation of calcium-transporting ATPase activity
negative regulation of RNA biosynthetic process
negative regulation of bone development
positive regulation of proteolysis involved in cellular protein catabolic process
negative regulation of calcium-dependent ATPase activity
negative regulation of DNA catabolic process
positive regulation of dUTP diphosphatase activity
negative regulation of leucine-tRNA ligase activity
negative regulation of DNA biosynthetic process
gluconolactonase activity
calcium ion binding
zinc ion binding
enzyme regulator activity
Back To Top
Related Products
POP-6
™
Polymer
POP-7
™
Polymer
BigDye
®
Terminator v1.1 Cycle Sequencing Kit
BigDye
®
Terminator v3.1 Cycle Sequencing Kit
BigDye
®
Direct Cycle Sequencing Kit
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-b49b87d85-n9r8p:80/100.66.76.150:80.
git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.47.0-Offline