Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Cell Analysis
Antibodies
Mass Spectrometry
Cell Culture
Laboratory Instruments
Clinical and Diagnostics
Chromatography
Laboratory Equipment
Laboratory Supplies
Molecular Biology and Nucleic Acid Analysis
Sequence-Specific Nucleic Acid Products
See all product categories
Applications
Cell Culture and Transfection
Flow Cytometry
Cancer Research
Chromatography
Sequencing
PCR
Lab Solutions
Allergy Diagnostics
See all applications and techniques
Services
Instrument Services
Cell Biology Services
Custom Services
Training Services
Enterprise Services
Lab Informatics Services
Financial and Leasing Services
Partnering and Licensing Services
CDMO and Clinical Trial Services
See all services
Help and Support
How to Order
Promotions and Online Offers
Contact Us
Change Location
Create a New Account
See all help and support topics
Popular
Our Instagram
Our Facebook
Blog Behind the Bench
Customer Experience Center (CEC)
Ecommerce Exclusives
Contact Us
Quick Order
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Contact Us
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
Gene Symbol:
AVPR1B
Entrez Gene ID
553
Gene Name:
arginine vasopressin receptor 1B
Gene Aliases
AVPR3
Gene Chromosome Location
Chr.1: on Build GRCh38
UniGene
Hs.1372
Species:
Human
Primer Chromosome Location
Chr.1: 206116765-206116261 on Build GRCh38
View in UCSC browser
Amplicon Length
505
Primer Sequences
Log in to view
Ampliseq ID's
Primer ID
Hs00312003_CE
Primer 5'-tail Option
Non-tailed
M13-tailed
Purification
Desalted
HPLC
Primer Pair /FWD/REV
Pair
Forward
Reverse
Synthesis Scale
25 nmol
Availability
Made To Order
Catalog #'s
A15629,A15630
Price
Add To Cart
Genomic Map
Assay Details
More Information
Genomic Map
LOADING...
×
Back To Top
Assay Details
Gene Information
AVPR1B
Accession
Protein
COSMIC ID
Mutation
AA
RefSeq
NM_000707.3
NP_000698.1
50331
c.386C>T
p.T129M
4993666
c.572_573insCGG
p.G191_F192insG
2215744
c.512delT
p.S173fs*5
3803639
c.208C>T
p.H70Y
2215741
c.455T>A
p.L152Q
902897
c.268C>T
p.P90S
3705429
c.394C>T
p.R132C
4664795
c.559T>C
p.W187R
2215738
c.411T>C
p.C137C
369759
c.202C>A
p.R68S
4652121
c.561G>T
p.W187C
1748112
c.224A>G
p.H75R
1683820
c.517delT
p.S173fs*5
4611707
c.414delC
p.L140fs*38
1668411
c.259C>A
p.Q87K
4576809
c.242T>C
p.L81P
7262069
c.196C>T
p.R66C
5825836
c.511_512insG
p.I171fs*55
4551313
c.525G>A
p.R175R
5735813
c.202C>T
p.R68C
3482370
c.366C>T
p.T122T
3482368
c.200C>T
p.S67F
7085164
c.195G>C
p.K65N
4488126
c.327C>T
p.A109A
3418666
c.508T>C
p.F170L
1235636
c.554A>G
p.D185G
230895
c.416_417CC>TT
p.P139>?
1127172
c.544G>T
p.G182W
4229979
c.331A>C
p.K111Q
6608977
c.247G>A
p.V83M
5352860
c.203G>A
p.R68H
6608975
c.421C>T
p.R141C
4143266
c.387G>T
p.T129T
3976903
c.365C>A
p.T122N
4994034
c.568T>C
p.F190L
6608969
c.384G>T
p.M128I
2215745
c.518C>T
p.S173F
6508562
c.242T>A
p.L81Q
2215742
c.463G>A
p.A155T
4764448
c.395G>A
p.R132H
902898
c.570C>T
p.F190F
2215739
c.412C>T
p.H138Y
902896
c.216C>T
p.F72F
369760
c.394C>G
p.R132G
73793
c.563C>A
p.A188E
2215736
c.252G>A
p.A84A
5969103
c.337C>G
p.L113V
3689314
c.310G>A
p.D104N
7303402
c.307C>T
p.P103S
1718127
c.459C>T
p.L153L
5938573
c.592G>A
p.A198T
358321
c.389T>C
p.L130P
1668412
c.316C>A
p.L106M
3482371
c.519C>T
p.S173S
7208943
c.205A>G
p.M69V
3482369
c.234G>A
p.L78L
279091
c.624C>T
p.F208F
678944
c.502C>T
p.Q168*
1184274
c.326C>T
p.A109V
5469857
c.454C>A
p.L152M
117400
c.488T>A
p.I163N
6608978
c.484G>A
p.A162T
4229980
c.552G>A
p.L184L
4229978
c.328G>A
p.V110I
6608976
c.217G>A
p.V73M
GenBank
AF030512.1
AAB84293.1
-
-
-
AF101726.1
AAD17892.1
-
-
-
D31833.1
BAA06621.1
-
-
-
DY317849.1
-
-
-
ENST00000367126.0
-
-
-
EU432111.1
-
-
-
L37112.1
AAA65687.1
-
-
-
Phenotype
MIM: 600264
Literature Links:
AVPR1B
PubMed Links
SNP ID
Back To Top
More Information
Set Membership:
Between 400-700 bp
Panther Classification:
Molecular Function -
G-protein coupled receptor
Gene Ontology Categories:
Function(s)
Process(es)
regulation of systemic arterial blood pressure by vasopressin
G-protein coupled receptor signaling pathway
activation of phospholipase C activity
positive regulation of cytosolic calcium ion concentration
cellular response to hormone stimulus
positive regulation of vasoconstriction
response to peptide
vasopressin receptor activity
protein kinase C binding
peptide binding
Format:
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer
™
Tool Quick Reference
Back To Top
Related Products
POP-6
™
Polymer
POP-7
™
Polymer
BigDye
®
Terminator v1.1 Cycle Sequencing Kit
BigDye
®
Terminator v3.1 Cycle Sequencing Kit
BigDye
®
Direct Cycle Sequencing Kit
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-74fd55b88b-zs8m7:80/10.239.167.245:80.
git-commit: e4d250c089da5f210ba2ef291b70bc15861d50fd
git-url: git@github.com:thermofisher/magellan-core.git
git-branch: origin/release/2.9.0-2024.05.21-1.0