Hamburger Menu Button
Thermo Fisher Scientific Logo
登入
Don't have an account ?
Create Account
Products
Antibodies
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Tubes
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Clinical Genomics
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO 和 CRO 服務
CDMO 服務
臨床試驗 CRO 服務
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
Register for an Account
How to Order
Instrument Support
Technical Support Centers
Learning Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
快速訂購
登入
登入
Don't have an account ?
Create Account
帳戶
訂單
訂製產品與專案
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
Gene:
KLLN
Transcript:
Assay Name:
Mutation genome location:
chr.10 87863232-87863720 on build GRCh38
Gene:
PTEN
Transcript:
Assay Name:
Mutation genome location:
chr.10 87863232-87863720 on build GRCh38
Assay ID
Hs00320882_CE
Purification
DS
HPLC
Size
25 nmol
25 nmol
50 nmol
50 nmol
25 nmol
25 nmol
50 nmol
50 nmol
25 nmol
25 nmol
50 nmol
50 nmol
25 nmol
25 nmol
50 nmol
50 nmol
Availability
Made To Order
Catalog #
A15629
Price
Your Price
Online offer:
Check your price ›
新增到購物車
Genomic Map
Assay Details
More Information
Genomic Map
LOADING...
×
Back To Top
Assay Details
Back To Top
More Information
Set Membership:
Between 400-700 bp
Panther Classification:
Molecular Function -
protein phosphatase
Gene Ontology Categories:
Function(s)
Process(es)
apoptotic process
cell cycle arrest
regulation of cyclin-dependent protein serine/threonine kinase activity
angiogenesis
negative regulation of protein phosphorylation
regulation of B cell apoptotic process
protein dephosphorylation
phosphatidylinositol biosynthetic process
neuron-neuron synaptic transmission
synapse assembly
central nervous system development
heart development
aging
response to nutrient
learning or memory
memory
locomotory behavior
cell proliferation
positive regulation of cell proliferation
negative regulation of cell proliferation
response to glucose
response to zinc ion
regulation of neuron projection development
negative regulation of phosphatidylinositol 3-kinase signaling
cell migration
dentate gyrus development
central nervous system neuron axonogenesis
negative regulation of cell migration
adult behavior
negative regulation of myelination
regulation of protein stability
negative regulation of cyclin-dependent protein serine/threonine kinase activity involved in G1/S transition of mitotic cell cycle
regulation of synaptic transmission, GABAergic
central nervous system myelin maintenance
response to estradiol
regulation of cellular component size
regulation of myeloid cell apoptotic process
response to ATP
multicellular organismal response to stress
social behavior
peptidyl-tyrosine dephosphorylation
response to drug
maternal behavior
negative regulation of apoptotic process
protein kinase B signaling
endothelial cell migration
inositol phosphate metabolic process
response to ethanol
locomotor rhythm
negative regulation of cell size
negative regulation of organ growth
response to arsenic-containing substance
inositol phosphate dephosphorylation
phosphatidylinositol dephosphorylation
platelet-derived growth factor receptor signaling pathway
phosphatidylinositol-mediated signaling
regulation of axon regeneration
cardiac muscle tissue development
forebrain morphogenesis
brain morphogenesis
negative regulation of epithelial cell proliferation
negative regulation of phagocytosis
negative regulation of axonogenesis
protein stabilization
T cell receptor signaling pathway
positive regulation of sequence-specific DNA binding transcription factor activity
negative regulation of focal adhesion assembly
negative regulation of protein kinase B signaling
rhythmic synaptic transmission
canonical Wnt signaling pathway
synapse maturation
prepulse inhibition
male mating behavior
long-term synaptic potentiation
long term synaptic depression
prostate gland growth
dendritic spine morphogenesis
negative regulation of dendritic spine morphogenesis
negative regulation of ERK1 and ERK2 cascade
positive regulation of ERK1 and ERK2 cascade
cellular response to hypoxia
negative regulation of ribosome biogenesis
negative regulation of cell aging
negative regulation of excitatory postsynaptic potential
presynaptic membrane assembly
postsynaptic density assembly
positive regulation of TRAIL-activated apoptotic signaling pathway
positive regulation of ubiquitin protein ligase activity
positive regulation of protein ubiquitination involved in ubiquitin-dependent protein catabolic process
negative regulation of G1/S transition of mitotic cell cycle
positive regulation of excitatory postsynaptic potential
negative regulation of synaptic vesicle clustering
DNA binding
magnesium ion binding
phosphatidylinositol-3-phosphatase activity
phosphoprotein phosphatase activity
protein serine/threonine phosphatase activity
protein tyrosine phosphatase activity
platelet-derived growth factor receptor binding
protein binding
protein tyrosine/serine/threonine phosphatase activity
lipid binding
anaphase-promoting complex binding
phosphatidylinositol-3,4,5-trisphosphate 3-phosphatase activity
enzyme binding
protein kinase binding
PDZ domain binding
identical protein binding
inositol-1,3,4,5-tetrakisphosphate 3-phosphatase activity
phosphatidylinositol-3,4-bisphosphate 3-phosphatase activity
Back To Top
Related Products
POP-6
™
Polymer
POP-7
™
Polymer
BigDye
®
Terminator v1.1 Cycle Sequencing Kit
BigDye
®
Terminator v3.1 Cycle Sequencing Kit
BigDye
®
Direct Cycle Sequencing Kit
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-67655d8755-x75fr:80/100.66.77.4:80.
git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.46.0-Offline